Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 197459695-197460203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COQ10B
Entrez Gene ID
80219
Gene Name
coenzyme Q10B
Gene Aliases
-
UniGene
Hs.606592
Gene Chromosome Location
Chr.2: 197453423-197475309 on Build GRCh38
Transcripts
NM_001320818.1 NM_001320819.1 NM_001320820.1 NM_025147.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6307807 c.121G>T p.G41C Chr.2: 197459948-197459948 on Build GRCh38
7076921 c.201C>T p.I67I Chr.2: 197460028-197460028 on Build GRCh38
6156127 c.202A>G p.T68A Chr.2: 197460029-197460029 on Build GRCh38
260260 c.242G>T p.R81I Chr.2: 197460069-197460069 on Build GRCh38
7204512 c.249_251delTTT p.L84delL Chr.2: 197460076-197460078 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375172756 Chr.2: 197460047-197460047 on Build GRCh38
rs765425646 Chr.2: 197459946-197459946 on Build GRCh38
rs779647756 Chr.2: 197459980-197459980 on Build GRCh38
rs145175790 Chr.2: 197460038-197460038 on Build GRCh38
rs754847466 Chr.2: 197460156-197460156 on Build GRCh38
rs576394326 Chr.2: 197459748-197459749 on Build GRCh38
rs746827856 Chr.2: 197460099-197460099 on Build GRCh38
rs760695787 Chr.2: 197460009-197460009 on Build GRCh38
rs571716881 Chr.2: 197460133-197460133 on Build GRCh38
rs770746808 Chr.2: 197460101-197460101 on Build GRCh38
rs964078892 Chr.2: 197459760-197459761 on Build GRCh38
rs79653400 Chr.2: 197459750-197459750 on Build GRCh38
rs200568950 Chr.2: 197460107-197460107 on Build GRCh38
rs376551176 Chr.2: 197459762-197459762 on Build GRCh38
rs755051832 Chr.2: 197459943-197459943 on Build GRCh38
rs34946819 Chr.2: 197459969-197459969 on Build GRCh38
rs750932058 Chr.2: 197460084-197460084 on Build GRCh38
rs895236731 Chr.2: 197459836-197459836 on Build GRCh38
rs182337005 Chr.2: 197460085-197460085 on Build GRCh38
rs187709462 Chr.2: 197460136-197460136 on Build GRCh38
rs778218802 Chr.2: 197459973-197459973 on Build GRCh38
rs750045220 Chr.2: 197459980-197459981 on Build GRCh38
rs768193206 Chr.2: 197459987-197459987 on Build GRCh38
rs770008465 Chr.2: 197460113-197460113 on Build GRCh38
rs764577127 Chr.2: 197460043-197460043 on Build GRCh38
rs1031488734 Chr.2: 197459742-197459743 on Build GRCh38
rs1037312955 Chr.2: 197459751-197459751 on Build GRCh38
rs954521463 Chr.2: 197459749-197459749 on Build GRCh38
rs747533671 Chr.2: 197459976-197459976 on Build GRCh38
rs536087990 Chr.2: 197459949-197459949 on Build GRCh38
rs760845886 Chr.2: 197459833-197459833 on Build GRCh38
rs752973192 Chr.2: 197460034-197460034 on Build GRCh38
rs757612479 Chr.2: 197460051-197460051 on Build GRCh38
rs141523172 Chr.2: 197460140-197460140 on Build GRCh38
rs747818512 Chr.2: 197460088-197460088 on Build GRCh38
rs982849377 Chr.2: 197459913-197459913 on Build GRCh38
rs781603123 Chr.2: 197460079-197460079 on Build GRCh38
rs1048394661 Chr.2: 197459799-197459799 on Build GRCh38
rs944194956 Chr.2: 197459748-197459748 on Build GRCh38
rs774678315 Chr.2: 197459911-197459911 on Build GRCh38
rs919931622 Chr.2: 197459853-197459857 on Build GRCh38
rs772939216 Chr.2: 197460008-197460008 on Build GRCh38
rs748792617 Chr.2: 197459983-197459983 on Build GRCh38
rs778599636 Chr.2: 197459994-197459994 on Build GRCh38
rs753820047 Chr.2: 197459917-197459917 on Build GRCh38
rs777372542 Chr.2: 197460097-197460097 on Build GRCh38
rs574348449 Chr.2: 197460131-197460131 on Build GRCh38
rs372019133 Chr.2: 197460130-197460130 on Build GRCh38
rs747728380 Chr.2: 197460004-197460004 on Build GRCh38
rs766499967 Chr.2: 197459902-197459902 on Build GRCh38
rs567723973 Chr.2: 197459842-197459842 on Build GRCh38
rs368302397 Chr.2: 197460124-197460124 on Build GRCh38
rs113527285 Chr.2: 197460170-197460170 on Build GRCh38
rs778331984 Chr.2: 197460087-197460087 on Build GRCh38
rs199765901 Chr.2: 197460106-197460106 on Build GRCh38
rs1012379174 Chr.2: 197459837-197459837 on Build GRCh38
rs770811878 Chr.2: 197460011-197460011 on Build GRCh38
rs759592148 Chr.2: 197460024-197460024 on Build GRCh38
rs766262666 Chr.2: 197460100-197460101 on Build GRCh38
rs757658883 Chr.2: 197459977-197459977 on Build GRCh38
rs776608625 Chr.2: 197460012-197460012 on Build GRCh38
rs1003102370 Chr.2: 197459984-197459984 on Build GRCh38
rs371673029 Chr.2: 197460092-197460092 on Build GRCh38
rs995536648 Chr.2: 197459795-197459795 on Build GRCh38
rs938803665 Chr.2: 197459972-197459972 on Build GRCh38
rs897379328 Chr.2: 197459760-197459760 on Build GRCh38
rs759452766 Chr.2: 197460110-197460110 on Build GRCh38
rs764161730 Chr.2: 197460025-197460025 on Build GRCh38
rs771865816 Chr.2: 197460005-197460005 on Build GRCh38
rs1025057092 Chr.2: 197459888-197459888 on Build GRCh38
rs542883919 Chr.2: 197460150-197460150 on Build GRCh38
rs758208281 Chr.2: 197460095-197460108 on Build GRCh38
rs200588862 Chr.2: 197459954-197459954 on Build GRCh38
rs975856976 Chr.2: 197459819-197459819 on Build GRCh38
rs374263983 Chr.2: 197459827-197459828 on Build GRCh38
rs952803873 Chr.2: 197459908-197459908 on Build GRCh38
rs150890135 Chr.2: 197460141-197460141 on Build GRCh38
rs1048676949 Chr.2: 197460149-197460149 on Build GRCh38

View Full Product Details