Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 197727757-197728259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BOLL
Entrez Gene ID
66037
Gene Name
boule homolog, RNA binding protein
Gene Aliases
BOULE
UniGene
Hs.169797
Gene Chromosome Location
Chr.2: 197726879-197786574 on Build GRCh38
Transcripts
NM_001284358.1 NM_001284361.1 NM_001284362.1 NM_033030.5 NM_197970.2 XM_006712715.3 XM_011511692.2 XM_011511693.2 XM_011511694.2

SNP Detail

SNP ID Chromosome Location
rs916980061 Chr.2: 197728005-197728005 on Build GRCh38
rs900316924 Chr.2: 197728027-197728027 on Build GRCh38
rs767772965 Chr.2: 197728072-197728072 on Build GRCh38
rs998792442 Chr.2: 197727806-197727806 on Build GRCh38
rs998344755 Chr.2: 197728125-197728125 on Build GRCh38
rs759819468 Chr.2: 197728069-197728069 on Build GRCh38
rs960242356 Chr.2: 197727783-197727783 on Build GRCh38
rs994669162 Chr.2: 197728032-197728032 on Build GRCh38
rs184065998 Chr.2: 197728009-197728009 on Build GRCh38
rs927087730 Chr.2: 197727944-197727944 on Build GRCh38
rs774765343 Chr.2: 197727980-197727980 on Build GRCh38
rs539310339 Chr.2: 197728083-197728083 on Build GRCh38
rs1039947940 Chr.2: 197728105-197728105 on Build GRCh38
rs963604397 Chr.2: 197727918-197727918 on Build GRCh38
rs1041340240 Chr.2: 197728054-197728054 on Build GRCh38
rs775148827 Chr.2: 197728087-197728089 on Build GRCh38
rs1004201411 Chr.2: 197727847-197727847 on Build GRCh38
rs993079638 Chr.2: 197727792-197727792 on Build GRCh38
rs1016960948 Chr.2: 197727859-197727859 on Build GRCh38
rs148574461 Chr.2: 197727996-197727996 on Build GRCh38
rs958557927 Chr.2: 197727981-197727981 on Build GRCh38
rs1029747833 Chr.2: 197727809-197727809 on Build GRCh38
rs532718791 Chr.2: 197728217-197728217 on Build GRCh38
rs951465139 Chr.2: 197727827-197727827 on Build GRCh38
rs887163265 Chr.2: 197728214-197728214 on Build GRCh38
rs1051292540 Chr.2: 197728204-197728204 on Build GRCh38
rs1027926725 Chr.2: 197728067-197728067 on Build GRCh38
rs992659907 Chr.2: 197727998-197727998 on Build GRCh38
rs900006200 Chr.2: 197727799-197727799 on Build GRCh38
rs1040237121 Chr.2: 197727786-197727786 on Build GRCh38
rs76873639 Chr.2: 197727782-197727782 on Build GRCh38
rs890128724 Chr.2: 197728003-197728003 on Build GRCh38
rs572654913 Chr.2: 197727922-197727922 on Build GRCh38
rs562157667 Chr.2: 197728124-197728126 on Build GRCh38
rs900120595 Chr.2: 197728113-197728113 on Build GRCh38
rs370062869 Chr.2: 197727997-197727997 on Build GRCh38
rs973348624 Chr.2: 197727920-197727920 on Build GRCh38
rs916056142 Chr.2: 197727804-197727804 on Build GRCh38

View Full Product Details