Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 199911489-199912023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf69
Entrez Gene ID
205327
Gene Name
chromosome 2 open reading frame 69
Gene Aliases
-
UniGene
Hs.154494
Gene Chromosome Location
Chr.2: 199911256-199928273 on Build GRCh38
Transcripts
NM_153689.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6204402 c.70G>T p.A24S Chr.2: 199911508-199911508 on Build GRCh38
399089 c.177C>A p.T59T Chr.2: 199911615-199911615 on Build GRCh38
3749627 c.192T>G p.D64E Chr.2: 199911630-199911630 on Build GRCh38
5978490 c.199C>T p.R67C Chr.2: 199911637-199911637 on Build GRCh38
5978489 c.107G>C p.G36A Chr.2: 199911545-199911545 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777904112 Chr.2: 199911591-199911591 on Build GRCh38
rs1008768652 Chr.2: 199911978-199911978 on Build GRCh38
rs548799391 Chr.2: 199911667-199911667 on Build GRCh38
rs751776602 Chr.2: 199911688-199911688 on Build GRCh38
rs762288716 Chr.2: 199911593-199911593 on Build GRCh38
rs1042359793 Chr.2: 199911845-199911845 on Build GRCh38
rs747711433 Chr.2: 199911785-199911785 on Build GRCh38
rs563083229 Chr.2: 199911633-199911633 on Build GRCh38
rs926493922 Chr.2: 199911706-199911706 on Build GRCh38
rs996936635 Chr.2: 199911607-199911607 on Build GRCh38
rs866052049 Chr.2: 199911626-199911626 on Build GRCh38
rs751455395 Chr.2: 199911893-199911893 on Build GRCh38
rs995885515 Chr.2: 199911933-199911933 on Build GRCh38
rs777065957 Chr.2: 199911509-199911509 on Build GRCh38
rs746636264 Chr.2: 199911806-199911806 on Build GRCh38
rs756678563 Chr.2: 199911778-199911778 on Build GRCh38
rs540650009 Chr.2: 199911545-199911545 on Build GRCh38
rs937498852 Chr.2: 199911615-199911615 on Build GRCh38
rs999584286 Chr.2: 199911891-199911891 on Build GRCh38
rs961961215 Chr.2: 199911997-199911997 on Build GRCh38
rs950280575 Chr.2: 199911831-199911831 on Build GRCh38
rs973748456 Chr.2: 199912002-199912002 on Build GRCh38
rs755050592 Chr.2: 199911570-199911570 on Build GRCh38
rs757604720 Chr.2: 199911616-199911616 on Build GRCh38
rs767488289 Chr.2: 199911983-199911983 on Build GRCh38
rs762310324 Chr.2: 199911790-199911790 on Build GRCh38
rs1056419384 Chr.2: 199911776-199911776 on Build GRCh38
rs546592325 Chr.2: 199911788-199911788 on Build GRCh38
rs954691185 Chr.2: 199911960-199911960 on Build GRCh38
rs752827106 Chr.2: 199911676-199911676 on Build GRCh38
rs767030169 Chr.2: 199911527-199911527 on Build GRCh38
rs370597918 Chr.2: 199911780-199911780 on Build GRCh38
rs568798827 Chr.2: 199911913-199911913 on Build GRCh38
rs760609777 Chr.2: 199911639-199911639 on Build GRCh38
rs1025039325 Chr.2: 199911512-199911512 on Build GRCh38
rs762882770 Chr.2: 199911684-199911684 on Build GRCh38
rs996572191 Chr.2: 199911848-199911848 on Build GRCh38
rs746356835 Chr.2: 199911628-199911628 on Build GRCh38
rs753818494 Chr.2: 199911569-199911569 on Build GRCh38
rs571126218 Chr.2: 199911794-199911794 on Build GRCh38
rs903458192 Chr.2: 199911856-199911856 on Build GRCh38
rs781715546 Chr.2: 199911617-199911617 on Build GRCh38
rs987757316 Chr.2: 199911726-199911726 on Build GRCh38
rs937405368 Chr.2: 199911804-199911804 on Build GRCh38
rs773176679 Chr.2: 199911541-199911541 on Build GRCh38
rs1028429885 Chr.2: 199911867-199911867 on Build GRCh38
rs973459800 Chr.2: 199911791-199911791 on Build GRCh38
rs536354517 Chr.2: 199911516-199911516 on Build GRCh38
rs572997218 Chr.2: 199911526-199911526 on Build GRCh38
rs764359748 Chr.2: 199911685-199911685 on Build GRCh38
rs974034199 Chr.2: 199912001-199912001 on Build GRCh38
rs1028805258 Chr.2: 199911699-199911699 on Build GRCh38
rs747576112 Chr.2: 199911631-199911631 on Build GRCh38
rs1020542975 Chr.2: 199911632-199911632 on Build GRCh38
rs538925979 Chr.2: 199911808-199911808 on Build GRCh38
rs187090652 Chr.2: 199911654-199911654 on Build GRCh38
rs781516621 Chr.2: 199911720-199911720 on Build GRCh38
rs766243890 Chr.2: 199911648-199911648 on Build GRCh38
rs940706762 Chr.2: 199911595-199911595 on Build GRCh38
rs139155183 Chr.2: 199911932-199911932 on Build GRCh38
rs961932669 Chr.2: 199911940-199911940 on Build GRCh38
rs114873734 Chr.2: 199911822-199911822 on Build GRCh38
rs1009726408 Chr.2: 199911566-199911566 on Build GRCh38
rs1053419303 Chr.2: 199911905-199911905 on Build GRCh38
rs554682984 Chr.2: 199911519-199911519 on Build GRCh38
rs759538791 Chr.2: 199911662-199911662 on Build GRCh38
rs560995925 Chr.2: 199911670-199911670 on Build GRCh38
rs892160938 Chr.2: 199911558-199911558 on Build GRCh38
rs1009197862 Chr.2: 199911825-199911825 on Build GRCh38
rs62178345 Chr.2: 199911630-199911630 on Build GRCh38
rs899281796 Chr.2: 199911613-199911613 on Build GRCh38
rs527992503 Chr.2: 199911943-199911943 on Build GRCh38
rs565435469 Chr.2: 199911552-199911552 on Build GRCh38
rs747388975 Chr.2: 199911611-199911611 on Build GRCh38
rs974597838 Chr.2: 199911543-199911543 on Build GRCh38
rs907817389 Chr.2: 199911740-199911740 on Build GRCh38
rs757661577 Chr.2: 199911710-199911710 on Build GRCh38
rs149905387 Chr.2: 199911999-199911999 on Build GRCh38
rs759369705 Chr.2: 199911778-199911779 on Build GRCh38
rs772982593 Chr.2: 199911637-199911637 on Build GRCh38
rs759433534 Chr.2: 199911974-199911974 on Build GRCh38
rs1022042628 Chr.2: 199911577-199911577 on Build GRCh38
rs766478215 Chr.2: 199911556-199911556 on Build GRCh38
rs777604598 Chr.2: 199911801-199911801 on Build GRCh38
rs758678127 Chr.2: 199911589-199911589 on Build GRCh38
rs1028752874 Chr.2: 199911945-199911945 on Build GRCh38
rs1015886989 Chr.2: 199911919-199911919 on Build GRCh38
rs752986093 Chr.2: 199911888-199911888 on Build GRCh38
rs868667563 Chr.2: 199911586-199911586 on Build GRCh38
rs748855175 Chr.2: 199911515-199911515 on Build GRCh38
rs1032126923 Chr.2: 199911521-199911521 on Build GRCh38
rs528160849 Chr.2: 199911750-199911750 on Build GRCh38
rs577488855 Chr.2: 199911588-199911588 on Build GRCh38
rs1055809344 Chr.2: 199911819-199911819 on Build GRCh38
rs780363295 Chr.2: 199911781-199911781 on Build GRCh38

View Full Product Details