Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 199927123-199927642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf69
Entrez Gene ID
205327
Gene Name
chromosome 2 open reading frame 69
Gene Aliases
-
UniGene
Hs.154494
Gene Chromosome Location
Chr.2: 199911256-199928273 on Build GRCh38
Transcripts
NM_153689.5

SNP Detail

SNP ID Chromosome Location
rs569314988 Chr.2: 199927214-199927214 on Build GRCh38
rs777421530 Chr.2: 199927255-199927256 on Build GRCh38
rs774281957 Chr.2: 199927408-199927408 on Build GRCh38
rs935096149 Chr.2: 199927330-199927330 on Build GRCh38
rs972746190 Chr.2: 199927564-199927564 on Build GRCh38
rs954276375 Chr.2: 199927145-199927145 on Build GRCh38
rs1023301535 Chr.2: 199927309-199927309 on Build GRCh38
rs1009394167 Chr.2: 199927160-199927160 on Build GRCh38
rs57970244 Chr.2: 199927328-199927329 on Build GRCh38
rs781535283 Chr.2: 199927471-199927471 on Build GRCh38
rs144383882 Chr.2: 199927596-199927596 on Build GRCh38
rs1036790315 Chr.2: 199927416-199927416 on Build GRCh38
rs887572086 Chr.2: 199927320-199927321 on Build GRCh38
rs972071470 Chr.2: 199927598-199927598 on Build GRCh38
rs923688330 Chr.2: 199927324-199927324 on Build GRCh38
rs766237117 Chr.2: 199927401-199927401 on Build GRCh38
rs1032098533 Chr.2: 199927458-199927458 on Build GRCh38
rs200930815 Chr.2: 199927331-199927331 on Build GRCh38
rs386654147 Chr.2: 199927381-199927383 on Build GRCh38
rs75346425 Chr.2: 199927212-199927212 on Build GRCh38
rs897869265 Chr.2: 199927447-199927447 on Build GRCh38
rs994979687 Chr.2: 199927198-199927198 on Build GRCh38
rs747351434 Chr.2: 199927294-199927294 on Build GRCh38
rs79900471 Chr.2: 199927308-199927308 on Build GRCh38
rs369650425 Chr.2: 199927308-199927309 on Build GRCh38
rs931427640 Chr.2: 199927304-199927304 on Build GRCh38
rs958909036 Chr.2: 199927232-199927232 on Build GRCh38
rs937850041 Chr.2: 199927338-199927339 on Build GRCh38
rs76660606 Chr.2: 199927208-199927208 on Build GRCh38
rs912816591 Chr.2: 199927163-199927163 on Build GRCh38
rs1020659871 Chr.2: 199927390-199927390 on Build GRCh38
rs536286247 Chr.2: 199927230-199927230 on Build GRCh38
rs982257801 Chr.2: 199927317-199927317 on Build GRCh38
rs1003187987 Chr.2: 199927616-199927616 on Build GRCh38
rs78800944 Chr.2: 199927211-199927211 on Build GRCh38
rs985693338 Chr.2: 199927157-199927157 on Build GRCh38
rs915084840 Chr.2: 199927367-199927367 on Build GRCh38
rs970441152 Chr.2: 199927316-199927316 on Build GRCh38
rs1048878054 Chr.2: 199927309-199927310 on Build GRCh38
rs368954862 Chr.2: 199927461-199927461 on Build GRCh38
rs200243225 Chr.2: 199927412-199927412 on Build GRCh38
rs386355527 Chr.2: 199927309-199927310 on Build GRCh38
rs559204664 Chr.2: 199927525-199927525 on Build GRCh38
rs577554955 Chr.2: 199927550-199927550 on Build GRCh38
rs386392283 Chr.2: 199927327-199927328 on Build GRCh38
rs17591036 Chr.2: 199927381-199927381 on Build GRCh38
rs1019841129 Chr.2: 199927526-199927526 on Build GRCh38
rs966038133 Chr.2: 199927535-199927535 on Build GRCh38
rs1013154086 Chr.2: 199927591-199927591 on Build GRCh38
rs878966306 Chr.2: 199927566-199927566 on Build GRCh38
rs994929831 Chr.2: 199927516-199927516 on Build GRCh38
rs1049249707 Chr.2: 199927543-199927543 on Build GRCh38
rs530293509 Chr.2: 199927383-199927383 on Build GRCh38
rs59958580 Chr.2: 199927307-199927307 on Build GRCh38
rs989235691 Chr.2: 199927337-199927337 on Build GRCh38
rs1027827117 Chr.2: 199927215-199927215 on Build GRCh38
rs991773360 Chr.2: 199927250-199927250 on Build GRCh38
rs894684724 Chr.2: 199927580-199927580 on Build GRCh38
rs918654352 Chr.2: 199927263-199927263 on Build GRCh38
rs199756580 Chr.2: 199927565-199927566 on Build GRCh38
rs75583675 Chr.2: 199927191-199927191 on Build GRCh38
rs769379091 Chr.2: 199927189-199927189 on Build GRCh38

View Full Product Details