Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200603175-200603674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AOX1
Entrez Gene ID
316
Gene Name
aldehyde oxidase 1
Gene Aliases
AO, AOH1
UniGene
Hs.406238
Gene Chromosome Location
Chr.2: 200585952-200677064 on Build GRCh38
Transcripts
NM_001159.3 XM_011511062.1 XM_017003946.1 XM_017003947.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6601428 c.510C>T p.C170C Chr.2: 200603278-200603278 on Build GRCh38
6358187 c.552A>G p.G184G Chr.2: 200603320-200603320 on Build GRCh38
6842302 c.499-1G>A p.? Chr.2: 200603266-200603266 on Build GRCh38
4571785 c.538T>G p.C180G Chr.2: 200603306-200603306 on Build GRCh38
7209787 c.521A>G p.K174R Chr.2: 200603289-200603289 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35057419 Chr.2: 200603443-200603443 on Build GRCh38
rs753264884 Chr.2: 200603516-200603516 on Build GRCh38
rs938879925 Chr.2: 200603380-200603380 on Build GRCh38
rs759762591 Chr.2: 200603362-200603362 on Build GRCh38
rs1057283119 Chr.2: 200603411-200603411 on Build GRCh38
rs760794916 Chr.2: 200603350-200603350 on Build GRCh38
rs1042085122 Chr.2: 200603439-200603439 on Build GRCh38
rs549050082 Chr.2: 200603536-200603536 on Build GRCh38
rs917699850 Chr.2: 200603415-200603415 on Build GRCh38
rs746443599 Chr.2: 200603251-200603252 on Build GRCh38
rs765474588 Chr.2: 200603356-200603356 on Build GRCh38
rs773687673 Chr.2: 200603596-200603596 on Build GRCh38
rs112973027 Chr.2: 200603499-200603499 on Build GRCh38
rs779900372 Chr.2: 200603294-200603294 on Build GRCh38
rs963388816 Chr.2: 200603589-200603589 on Build GRCh38
rs201052951 Chr.2: 200603371-200603371 on Build GRCh38
rs748223634 Chr.2: 200603502-200603502 on Build GRCh38
rs773171778 Chr.2: 200603339-200603339 on Build GRCh38
rs374371168 Chr.2: 200603384-200603384 on Build GRCh38
rs950497749 Chr.2: 200603533-200603533 on Build GRCh38
rs374681400 Chr.2: 200603230-200603230 on Build GRCh38
rs552108524 Chr.2: 200603456-200603456 on Build GRCh38
rs542431016 Chr.2: 200603298-200603298 on Build GRCh38
rs753043755 Chr.2: 200603242-200603242 on Build GRCh38
rs1016334315 Chr.2: 200603567-200603567 on Build GRCh38
rs778585898 Chr.2: 200603319-200603319 on Build GRCh38
rs950811268 Chr.2: 200603202-200603202 on Build GRCh38
rs755652086 Chr.2: 200603285-200603285 on Build GRCh38
rs570559676 Chr.2: 200603513-200603513 on Build GRCh38
rs765519366 Chr.2: 200603240-200603240 on Build GRCh38
rs916703329 Chr.2: 200603633-200603633 on Build GRCh38
rs202026674 Chr.2: 200603503-200603504 on Build GRCh38
rs980368912 Chr.2: 200603312-200603312 on Build GRCh38
rs368656069 Chr.2: 200603271-200603271 on Build GRCh38
rs770987381 Chr.2: 200603352-200603352 on Build GRCh38
rs778290032 Chr.2: 200603248-200603248 on Build GRCh38
rs997781445 Chr.2: 200603519-200603519 on Build GRCh38
rs906812759 Chr.2: 200603353-200603353 on Build GRCh38
rs903529362 Chr.2: 200603454-200603454 on Build GRCh38
rs758838211 Chr.2: 200603244-200603244 on Build GRCh38
rs907887248 Chr.2: 200603593-200603593 on Build GRCh38
rs900403368 Chr.2: 200603509-200603509 on Build GRCh38
rs957742121 Chr.2: 200603396-200603396 on Build GRCh38
rs765438502 Chr.2: 200603368-200603368 on Build GRCh38
rs113393173 Chr.2: 200603520-200603520 on Build GRCh38
rs764547298 Chr.2: 200603397-200603397 on Build GRCh38
rs752088394 Chr.2: 200603404-200603404 on Build GRCh38
rs763308671 Chr.2: 200603621-200603621 on Build GRCh38
rs768639406 Chr.2: 200603300-200603300 on Build GRCh38
rs1018063448 Chr.2: 200603444-200603444 on Build GRCh38
rs148871699 Chr.2: 200603357-200603357 on Build GRCh38
rs771935952 Chr.2: 200603333-200603333 on Build GRCh38
rs964215185 Chr.2: 200603449-200603449 on Build GRCh38
rs531684874 Chr.2: 200603515-200603515 on Build GRCh38
rs747927893 Chr.2: 200603328-200603328 on Build GRCh38
rs560153347 Chr.2: 200603428-200603428 on Build GRCh38

View Full Product Details