Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200603896-200604401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AOX1
Entrez Gene ID
316
Gene Name
aldehyde oxidase 1
Gene Aliases
AO, AOH1
UniGene
Hs.406238
Gene Chromosome Location
Chr.2: 200585952-200677064 on Build GRCh38
Transcripts
NM_001159.3 XM_011511062.1 XM_017003946.1 XM_017003947.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6543796 c.596C>T p.P199L Chr.2: 200604024-200604024 on Build GRCh38
7277673 c.625C>T p.P209S Chr.2: 200604053-200604053 on Build GRCh38
6090108 c.669G>T p.M223I Chr.2: 200604097-200604097 on Build GRCh38
3576187 c.655C>T p.P219S Chr.2: 200604083-200604083 on Build GRCh38
5651088 c.664C>A p.L222I Chr.2: 200604092-200604092 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140808647 Chr.2: 200604218-200604218 on Build GRCh38
rs759568130 Chr.2: 200604035-200604035 on Build GRCh38
rs548078335 Chr.2: 200604134-200604134 on Build GRCh38
rs1020522196 Chr.2: 200604301-200604301 on Build GRCh38
rs770784669 Chr.2: 200604024-200604024 on Build GRCh38
rs2293527 Chr.2: 200604366-200604366 on Build GRCh38
rs1034349024 Chr.2: 200603941-200603941 on Build GRCh38
rs747931915 Chr.2: 200603989-200603989 on Build GRCh38
rs753332584 Chr.2: 200603982-200603982 on Build GRCh38
rs369101350 Chr.2: 200604111-200604111 on Build GRCh38
rs529389023 Chr.2: 200604062-200604062 on Build GRCh38
rs776206162 Chr.2: 200603961-200603961 on Build GRCh38
rs895832706 Chr.2: 200604269-200604269 on Build GRCh38
rs1034806187 Chr.2: 200604221-200604221 on Build GRCh38
rs1052680889 Chr.2: 200603952-200603952 on Build GRCh38
rs879194388 Chr.2: 200604274-200604274 on Build GRCh38
rs564126941 Chr.2: 200603958-200603958 on Build GRCh38
rs753765495 Chr.2: 200604118-200604118 on Build GRCh38
rs752379517 Chr.2: 200604130-200604130 on Build GRCh38
rs780842453 Chr.2: 200603932-200603933 on Build GRCh38
rs899498809 Chr.2: 200604333-200604333 on Build GRCh38
rs552166198 Chr.2: 200604187-200604187 on Build GRCh38
rs973786299 Chr.2: 200604169-200604169 on Build GRCh38
rs1037538619 Chr.2: 200604205-200604205 on Build GRCh38
rs758192003 Chr.2: 200604131-200604131 on Build GRCh38
rs757007070 Chr.2: 200604173-200604173 on Build GRCh38
rs201006322 Chr.2: 200604110-200604110 on Build GRCh38
rs376036556 Chr.2: 200603931-200603931 on Build GRCh38
rs779706214 Chr.2: 200603975-200603975 on Build GRCh38
rs932958420 Chr.2: 200604096-200604096 on Build GRCh38
rs746941935 Chr.2: 200604023-200604023 on Build GRCh38
rs960278387 Chr.2: 200603943-200603943 on Build GRCh38
rs993095528 Chr.2: 200603956-200603956 on Build GRCh38
rs376178128 Chr.2: 200604034-200604034 on Build GRCh38
rs750169592 Chr.2: 200604196-200604196 on Build GRCh38
rs947004381 Chr.2: 200604253-200604253 on Build GRCh38
rs775752418 Chr.2: 200604042-200604042 on Build GRCh38
rs946349784 Chr.2: 200603980-200603980 on Build GRCh38
rs768272109 Chr.2: 200604126-200604128 on Build GRCh38
rs777545751 Chr.2: 200604135-200604135 on Build GRCh38
rs531568101 Chr.2: 200603983-200603983 on Build GRCh38
rs764845334 Chr.2: 200604119-200604119 on Build GRCh38
rs368304992 Chr.2: 200603977-200603977 on Build GRCh38
rs1014639162 Chr.2: 200604277-200604277 on Build GRCh38
rs2293526 Chr.2: 200604321-200604321 on Build GRCh38
rs143374796 Chr.2: 200604022-200604022 on Build GRCh38
rs1039391555 Chr.2: 200604007-200604007 on Build GRCh38
rs928069135 Chr.2: 200604191-200604191 on Build GRCh38
rs775065999 Chr.2: 200604063-200604063 on Build GRCh38
rs935525646 Chr.2: 200603922-200603922 on Build GRCh38
rs763279070 Chr.2: 200604055-200604055 on Build GRCh38
rs1016793300 Chr.2: 200604146-200604146 on Build GRCh38
rs768088310 Chr.2: 200604098-200604098 on Build GRCh38
rs2293525 Chr.2: 200604009-200604009 on Build GRCh38
rs762415096 Chr.2: 200604080-200604080 on Build GRCh38
rs962625631 Chr.2: 200604155-200604155 on Build GRCh38
rs112010507 Chr.2: 200604207-200604207 on Build GRCh38
rs755742002 Chr.2: 200603966-200603966 on Build GRCh38
rs537807454 Chr.2: 200604332-200604332 on Build GRCh38

View Full Product Details