Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200854596-200855115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLK1
Entrez Gene ID
1195
Gene Name
CDC like kinase 1
Gene Aliases
CLK, CLK/STY, STY
UniGene
Hs.433732
Gene Chromosome Location
Chr.2: 200853009-200864744 on Build GRCh38
Transcripts
NM_001162407.1 NM_004071.3 NR_027855.1 NR_027856.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5562415 c.1264C>T p.P422S Chr.2: 200855006-200855006 on Build GRCh38
5964806 c.1129A>G p.T377A Chr.2: 200855015-200855015 on Build GRCh38
1142373 c.1267-1G>T p.? Chr.2: 200854696-200854696 on Build GRCh38
3372602 c.1132G>T p.V378L Chr.2: 200855012-200855012 on Build GRCh38
5923582 c.1061T>C p.L354P Chr.2: 200855083-200855083 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs181420491 Chr.2: 200854902-200854902 on Build GRCh38
rs375541172 Chr.2: 200854959-200854959 on Build GRCh38
rs1026509128 Chr.2: 200854820-200854820 on Build GRCh38
rs771140179 Chr.2: 200854664-200854664 on Build GRCh38
rs770429089 Chr.2: 200854750-200854750 on Build GRCh38
rs762207952 Chr.2: 200855088-200855089 on Build GRCh38
rs112371737 Chr.2: 200854878-200854878 on Build GRCh38
rs751448751 Chr.2: 200855076-200855076 on Build GRCh38
rs898686830 Chr.2: 200854635-200854635 on Build GRCh38
rs765813649 Chr.2: 200854703-200854703 on Build GRCh38
rs372587239 Chr.2: 200854991-200854991 on Build GRCh38
rs772148575 Chr.2: 200854648-200854648 on Build GRCh38
rs753434038 Chr.2: 200854737-200854737 on Build GRCh38
rs111682852 Chr.2: 200854686-200854686 on Build GRCh38
rs377039077 Chr.2: 200855042-200855042 on Build GRCh38
rs778279257 Chr.2: 200854920-200854920 on Build GRCh38
rs757514465 Chr.2: 200855079-200855079 on Build GRCh38
rs578240338 Chr.2: 200854883-200854886 on Build GRCh38
rs201819229 Chr.2: 200854730-200854730 on Build GRCh38
rs898544489 Chr.2: 200854916-200854916 on Build GRCh38
rs368653939 Chr.2: 200854979-200854979 on Build GRCh38
rs887146464 Chr.2: 200854886-200854889 on Build GRCh38
rs1042600 Chr.2: 200855038-200855038 on Build GRCh38
rs923213694 Chr.2: 200854843-200854843 on Build GRCh38
rs1040082581 Chr.2: 200855026-200855026 on Build GRCh38
rs753251818 Chr.2: 200855031-200855031 on Build GRCh38
rs985097332 Chr.2: 200854842-200854842 on Build GRCh38
rs916149069 Chr.2: 200854848-200854848 on Build GRCh38
rs981419678 Chr.2: 200854895-200854895 on Build GRCh38
rs765857921 Chr.2: 200854736-200854736 on Build GRCh38
rs974955336 Chr.2: 200854825-200854825 on Build GRCh38
rs1013104144 Chr.2: 200854932-200854932 on Build GRCh38
rs759112804 Chr.2: 200855035-200855035 on Build GRCh38
rs200832543 Chr.2: 200854986-200854986 on Build GRCh38
rs748018926 Chr.2: 200854647-200854647 on Build GRCh38
rs776077029 Chr.2: 200854994-200854994 on Build GRCh38
rs747851768 Chr.2: 200854745-200854745 on Build GRCh38
rs768894843 Chr.2: 200854744-200854745 on Build GRCh38
rs190163896 Chr.2: 200854837-200854837 on Build GRCh38
rs777698575 Chr.2: 200855074-200855074 on Build GRCh38
rs994050821 Chr.2: 200854812-200854812 on Build GRCh38
rs138594554 Chr.2: 200854690-200854690 on Build GRCh38
rs530765532 Chr.2: 200854831-200854831 on Build GRCh38
rs981107368 Chr.2: 200854700-200854704 on Build GRCh38
rs14379 Chr.2: 200854666-200854666 on Build GRCh38
rs763396314 Chr.2: 200854711-200854711 on Build GRCh38
rs372629858 Chr.2: 200854822-200854822 on Build GRCh38
rs549157908 Chr.2: 200854846-200854846 on Build GRCh38
rs764435537 Chr.2: 200855012-200855012 on Build GRCh38
rs758478473 Chr.2: 200855064-200855064 on Build GRCh38
rs952384843 Chr.2: 200854821-200854821 on Build GRCh38
rs545702098 Chr.2: 200854906-200854909 on Build GRCh38
rs550616258 Chr.2: 200854740-200854740 on Build GRCh38
rs775676991 Chr.2: 200854708-200854708 on Build GRCh38
rs563152323 Chr.2: 200854747-200854747 on Build GRCh38
rs199843052 Chr.2: 200854651-200854651 on Build GRCh38
rs747231237 Chr.2: 200854975-200854975 on Build GRCh38
rs75615141 Chr.2: 200854668-200854668 on Build GRCh38
rs965800046 Chr.2: 200854776-200854776 on Build GRCh38
rs577465397 Chr.2: 200854988-200854988 on Build GRCh38
rs763159000 Chr.2: 200854996-200854996 on Build GRCh38
rs969995565 Chr.2: 200854876-200854876 on Build GRCh38
rs141001362 Chr.2: 200854943-200854943 on Build GRCh38
rs777090158 Chr.2: 200854988-200854988 on Build GRCh38
rs940005701 Chr.2: 200854887-200854890 on Build GRCh38
rs781498072 Chr.2: 200855084-200855084 on Build GRCh38
rs913769676 Chr.2: 200854950-200854950 on Build GRCh38
rs767097735 Chr.2: 200854720-200854720 on Build GRCh38
rs774751838 Chr.2: 200855013-200855013 on Build GRCh38
rs943247060 Chr.2: 200854799-200854799 on Build GRCh38
rs745876196 Chr.2: 200855085-200855085 on Build GRCh38
rs778944129 Chr.2: 200854742-200854742 on Build GRCh38
rs563370189 Chr.2: 200854631-200854631 on Build GRCh38
rs750078109 Chr.2: 200854728-200854728 on Build GRCh38
rs770149713 Chr.2: 200854993-200854993 on Build GRCh38
rs564904754 Chr.2: 200854699-200854699 on Build GRCh38
rs766056400 Chr.2: 200855029-200855029 on Build GRCh38
rs955683889 Chr.2: 200854717-200854717 on Build GRCh38
rs988365784 Chr.2: 200854935-200854935 on Build GRCh38
rs1018673337 Chr.2: 200854743-200854743 on Build GRCh38

View Full Product Details