Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200975068-200975585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM126B
Entrez Gene ID
285172
Gene Name
family with sequence similarity 126 member B
Gene Aliases
HYCC2
UniGene
Hs.24701
Gene Chromosome Location
Chr.2: 200973718-201071671 on Build GRCh38
Transcripts
NM_001321618.1 NM_001321619.1 NM_001321621.1 NM_001321622.1 NM_001321623.1 NM_001321624.1 NM_001321625.1 NM_001321626.1 NM_001321627.1 NM_001321628.1 NM_001321629.1 NM_173822.4

SNP Detail

SNP ID Chromosome Location
rs368505659 Chr.2: 200975188-200975188 on Build GRCh38
rs779388932 Chr.2: 200975313-200975313 on Build GRCh38
rs893364667 Chr.2: 200975330-200975330 on Build GRCh38
rs72929038 Chr.2: 200975171-200975171 on Build GRCh38
rs985003713 Chr.2: 200975420-200975420 on Build GRCh38
rs909164850 Chr.2: 200975460-200975463 on Build GRCh38
rs1039180505 Chr.2: 200975505-200975505 on Build GRCh38
rs144109477 Chr.2: 200975360-200975360 on Build GRCh38
rs185287103 Chr.2: 200975232-200975232 on Build GRCh38
rs922096302 Chr.2: 200975512-200975512 on Build GRCh38
rs886667407 Chr.2: 200975525-200975525 on Build GRCh38
rs959313985 Chr.2: 200975354-200975354 on Build GRCh38
rs936389945 Chr.2: 200975477-200975477 on Build GRCh38
rs753847507 Chr.2: 200975239-200975239 on Build GRCh38
rs146473046 Chr.2: 200975541-200975541 on Build GRCh38
rs763843872 Chr.2: 200975118-200975118 on Build GRCh38
rs900770477 Chr.2: 200975205-200975205 on Build GRCh38
rs991490883 Chr.2: 200975496-200975496 on Build GRCh38
rs4035023 Chr.2: 200975295-200975295 on Build GRCh38
rs904053292 Chr.2: 200975124-200975124 on Build GRCh38
rs903357074 Chr.2: 200975242-200975242 on Build GRCh38
rs552424382 Chr.2: 200975394-200975394 on Build GRCh38
rs562232874 Chr.2: 200975252-200975252 on Build GRCh38
rs181842367 Chr.2: 200975349-200975349 on Build GRCh38
rs1042123155 Chr.2: 200975119-200975119 on Build GRCh38
rs757250925 Chr.2: 200975194-200975194 on Build GRCh38
rs945041198 Chr.2: 200975374-200975374 on Build GRCh38
rs917586395 Chr.2: 200975114-200975114 on Build GRCh38
rs16836427 Chr.2: 200975370-200975370 on Build GRCh38
rs1030783135 Chr.2: 200975249-200975249 on Build GRCh38
rs570688577 Chr.2: 200975515-200975515 on Build GRCh38
rs564311342 Chr.2: 200975185-200975185 on Build GRCh38
rs755621808 Chr.2: 200975397-200975397 on Build GRCh38
rs139377954 Chr.2: 200975240-200975240 on Build GRCh38
rs978137662 Chr.2: 200975376-200975376 on Build GRCh38
rs1005400772 Chr.2: 200975265-200975265 on Build GRCh38
rs771144549 Chr.2: 200975513-200975514 on Build GRCh38
rs186908792 Chr.2: 200975522-200975522 on Build GRCh38
rs1016370250 Chr.2: 200975271-200975271 on Build GRCh38
rs1026444076 Chr.2: 200975403-200975403 on Build GRCh38
rs903241211 Chr.2: 200975559-200975559 on Build GRCh38
rs925046584 Chr.2: 200975406-200975406 on Build GRCh38
rs189628234 Chr.2: 200975293-200975293 on Build GRCh38
rs937912881 Chr.2: 200975486-200975487 on Build GRCh38

View Full Product Details