Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200976265-200976766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM126B
Entrez Gene ID
285172
Gene Name
family with sequence similarity 126 member B
Gene Aliases
HYCC2
UniGene
Hs.24701
Gene Chromosome Location
Chr.2: 200973718-201071671 on Build GRCh38
Transcripts
NM_001321618.1 NM_001321619.1 NM_001321621.1 NM_001321622.1 NM_001321623.1 NM_001321624.1 NM_001321625.1 NM_001321626.1 NM_001321627.1 NM_001321628.1 NM_001321629.1 NM_173822.4

SNP Detail

SNP ID Chromosome Location
rs1037429619 Chr.2: 200976459-200976459 on Build GRCh38
rs974357169 Chr.2: 200976287-200976287 on Build GRCh38
rs895907339 Chr.2: 200976675-200976675 on Build GRCh38
rs974949622 Chr.2: 200976680-200976680 on Build GRCh38
rs764210369 Chr.2: 200976686-200976686 on Build GRCh38
rs748521259 Chr.2: 200976677-200976677 on Build GRCh38
rs549668090 Chr.2: 200976372-200976372 on Build GRCh38
rs984110436 Chr.2: 200976714-200976714 on Build GRCh38
rs908035902 Chr.2: 200976452-200976452 on Build GRCh38
rs34896158 Chr.2: 200976295-200976295 on Build GRCh38
rs114272559 Chr.2: 200976685-200976685 on Build GRCh38
rs886111041 Chr.2: 200976416-200976416 on Build GRCh38
rs558787297 Chr.2: 200976479-200976479 on Build GRCh38
rs183316404 Chr.2: 200976627-200976627 on Build GRCh38
rs987661710 Chr.2: 200976412-200976412 on Build GRCh38
rs557473997 Chr.2: 200976659-200976659 on Build GRCh38
rs940989414 Chr.2: 200976490-200976490 on Build GRCh38
rs914637766 Chr.2: 200976311-200976311 on Build GRCh38
rs1025209846 Chr.2: 200976553-200976553 on Build GRCh38
rs1039209380 Chr.2: 200976354-200976354 on Build GRCh38
rs565370119 Chr.2: 200976588-200976588 on Build GRCh38
rs906489454 Chr.2: 200976556-200976556 on Build GRCh38
rs937342083 Chr.2: 200976650-200976650 on Build GRCh38
rs770781856 Chr.2: 200976369-200976369 on Build GRCh38
rs535357847 Chr.2: 200976483-200976483 on Build GRCh38
rs1012062956 Chr.2: 200976550-200976550 on Build GRCh38
rs943530772 Chr.2: 200976353-200976353 on Build GRCh38
rs762889438 Chr.2: 200976355-200976355 on Build GRCh38
rs894866831 Chr.2: 200976468-200976468 on Build GRCh38
rs547265829 Chr.2: 200976528-200976528 on Build GRCh38
rs1047426810 Chr.2: 200976389-200976389 on Build GRCh38
rs773975118 Chr.2: 200976605-200976605 on Build GRCh38
rs537663512 Chr.2: 200976727-200976727 on Build GRCh38
rs990121210 Chr.2: 200976661-200976661 on Build GRCh38
rs1021747190 Chr.2: 200976672-200976672 on Build GRCh38

View Full Product Details