Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200976548-200977089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM126B
Entrez Gene ID
285172
Gene Name
family with sequence similarity 126 member B
Gene Aliases
HYCC2
UniGene
Hs.24701
Gene Chromosome Location
Chr.2: 200973718-201071671 on Build GRCh38
Transcripts
NM_001321618.1 NM_001321619.1 NM_001321621.1 NM_001321622.1 NM_001321623.1 NM_001321624.1 NM_001321625.1 NM_001321626.1 NM_001321627.1 NM_001321628.1 NM_001321629.1 NM_173822.4

SNP Detail

SNP ID Chromosome Location
rs893715496 Chr.2: 200976937-200976937 on Build GRCh38
rs895907339 Chr.2: 200976675-200976675 on Build GRCh38
rs867076523 Chr.2: 200976889-200976889 on Build GRCh38
rs764210369 Chr.2: 200976686-200976686 on Build GRCh38
rs974949622 Chr.2: 200976680-200976680 on Build GRCh38
rs748521259 Chr.2: 200976677-200976677 on Build GRCh38
rs765782291 Chr.2: 200976910-200976910 on Build GRCh38
rs1037314038 Chr.2: 200976778-200976778 on Build GRCh38
rs984110436 Chr.2: 200976714-200976714 on Build GRCh38
rs1032240613 Chr.2: 200976955-200976955 on Build GRCh38
rs1046260813 Chr.2: 200976804-200976804 on Build GRCh38
rs1031350098 Chr.2: 200976921-200976921 on Build GRCh38
rs773872021 Chr.2: 200976832-200976832 on Build GRCh38
rs995807659 Chr.2: 200977003-200977003 on Build GRCh38
rs114272559 Chr.2: 200976685-200976685 on Build GRCh38
rs183316404 Chr.2: 200976627-200976627 on Build GRCh38
rs954526800 Chr.2: 200977019-200977019 on Build GRCh38
rs557473997 Chr.2: 200976659-200976659 on Build GRCh38
rs1011609035 Chr.2: 200976959-200976959 on Build GRCh38
rs113219594 Chr.2: 200976761-200976761 on Build GRCh38
rs908094197 Chr.2: 200976750-200976750 on Build GRCh38
rs140119277 Chr.2: 200976757-200976757 on Build GRCh38
rs565370119 Chr.2: 200976588-200976588 on Build GRCh38
rs937342083 Chr.2: 200976650-200976650 on Build GRCh38
rs573355402 Chr.2: 200976803-200976803 on Build GRCh38
rs893696236 Chr.2: 200976997-200976997 on Build GRCh38
rs947803697 Chr.2: 200976802-200976802 on Build GRCh38
rs773975118 Chr.2: 200976605-200976605 on Build GRCh38
rs537663512 Chr.2: 200976727-200976727 on Build GRCh38
rs1029038997 Chr.2: 200977009-200977009 on Build GRCh38
rs753297380 Chr.2: 200976779-200976781 on Build GRCh38
rs1021747190 Chr.2: 200976672-200976672 on Build GRCh38
rs990121210 Chr.2: 200976661-200976661 on Build GRCh38
rs574298724 Chr.2: 200976782-200976782 on Build GRCh38
rs999344062 Chr.2: 200976897-200976897 on Build GRCh38

View Full Product Details