Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200978336-200978885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM126B
Entrez Gene ID
285172
Gene Name
family with sequence similarity 126 member B
Gene Aliases
HYCC2
UniGene
Hs.24701
Gene Chromosome Location
Chr.2: 200973718-201071671 on Build GRCh38
Transcripts
NM_001321618.1 NM_001321619.1 NM_001321621.1 NM_001321622.1 NM_001321623.1 NM_001321624.1 NM_001321625.1 NM_001321626.1 NM_001321627.1 NM_001321628.1 NM_001321629.1 NM_173822.4

SNP Detail

SNP ID Chromosome Location
rs930309049 Chr.2: 200978425-200978425 on Build GRCh38
rs967769892 Chr.2: 200978669-200978669 on Build GRCh38
rs906926742 Chr.2: 200978853-200978853 on Build GRCh38
rs10198771 Chr.2: 200978663-200978663 on Build GRCh38
rs948505943 Chr.2: 200978771-200978771 on Build GRCh38
rs1023757008 Chr.2: 200978429-200978430 on Build GRCh38
rs13418869 Chr.2: 200978496-200978496 on Build GRCh38
rs796198054 Chr.2: 200978481-200978483 on Build GRCh38
rs10198684 Chr.2: 200978577-200978577 on Build GRCh38
rs774167011 Chr.2: 200978465-200978469 on Build GRCh38
rs894219621 Chr.2: 200978499-200978499 on Build GRCh38
rs1035643682 Chr.2: 200978602-200978602 on Build GRCh38
rs954308247 Chr.2: 200978696-200978696 on Build GRCh38
rs557615655 Chr.2: 200978491-200978491 on Build GRCh38
rs907357448 Chr.2: 200978622-200978622 on Build GRCh38
rs866799985 Chr.2: 200978742-200978742 on Build GRCh38
rs149403419 Chr.2: 200978765-200978765 on Build GRCh38
rs750437984 Chr.2: 200978810-200978810 on Build GRCh38
rs536645782 Chr.2: 200978601-200978601 on Build GRCh38
rs10198792 Chr.2: 200978736-200978736 on Build GRCh38
rs10198870 Chr.2: 200978777-200978777 on Build GRCh38
rs575268162 Chr.2: 200978672-200978672 on Build GRCh38
rs761978159 Chr.2: 200978764-200978764 on Build GRCh38
rs10198881 Chr.2: 200978818-200978818 on Build GRCh38
rs560975271 Chr.2: 200978808-200978808 on Build GRCh38
rs763533025 Chr.2: 200978847-200978847 on Build GRCh38
rs986896023 Chr.2: 200978708-200978708 on Build GRCh38
rs1019223007 Chr.2: 200978740-200978740 on Build GRCh38
rs76181344 Chr.2: 200978486-200978486 on Build GRCh38
rs555409709 Chr.2: 200978570-200978570 on Build GRCh38
rs544376221 Chr.2: 200978673-200978673 on Build GRCh38
rs940725262 Chr.2: 200978767-200978767 on Build GRCh38
rs930721740 Chr.2: 200978778-200978778 on Build GRCh38
rs941282248 Chr.2: 200978770-200978770 on Build GRCh38
rs905478052 Chr.2: 200978507-200978507 on Build GRCh38
rs987775751 Chr.2: 200978631-200978631 on Build GRCh38
rs72929040 Chr.2: 200978807-200978807 on Build GRCh38
rs971862626 Chr.2: 200978755-200978755 on Build GRCh38
rs905364507 Chr.2: 200978809-200978809 on Build GRCh38
rs1046356303 Chr.2: 200978550-200978550 on Build GRCh38
rs778614995 Chr.2: 200978510-200978510 on Build GRCh38
rs1035294207 Chr.2: 200978814-200978814 on Build GRCh38
rs1003809019 Chr.2: 200978512-200978512 on Build GRCh38
rs10198686 Chr.2: 200978579-200978579 on Build GRCh38
rs527453236 Chr.2: 200978402-200978402 on Build GRCh38
rs572957427 Chr.2: 200978633-200978633 on Build GRCh38
rs188671480 Chr.2: 200978632-200978632 on Build GRCh38
rs975281123 Chr.2: 200978678-200978678 on Build GRCh38
rs11888171 Chr.2: 200978534-200978534 on Build GRCh38
rs545011555 Chr.2: 200978757-200978757 on Build GRCh38
rs10198597 Chr.2: 200978494-200978494 on Build GRCh38
rs965887573 Chr.2: 200978754-200978754 on Build GRCh38
rs546464825 Chr.2: 200978811-200978812 on Build GRCh38
rs139286738 Chr.2: 200978761-200978761 on Build GRCh38
rs921097900 Chr.2: 200978737-200978737 on Build GRCh38

View Full Product Details