Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200981064-200981607 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM126B
Entrez Gene ID
285172
Gene Name
family with sequence similarity 126 member B
Gene Aliases
HYCC2
UniGene
Hs.24701
Gene Chromosome Location
Chr.2: 200973718-201071671 on Build GRCh38
Transcripts
NM_001321618.1 NM_001321619.1 NM_001321621.1 NM_001321622.1 NM_001321623.1 NM_001321624.1 NM_001321625.1 NM_001321626.1 NM_001321627.1 NM_001321628.1 NM_001321629.1 NM_173822.4 XM_006712443.3 XM_011511002.2 XM_017003879.1 XM_017003880.1 XM_017003881.1 XM_017003882.1 XM_017003883.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7319815 c.1457G>A p.R486Q Chr.2: 200981406-200981406 on Build GRCh38
7331807 c.1466C>T p.T489I Chr.2: 200981397-200981397 on Build GRCh38
1129385 c.1477C>T p.Q493* Chr.2: 200981386-200981386 on Build GRCh38
7366978 c.1259T>A p.V420D Chr.2: 200981604-200981604 on Build GRCh38
1168924 c.1415G>C p.G472A Chr.2: 200981448-200981448 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748345663 Chr.2: 200981269-200981269 on Build GRCh38
rs942628706 Chr.2: 200981552-200981552 on Build GRCh38
rs900955110 Chr.2: 200981137-200981137 on Build GRCh38
rs763334144 Chr.2: 200981383-200981383 on Build GRCh38
rs201168147 Chr.2: 200981276-200981276 on Build GRCh38
rs925862134 Chr.2: 200981503-200981503 on Build GRCh38
rs1053787624 Chr.2: 200981253-200981253 on Build GRCh38
rs960806440 Chr.2: 200981345-200981345 on Build GRCh38
rs759100696 Chr.2: 200981538-200981538 on Build GRCh38
rs751717333 Chr.2: 200981570-200981570 on Build GRCh38
rs958575569 Chr.2: 200981406-200981406 on Build GRCh38
rs749285810 Chr.2: 200981230-200981230 on Build GRCh38
rs777078217 Chr.2: 200981295-200981295 on Build GRCh38
rs760435799 Chr.2: 200981466-200981466 on Build GRCh38
rs143404685 Chr.2: 200981431-200981431 on Build GRCh38
rs185335735 Chr.2: 200981168-200981168 on Build GRCh38
rs755748322 Chr.2: 200981224-200981224 on Build GRCh38
rs772949644 Chr.2: 200981457-200981457 on Build GRCh38
rs889899905 Chr.2: 200981254-200981254 on Build GRCh38
rs1039813790 Chr.2: 200981559-200981559 on Build GRCh38
rs779840554 Chr.2: 200981310-200981310 on Build GRCh38
rs13028562 Chr.2: 200981159-200981159 on Build GRCh38
rs867011871 Chr.2: 200981250-200981250 on Build GRCh38
rs773272054 Chr.2: 200981437-200981437 on Build GRCh38
rs770962602 Chr.2: 200981412-200981412 on Build GRCh38
rs13028001 Chr.2: 200981126-200981126 on Build GRCh38
rs774908769 Chr.2: 200981550-200981560 on Build GRCh38
rs752731064 Chr.2: 200981502-200981502 on Build GRCh38
rs1017618417 Chr.2: 200981203-200981203 on Build GRCh38
rs190216711 Chr.2: 200981118-200981118 on Build GRCh38
rs747892607 Chr.2: 200981376-200981376 on Build GRCh38
rs770065299 Chr.2: 200981407-200981407 on Build GRCh38
rs763430862 Chr.2: 200981326-200981326 on Build GRCh38
rs750139679 Chr.2: 200981339-200981339 on Build GRCh38
rs766820309 Chr.2: 200981427-200981427 on Build GRCh38
rs945894876 Chr.2: 200981432-200981432 on Build GRCh38
rs765920830 Chr.2: 200981470-200981470 on Build GRCh38
rs978529728 Chr.2: 200981461-200981461 on Build GRCh38
rs138302584 Chr.2: 200981282-200981282 on Build GRCh38
rs759153730 Chr.2: 200981487-200981487 on Build GRCh38
rs753350105 Chr.2: 200981501-200981501 on Build GRCh38
rs758254199 Chr.2: 200981377-200981377 on Build GRCh38
rs997929117 Chr.2: 200981147-200981147 on Build GRCh38
rs771050626 Chr.2: 200981293-200981293 on Build GRCh38
rs753385948 Chr.2: 200981486-200981486 on Build GRCh38
rs201247752 Chr.2: 200981462-200981462 on Build GRCh38
rs777952927 Chr.2: 200981403-200981403 on Build GRCh38
rs776699767 Chr.2: 200981418-200981418 on Build GRCh38
rs143338916 Chr.2: 200981347-200981347 on Build GRCh38
rs541399085 Chr.2: 200981132-200981132 on Build GRCh38
rs569072628 Chr.2: 200981520-200981520 on Build GRCh38
rs560019737 Chr.2: 200981144-200981144 on Build GRCh38
rs757479553 Chr.2: 200981577-200981577 on Build GRCh38
rs945838402 Chr.2: 200981150-200981150 on Build GRCh38
rs372270432 Chr.2: 200981359-200981359 on Build GRCh38
rs778603905 Chr.2: 200981260-200981260 on Build GRCh38
rs17467902 Chr.2: 200981360-200981360 on Build GRCh38
rs139514675 Chr.2: 200981489-200981489 on Build GRCh38
rs763193756 Chr.2: 200981439-200981439 on Build GRCh38
rs767068954 Chr.2: 200981329-200981329 on Build GRCh38
rs181143027 Chr.2: 200981166-200981166 on Build GRCh38
rs770184297 Chr.2: 200981422-200981422 on Build GRCh38
rs376627039 Chr.2: 200981467-200981467 on Build GRCh38
rs777578552 Chr.2: 200981523-200981523 on Build GRCh38
rs764567449 Chr.2: 200981442-200981442 on Build GRCh38
rs538309573 Chr.2: 200981355-200981355 on Build GRCh38
rs778968205 Chr.2: 200981369-200981369 on Build GRCh38
rs139592690 Chr.2: 200981167-200981167 on Build GRCh38
rs775726332 Chr.2: 200981321-200981321 on Build GRCh38
rs760071040 Chr.2: 200981302-200981302 on Build GRCh38
rs778438869 Chr.2: 200981124-200981124 on Build GRCh38
rs780107389 Chr.2: 200981229-200981229 on Build GRCh38
rs754752409 Chr.2: 200981244-200981244 on Build GRCh38

View Full Product Details