Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201135589-201136133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFLAR
Entrez Gene ID
8837
Gene Name
CASP8 and FADD like apoptosis regulator
Gene Aliases
CASH, CASP8AP1, CLARP, Casper, FLAME, FLAME-1, FLAME1, FLIP, I-FLICE, MRIT, c-FLIP, c-FLIPL, c-FLIPR, c-FLIPS
UniGene
Hs.390736
Gene Chromosome Location
Chr.2: 201116015-201172688 on Build GRCh38
Transcripts
NM_001127183.2 NM_001127184.2 NM_001202516.1 NM_001202517.1 NM_001202518.1 NM_001202519.1 NM_001308042.1 NM_001308043.1 NM_003879.5 XM_017005189.1 XM_017005190.1 XM_017005191.1 XM_017005192.1 XM_017005193.1 XM_017005194.1 XM_017005195.1 XM_017005196.1 XM_017005197.1 XM_017005198.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6627570 c.510G>A p.K170K Chr.2: 201136094-201136094 on Build GRCh38
6987864 c.507G>T p.Q169H Chr.2: 201136091-201136091 on Build GRCh38
6543802 c.492G>T p.L164L Chr.2: 201136076-201136076 on Build GRCh38
1306293 c.409G>A p.E137K Chr.2: 201135993-201135993 on Build GRCh38
6849219 c.416A>G p.E139G Chr.2: 201136000-201136000 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991524026 Chr.2: 201135755-201135755 on Build GRCh38
rs781033253 Chr.2: 201135778-201135778 on Build GRCh38
rs752712939 Chr.2: 201136040-201136040 on Build GRCh38
rs79528267 Chr.2: 201135647-201135647 on Build GRCh38
rs750022461 Chr.2: 201135938-201135938 on Build GRCh38
rs754566160 Chr.2: 201135962-201135962 on Build GRCh38
rs747799016 Chr.2: 201135779-201135779 on Build GRCh38
rs545097642 Chr.2: 201135727-201135727 on Build GRCh38
rs755768732 Chr.2: 201135820-201135820 on Build GRCh38
rs905024221 Chr.2: 201135802-201135802 on Build GRCh38
rs777427018 Chr.2: 201135876-201135876 on Build GRCh38
rs200873517 Chr.2: 201135703-201135704 on Build GRCh38
rs1036007422 Chr.2: 201135769-201135769 on Build GRCh38
rs549597369 Chr.2: 201135933-201135933 on Build GRCh38
rs923787849 Chr.2: 201135826-201135826 on Build GRCh38
rs528965856 Chr.2: 201135636-201135636 on Build GRCh38
rs180961724 Chr.2: 201135621-201135621 on Build GRCh38
rs4034920 Chr.2: 201135706-201135707 on Build GRCh38
rs766125704 Chr.2: 201135941-201135941 on Build GRCh38
rs932162099 Chr.2: 201136055-201136055 on Build GRCh38
rs531410289 Chr.2: 201135939-201135939 on Build GRCh38
rs1015536728 Chr.2: 201135661-201135661 on Build GRCh38
rs922089982 Chr.2: 201135646-201135646 on Build GRCh38
rs73045306 Chr.2: 201135672-201135672 on Build GRCh38
rs866121768 Chr.2: 201135760-201135760 on Build GRCh38
rs898852302 Chr.2: 201135776-201135776 on Build GRCh38
rs776083486 Chr.2: 201135926-201135926 on Build GRCh38
rs994682661 Chr.2: 201135785-201135785 on Build GRCh38
rs968714953 Chr.2: 201135955-201135955 on Build GRCh38
rs764449760 Chr.2: 201135929-201135929 on Build GRCh38
rs563981967 Chr.2: 201135818-201135818 on Build GRCh38
rs763261328 Chr.2: 201135927-201135927 on Build GRCh38
rs758482965 Chr.2: 201135969-201135970 on Build GRCh38
rs535524387 Chr.2: 201135791-201135792 on Build GRCh38
rs868581421 Chr.2: 201135685-201135685 on Build GRCh38
rs890746469 Chr.2: 201135633-201135633 on Build GRCh38
rs988840859 Chr.2: 201135612-201135612 on Build GRCh38
rs915029478 Chr.2: 201135888-201135888 on Build GRCh38
rs77669745 Chr.2: 201135648-201135648 on Build GRCh38
rs753256043 Chr.2: 201135956-201135956 on Build GRCh38
rs935054577 Chr.2: 201135838-201135838 on Build GRCh38
rs778667629 Chr.2: 201135974-201135974 on Build GRCh38
rs978969583 Chr.2: 201135958-201135958 on Build GRCh38
rs970659374 Chr.2: 201135817-201135817 on Build GRCh38
rs755277403 Chr.2: 201135733-201135733 on Build GRCh38
rs1009002881 Chr.2: 201135660-201135660 on Build GRCh38

View Full Product Details