Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201141248-201141748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFLAR
Entrez Gene ID
8837
Gene Name
CASP8 and FADD like apoptosis regulator
Gene Aliases
CASH, CASP8AP1, CLARP, Casper, FLAME, FLAME-1, FLAME1, FLIP, I-FLICE, MRIT, c-FLIP, c-FLIPL, c-FLIPR, c-FLIPS
UniGene
Hs.390736
Gene Chromosome Location
Chr.2: 201116015-201172688 on Build GRCh38
Transcripts
NM_001127183.2 NM_001127184.2 NM_001202515.1 NM_001202516.1 NM_001202517.1 NM_001202518.1 NM_001202519.1 NM_001308042.1 NM_003879.5 XM_017005189.1 XM_017005190.1 XM_017005191.1 XM_017005192.1 XM_017005193.1 XM_017005194.1 XM_017005195.1 XM_017005196.1 XM_017005197.1 XM_017005198.1 NR_040030.1

Gene Symbol
CFLAR-AS1
Entrez Gene ID
65072
Gene Name
CFLAR antisense RNA 1
Gene Aliases
ALS2CR10
UniGene
Hs.664613
Gene Chromosome Location
Chr.2: 201140289-201157792 on Build GRCh38
Transcripts
NM_001127183.2 NM_001127184.2 NM_001202515.1 NM_001202516.1 NM_001202517.1 NM_001202518.1 NM_001202519.1 NM_001308042.1 NM_003879.5 XM_017005189.1 XM_017005190.1 XM_017005191.1 XM_017005192.1 XM_017005193.1 XM_017005194.1 XM_017005195.1 XM_017005196.1 XM_017005197.1 XM_017005198.1 NR_040030.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6301286 c.607-4A>G p.? Chr.2: 201141370-201141370 on Build GRCh38
4504224 c.659C>T p.S220F Chr.2: 201141426-201141426 on Build GRCh38
3757874 c.607-1G>A p.? Chr.2: 201141373-201141373 on Build GRCh38
1014804 c.646C>A p.L216I Chr.2: 201141413-201141413 on Build GRCh38
5395926 c.631C>T p.P211S Chr.2: 201141398-201141398 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs866743181 Chr.2: 201141417-201141417 on Build GRCh38
rs1020120695 Chr.2: 201141712-201141712 on Build GRCh38
rs765391129 Chr.2: 201141475-201141475 on Build GRCh38
rs978920447 Chr.2: 201141340-201141340 on Build GRCh38
rs752889951 Chr.2: 201141479-201141479 on Build GRCh38
rs972383917 Chr.2: 201141465-201141465 on Build GRCh38
rs397714976 Chr.2: 201141705-201141706 on Build GRCh38
rs1002231003 Chr.2: 201141573-201141573 on Build GRCh38
rs905036379 Chr.2: 201141545-201141545 on Build GRCh38
rs752012429 Chr.2: 201141325-201141325 on Build GRCh38
rs577683719 Chr.2: 201141609-201141609 on Build GRCh38
rs770915594 Chr.2: 201141418-201141418 on Build GRCh38
rs55941675 Chr.2: 201141703-201141704 on Build GRCh38
rs969280906 Chr.2: 201141273-201141273 on Build GRCh38
rs545631305 Chr.2: 201141714-201141714 on Build GRCh38
rs934150591 Chr.2: 201141395-201141395 on Build GRCh38
rs1010269608 Chr.2: 201141689-201141689 on Build GRCh38
rs776376711 Chr.2: 201141419-201141419 on Build GRCh38
rs904747757 Chr.2: 201141699-201141699 on Build GRCh38
rs184131473 Chr.2: 201141501-201141501 on Build GRCh38
rs937370160 Chr.2: 201141587-201141587 on Build GRCh38
rs771081473 Chr.2: 201141467-201141467 on Build GRCh38
rs867314690 Chr.2: 201141416-201141416 on Build GRCh38
rs746303316 Chr.2: 201141365-201141365 on Build GRCh38
rs556568607 Chr.2: 201141723-201141723 on Build GRCh38
rs1034680070 Chr.2: 201141588-201141588 on Build GRCh38
rs781679266 Chr.2: 201141363-201141363 on Build GRCh38
rs768392980 Chr.2: 201141366-201141366 on Build GRCh38
rs1029082332 Chr.2: 201141722-201141722 on Build GRCh38
rs1020940004 Chr.2: 201141693-201141693 on Build GRCh38
rs866615741 Chr.2: 201141389-201141389 on Build GRCh38
rs556198239 Chr.2: 201141647-201141651 on Build GRCh38
rs773138912 Chr.2: 201141383-201141383 on Build GRCh38
rs771517176 Chr.2: 201141374-201141374 on Build GRCh38
rs759372219 Chr.2: 201141444-201141444 on Build GRCh38
rs905205311 Chr.2: 201141514-201141514 on Build GRCh38
rs757637682 Chr.2: 201141343-201141343 on Build GRCh38
rs10190751 Chr.2: 201141373-201141373 on Build GRCh38
rs371458989 Chr.2: 201141370-201141370 on Build GRCh38
rs1014471140 Chr.2: 201141608-201141608 on Build GRCh38
rs960019927 Chr.2: 201141590-201141590 on Build GRCh38
rs186755151 Chr.2: 201141703-201141703 on Build GRCh38
rs920551436 Chr.2: 201141478-201141478 on Build GRCh38
rs760546684 Chr.2: 201141415-201141415 on Build GRCh38
rs1057277477 Chr.2: 201141635-201141635 on Build GRCh38
rs1005640282 Chr.2: 201141449-201141449 on Build GRCh38

View Full Product Details