Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201208040-201208587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP10
Entrez Gene ID
843
Gene Name
caspase 10
Gene Aliases
ALPS2, FLICE2, MCH4
UniGene
Hs.5353
Gene Chromosome Location
Chr.2: 201182881-201229406 on Build GRCh38
Transcripts
NM_001206524.1 NM_001206542.1 NM_001230.4 NM_032974.4 NM_032976.3 NM_032977.3 XM_005246907.2 XM_006712796.3 XM_011511990.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5861487 c.874C>T p.H292Y Chr.2: 201208135-201208135 on Build GRCh38
3909328 c.852C>T p.G284G Chr.2: 201208113-201208113 on Build GRCh38
4764375 c.922+1G>A p.? Chr.2: 201208184-201208184 on Build GRCh38
3909329 c.852C>T p.G284G Chr.2: 201208113-201208113 on Build GRCh38
4764376 c.922+1G>A p.? Chr.2: 201208184-201208184 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367848313 Chr.2: 201208221-201208221 on Build GRCh38
rs769651296 Chr.2: 201208177-201208178 on Build GRCh38
rs932870591 Chr.2: 201208542-201208542 on Build GRCh38
rs559244194 Chr.2: 201208188-201208188 on Build GRCh38
rs538123927 Chr.2: 201208519-201208519 on Build GRCh38
rs746636209 Chr.2: 201208124-201208124 on Build GRCh38
rs1014775414 Chr.2: 201208277-201208277 on Build GRCh38
rs6750157 Chr.2: 201208426-201208426 on Build GRCh38
rs113096171 Chr.2: 201208211-201208211 on Build GRCh38
rs981465275 Chr.2: 201208172-201208172 on Build GRCh38
rs566998504 Chr.2: 201208451-201208451 on Build GRCh38
rs748988477 Chr.2: 201208171-201208171 on Build GRCh38
rs756914853 Chr.2: 201208218-201208218 on Build GRCh38
rs753175958 Chr.2: 201208215-201208215 on Build GRCh38
rs762072082 Chr.2: 201208069-201208069 on Build GRCh38
rs113867411 Chr.2: 201208077-201208077 on Build GRCh38
rs936253557 Chr.2: 201208162-201208162 on Build GRCh38
rs187232960 Chr.2: 201208478-201208478 on Build GRCh38
rs760814428 Chr.2: 201208200-201208200 on Build GRCh38
rs150160468 Chr.2: 201208253-201208253 on Build GRCh38
rs912672004 Chr.2: 201208563-201208563 on Build GRCh38
rs775531682 Chr.2: 201208155-201208155 on Build GRCh38
rs897717908 Chr.2: 201208282-201208282 on Build GRCh38
rs138607846 Chr.2: 201208288-201208288 on Build GRCh38
rs71022355 Chr.2: 201208406-201208407 on Build GRCh38
rs757006791 Chr.2: 201208128-201208128 on Build GRCh38
rs774579828 Chr.2: 201208065-201208065 on Build GRCh38
rs371741849 Chr.2: 201208345-201208345 on Build GRCh38
rs1013074144 Chr.2: 201208331-201208331 on Build GRCh38
rs148867428 Chr.2: 201208558-201208558 on Build GRCh38
rs114959391 Chr.2: 201208435-201208435 on Build GRCh38
rs41419046 Chr.2: 201208250-201208250 on Build GRCh38
rs200221261 Chr.2: 201208184-201208184 on Build GRCh38
rs751757781 Chr.2: 201208283-201208283 on Build GRCh38
rs767555790 Chr.2: 201208075-201208075 on Build GRCh38
rs754175962 Chr.2: 201208541-201208541 on Build GRCh38
rs957577647 Chr.2: 201208441-201208441 on Build GRCh38
rs17860403 Chr.2: 201208114-201208114 on Build GRCh38
rs773105887 Chr.2: 201208194-201208194 on Build GRCh38
rs958605793 Chr.2: 201208329-201208329 on Build GRCh38
rs146654699 Chr.2: 201208148-201208148 on Build GRCh38
rs971449971 Chr.2: 201208388-201208388 on Build GRCh38
rs768371369 Chr.2: 201208170-201208170 on Build GRCh38
rs373189673 Chr.2: 201208206-201208206 on Build GRCh38
rs759691578 Chr.2: 201208381-201208381 on Build GRCh38
rs958667368 Chr.2: 201208091-201208091 on Build GRCh38
rs182849481 Chr.2: 201208474-201208474 on Build GRCh38
rs17672977 Chr.2: 201208509-201208509 on Build GRCh38
rs111719534 Chr.2: 201208117-201208117 on Build GRCh38
rs372906837 Chr.2: 201208107-201208107 on Build GRCh38
rs1012064973 Chr.2: 201208386-201208386 on Build GRCh38
rs774173689 Chr.2: 201208177-201208177 on Build GRCh38
rs767575167 Chr.2: 201208391-201208391 on Build GRCh38
rs540207844 Chr.2: 201208158-201208158 on Build GRCh38
rs745398864 Chr.2: 201208141-201208141 on Build GRCh38
rs149912574 Chr.2: 201208174-201208174 on Build GRCh38
rs886067197 Chr.2: 201208289-201208289 on Build GRCh38
rs750581097 Chr.2: 201208100-201208100 on Build GRCh38
rs369897442 Chr.2: 201208140-201208140 on Build GRCh38
rs991257476 Chr.2: 201208106-201208106 on Build GRCh38
rs753572822 Chr.2: 201208209-201208209 on Build GRCh38
rs895097193 Chr.2: 201208374-201208374 on Build GRCh38

View Full Product Details