Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201807416-201807926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK15
Entrez Gene ID
65061
Gene Name
cyclin dependent kinase 15
Gene Aliases
ALS2CR7, PFTAIRE2, PFTK2
UniGene
Hs.348711
Gene Chromosome Location
Chr.2: 201790324-201895550 on Build GRCh38
Transcripts
NM_001261435.1 NM_001261436.1 NM_139158.2 XM_005246781.3 XM_005246782.4 XM_011511650.2 XM_011511652.1 XM_011511654.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2907731 c.220C>T p.R74W Chr.2: 201807590-201807590 on Build GRCh38
1614156 c.80A>G p.D27G Chr.2: 201807603-201807603 on Build GRCh38
4671110 c.191G>A p.R64H Chr.2: 201807561-201807561 on Build GRCh38
6156573 c.312G>T p.L104F Chr.2: 201807896-201807896 on Build GRCh38
5861510 c.120+1G>A p.? Chr.2: 201807644-201807644 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767765738 Chr.2: 201807629-201807629 on Build GRCh38
rs1042987895 Chr.2: 201807745-201807745 on Build GRCh38
rs373974199 Chr.2: 201807829-201807829 on Build GRCh38
rs967874570 Chr.2: 201807514-201807514 on Build GRCh38
rs777340501 Chr.2: 201807461-201807461 on Build GRCh38
rs371388573 Chr.2: 201807666-201807666 on Build GRCh38
rs377366156 Chr.2: 201807538-201807538 on Build GRCh38
rs569840064 Chr.2: 201807507-201807507 on Build GRCh38
rs760090280 Chr.2: 201807476-201807479 on Build GRCh38
rs776537698 Chr.2: 201807681-201807681 on Build GRCh38
rs758944433 Chr.2: 201807466-201807466 on Build GRCh38
rs766895293 Chr.2: 201807598-201807599 on Build GRCh38
rs759851495 Chr.2: 201807462-201807462 on Build GRCh38
rs778091053 Chr.2: 201807490-201807490 on Build GRCh38
rs774553691 Chr.2: 201807804-201807804 on Build GRCh38
rs768959454 Chr.2: 201807888-201807888 on Build GRCh38
rs913071578 Chr.2: 201807720-201807720 on Build GRCh38
rs747832628 Chr.2: 201807658-201807658 on Build GRCh38
rs193603 Chr.2: 201807806-201807806 on Build GRCh38
rs764238297 Chr.2: 201807605-201807605 on Build GRCh38
rs1038980317 Chr.2: 201807695-201807695 on Build GRCh38
rs747406662 Chr.2: 201807774-201807774 on Build GRCh38
rs752276525 Chr.2: 201807668-201807670 on Build GRCh38
rs368340552 Chr.2: 201807665-201807665 on Build GRCh38
rs142237246 Chr.2: 201807523-201807523 on Build GRCh38
rs746974802 Chr.2: 201807543-201807543 on Build GRCh38
rs1051456255 Chr.2: 201807793-201807794 on Build GRCh38
rs896171307 Chr.2: 201807731-201807731 on Build GRCh38
rs781234061 Chr.2: 201807852-201807852 on Build GRCh38
rs926792005 Chr.2: 201807703-201807703 on Build GRCh38
rs763085191 Chr.2: 201807601-201807601 on Build GRCh38
rs997863043 Chr.2: 201807870-201807870 on Build GRCh38
rs748922435 Chr.2: 201807509-201807509 on Build GRCh38
rs200245595 Chr.2: 201807519-201807519 on Build GRCh38
rs77557988 Chr.2: 201807887-201807887 on Build GRCh38
rs765394945 Chr.2: 201807463-201807463 on Build GRCh38
rs570865727 Chr.2: 201807724-201807724 on Build GRCh38
rs758127460 Chr.2: 201807837-201807837 on Build GRCh38
rs373141817 Chr.2: 201807803-201807803 on Build GRCh38
rs548746565 Chr.2: 201807573-201807573 on Build GRCh38
rs766232066 Chr.2: 201807452-201807452 on Build GRCh38
rs757496592 Chr.2: 201807506-201807506 on Build GRCh38
rs201632234 Chr.2: 201807591-201807591 on Build GRCh38
rs746213688 Chr.2: 201807464-201807464 on Build GRCh38
rs769653908 Chr.2: 201807879-201807879 on Build GRCh38
rs200222460 Chr.2: 201807659-201807659 on Build GRCh38
rs964549232 Chr.2: 201807834-201807834 on Build GRCh38
rs745709326 Chr.2: 201807682-201807682 on Build GRCh38
rs770969647 Chr.2: 201807672-201807672 on Build GRCh38
rs369713551 Chr.2: 201807446-201807446 on Build GRCh38
rs201753010 Chr.2: 201807840-201807840 on Build GRCh38
rs1021459389 Chr.2: 201807473-201807473 on Build GRCh38
rs754599551 Chr.2: 201807644-201807644 on Build GRCh38
rs1009198363 Chr.2: 201807755-201807755 on Build GRCh38
rs879067602 Chr.2: 201807457-201807457 on Build GRCh38
rs780051899 Chr.2: 201807883-201807883 on Build GRCh38
rs955385484 Chr.2: 201807889-201807889 on Build GRCh38
rs373450112 Chr.2: 201807503-201807503 on Build GRCh38
rs756433667 Chr.2: 201807814-201807814 on Build GRCh38
rs369158011 Chr.2: 201807502-201807502 on Build GRCh38
rs941448846 Chr.2: 201807686-201807686 on Build GRCh38
rs757121648 Chr.2: 201807847-201807847 on Build GRCh38
rs759835104 Chr.2: 201807561-201807561 on Build GRCh38
rs1056689497 Chr.2: 201807715-201807715 on Build GRCh38
rs996268314 Chr.2: 201807779-201807779 on Build GRCh38
rs556099680 Chr.2: 201807795-201807795 on Build GRCh38
rs938289954 Chr.2: 201807704-201807704 on Build GRCh38
rs745803504 Chr.2: 201807854-201807854 on Build GRCh38
rs769740847 Chr.2: 201807690-201807690 on Build GRCh38
rs771636326 Chr.2: 201807530-201807530 on Build GRCh38
rs757836967 Chr.2: 201807620-201807620 on Build GRCh38
rs766738042 Chr.2: 201807816-201807816 on Build GRCh38
rs1005013404 Chr.2: 201807557-201807557 on Build GRCh38
rs34776344 Chr.2: 201807560-201807560 on Build GRCh38
rs201182481 Chr.2: 201807653-201807653 on Build GRCh38
rs982783078 Chr.2: 201807890-201807890 on Build GRCh38
rs747703696 Chr.2: 201807525-201807525 on Build GRCh38
rs900413689 Chr.2: 201807758-201807758 on Build GRCh38
rs138441846 Chr.2: 201807891-201807891 on Build GRCh38
rs750823228 Chr.2: 201807641-201807641 on Build GRCh38
rs772035580 Chr.2: 201807444-201807444 on Build GRCh38
rs746569767 Chr.2: 201807842-201807842 on Build GRCh38
rs768115412 Chr.2: 201807809-201807809 on Build GRCh38

View Full Product Details