Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201854733-201855255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK15
Entrez Gene ID
65061
Gene Name
cyclin dependent kinase 15
Gene Aliases
ALS2CR7, PFTAIRE2, PFTK2
UniGene
Hs.348711
Gene Chromosome Location
Chr.2: 201790324-201895550 on Build GRCh38
Transcripts
NM_001261435.1 NM_001261436.1 NM_139158.2 XM_005246781.3 XM_005246782.4 XM_011511650.2 XM_011511652.1 XM_011511654.2 XM_011511655.2 XM_011511656.2 XM_011511657.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6625410 c.956T>C p.V319A Chr.2: 201854884-201854884 on Build GRCh38
7360651 c.836A>T p.K279M Chr.2: 201854917-201854917 on Build GRCh38
6625403 c.849C>T p.Y283Y Chr.2: 201854930-201854930 on Build GRCh38
3962184 c.946-7T>C p.? Chr.2: 201854867-201854867 on Build GRCh38
7360652 c.989A>T p.K330M Chr.2: 201854917-201854917 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561188601 Chr.2: 201854770-201854770 on Build GRCh38
rs780484846 Chr.2: 201855184-201855184 on Build GRCh38
rs754401995 Chr.2: 201854938-201854938 on Build GRCh38
rs1040383352 Chr.2: 201855165-201855165 on Build GRCh38
rs550695496 Chr.2: 201854927-201854927 on Build GRCh38
rs750958888 Chr.2: 201854851-201854851 on Build GRCh38
rs981092591 Chr.2: 201855058-201855058 on Build GRCh38
rs933086700 Chr.2: 201855144-201855144 on Build GRCh38
rs368326843 Chr.2: 201854988-201854988 on Build GRCh38
rs180872991 Chr.2: 201855033-201855033 on Build GRCh38
rs768045931 Chr.2: 201854865-201854866 on Build GRCh38
rs775432947 Chr.2: 201854899-201854899 on Build GRCh38
rs893997352 Chr.2: 201855128-201855128 on Build GRCh38
rs925549967 Chr.2: 201855052-201855052 on Build GRCh38
rs747522548 Chr.2: 201855186-201855186 on Build GRCh38
rs921594169 Chr.2: 201855139-201855139 on Build GRCh38
rs777481168 Chr.2: 201854866-201854866 on Build GRCh38
rs900281973 Chr.2: 201854773-201854773 on Build GRCh38
rs755424972 Chr.2: 201854873-201854873 on Build GRCh38
rs886232315 Chr.2: 201854869-201854869 on Build GRCh38
rs779394508 Chr.2: 201854968-201854968 on Build GRCh38
rs992615179 Chr.2: 201855001-201855001 on Build GRCh38
rs370311295 Chr.2: 201854838-201854838 on Build GRCh38
rs554968045 Chr.2: 201855034-201855034 on Build GRCh38
rs762160050 Chr.2: 201854912-201854912 on Build GRCh38
rs752278791 Chr.2: 201854763-201854763 on Build GRCh38
rs762701067 Chr.2: 201854900-201854900 on Build GRCh38
rs367719089 Chr.2: 201854903-201854903 on Build GRCh38
rs377762320 Chr.2: 201854906-201854906 on Build GRCh38
rs149899531 Chr.2: 201854993-201854993 on Build GRCh38
rs779117119 Chr.2: 201854858-201854858 on Build GRCh38
rs924246800 Chr.2: 201855061-201855061 on Build GRCh38
rs547137967 Chr.2: 201854824-201854824 on Build GRCh38
rs114195966 Chr.2: 201855215-201855215 on Build GRCh38
rs886286939 Chr.2: 201855205-201855205 on Build GRCh38
rs73055701 Chr.2: 201854792-201854792 on Build GRCh38
rs776273369 Chr.2: 201854886-201854886 on Build GRCh38
rs753292347 Chr.2: 201854894-201854906 on Build GRCh38
rs1004263000 Chr.2: 201855217-201855217 on Build GRCh38
rs756725510 Chr.2: 201854854-201854854 on Build GRCh38
rs1025151255 Chr.2: 201855011-201855011 on Build GRCh38
rs185762408 Chr.2: 201855204-201855204 on Build GRCh38
rs769775034 Chr.2: 201854898-201854898 on Build GRCh38
rs199639155 Chr.2: 201854874-201854874 on Build GRCh38
rs974439476 Chr.2: 201855085-201855085 on Build GRCh38
rs746688089 Chr.2: 201854875-201854875 on Build GRCh38
rs745777451 Chr.2: 201854891-201854891 on Build GRCh38
rs899868300 Chr.2: 201854816-201854816 on Build GRCh38
rs991849636 Chr.2: 201854974-201854974 on Build GRCh38
rs371256915 Chr.2: 201854932-201854932 on Build GRCh38
rs375313182 Chr.2: 201854967-201854967 on Build GRCh38
rs781387700 Chr.2: 201854843-201854843 on Build GRCh38
rs936894892 Chr.2: 201855055-201855055 on Build GRCh38
rs749620772 Chr.2: 201854870-201854870 on Build GRCh38
rs1054175878 Chr.2: 201855084-201855084 on Build GRCh38
rs528295236 Chr.2: 201854777-201854777 on Build GRCh38
rs377132693 Chr.2: 201854830-201854830 on Build GRCh38
rs142560911 Chr.2: 201854879-201854879 on Build GRCh38
rs1053047620 Chr.2: 201854756-201854756 on Build GRCh38
rs1033625521 Chr.2: 201854928-201854928 on Build GRCh38
rs900080303 Chr.2: 201855178-201855178 on Build GRCh38
rs756977520 Chr.2: 201854980-201854980 on Build GRCh38
rs934418101 Chr.2: 201855074-201855074 on Build GRCh38
rs1051915227 Chr.2: 201855200-201855200 on Build GRCh38
rs1016105643 Chr.2: 201854882-201854882 on Build GRCh38
rs746638111 Chr.2: 201854975-201854975 on Build GRCh38
rs746192131 Chr.2: 201854936-201854936 on Build GRCh38
rs757221913 Chr.2: 201854839-201854839 on Build GRCh38
rs571449252 Chr.2: 201854806-201854806 on Build GRCh38
rs763959488 Chr.2: 201854905-201854905 on Build GRCh38
rs532275660 Chr.2: 201854818-201854818 on Build GRCh38
rs539318970 Chr.2: 201855232-201855232 on Build GRCh38

View Full Product Details