Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201872176-201872672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK15
Entrez Gene ID
65061
Gene Name
cyclin dependent kinase 15
Gene Aliases
ALS2CR7, PFTAIRE2, PFTK2
UniGene
Hs.348711
Gene Chromosome Location
Chr.2: 201790324-201895550 on Build GRCh38
Transcripts
NM_001261435.1 NM_001261436.1 NM_139158.2 XM_005246781.3 XM_005246782.4 XM_011511650.2 XM_011511652.1 XM_011511655.2 XM_011511656.2 XM_011511657.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7326145 c.875C>T p.T292M Chr.2: 201872296-201872296 on Build GRCh38
6156569 c.1058+1G>T p.? Chr.2: 201872327-201872327 on Build GRCh38
7326146 c.1028C>T p.T343M Chr.2: 201872296-201872296 on Build GRCh38
4090280 c.1033C>T p.R345* Chr.2: 201872301-201872301 on Build GRCh38
6090053 c.857-1G>A p.? Chr.2: 201872277-201872277 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368174315 Chr.2: 201872297-201872297 on Build GRCh38
rs778526553 Chr.2: 201872357-201872358 on Build GRCh38
rs762097278 Chr.2: 201872355-201872355 on Build GRCh38
rs1027416643 Chr.2: 201872436-201872436 on Build GRCh38
rs781412173 Chr.2: 201872327-201872327 on Build GRCh38
rs1006714885 Chr.2: 201872631-201872631 on Build GRCh38
rs146077160 Chr.2: 201872402-201872402 on Build GRCh38
rs764086373 Chr.2: 201872376-201872376 on Build GRCh38
rs929343511 Chr.2: 201872433-201872433 on Build GRCh38
rs570755737 Chr.2: 201872483-201872483 on Build GRCh38
rs370382391 Chr.2: 201872301-201872301 on Build GRCh38
rs116483098 Chr.2: 201872457-201872457 on Build GRCh38
rs1047888870 Chr.2: 201872285-201872285 on Build GRCh38
rs184103705 Chr.2: 201872598-201872598 on Build GRCh38
rs368742674 Chr.2: 201872257-201872257 on Build GRCh38
rs576725269 Chr.2: 201872374-201872374 on Build GRCh38
rs13419220 Chr.2: 201872365-201872365 on Build GRCh38
rs983633103 Chr.2: 201872438-201872438 on Build GRCh38
rs757153144 Chr.2: 201872324-201872324 on Build GRCh38
rs1042766872 Chr.2: 201872603-201872603 on Build GRCh38
rs140412753 Chr.2: 201872344-201872354 on Build GRCh38
rs1035442017 Chr.2: 201872321-201872321 on Build GRCh38
rs767786132 Chr.2: 201872356-201872356 on Build GRCh38
rs549338953 Chr.2: 201872295-201872295 on Build GRCh38
rs386654233 Chr.2: 201872354-201872365 on Build GRCh38
rs1010032423 Chr.2: 201872328-201872328 on Build GRCh38
rs140027574 Chr.2: 201872581-201872581 on Build GRCh38
rs185229780 Chr.2: 201872258-201872258 on Build GRCh38
rs891559886 Chr.2: 201872578-201872578 on Build GRCh38
rs755801588 Chr.2: 201872227-201872227 on Build GRCh38
rs777778319 Chr.2: 201872240-201872240 on Build GRCh38
rs375122798 Chr.2: 201872296-201872296 on Build GRCh38
rs200862859 Chr.2: 201872236-201872236 on Build GRCh38
rs926078568 Chr.2: 201872629-201872629 on Build GRCh38
rs1002530733 Chr.2: 201872306-201872306 on Build GRCh38
rs889051087 Chr.2: 201872410-201872410 on Build GRCh38
rs769099964 Chr.2: 201872351-201872351 on Build GRCh38
rs771329961 Chr.2: 201872243-201872243 on Build GRCh38
rs1024765475 Chr.2: 201872491-201872491 on Build GRCh38
rs1018176515 Chr.2: 201872405-201872405 on Build GRCh38
rs747204620 Chr.2: 201872241-201872241 on Build GRCh38
rs746754824 Chr.2: 201872312-201872312 on Build GRCh38
rs535256477 Chr.2: 201872342-201872342 on Build GRCh38
rs925812141 Chr.2: 201872368-201872368 on Build GRCh38
rs181365974 Chr.2: 201872576-201872576 on Build GRCh38
rs764712442 Chr.2: 201872300-201872300 on Build GRCh38
rs752199302 Chr.2: 201872373-201872373 on Build GRCh38
rs73057628 Chr.2: 201872430-201872430 on Build GRCh38
rs552212214 Chr.2: 201872471-201872471 on Build GRCh38
rs35273085 Chr.2: 201872230-201872230 on Build GRCh38
rs13032979 Chr.2: 201872466-201872466 on Build GRCh38
rs374280027 Chr.2: 201872363-201872365 on Build GRCh38
rs1040469523 Chr.2: 201872423-201872423 on Build GRCh38
rs531478255 Chr.2: 201872527-201872527 on Build GRCh38
rs374734881 Chr.2: 201872239-201872239 on Build GRCh38
rs13032025 Chr.2: 201872360-201872360 on Build GRCh38
rs370770162 Chr.2: 201872421-201872421 on Build GRCh38
rs372801952 Chr.2: 201872354-201872354 on Build GRCh38
rs562245939 Chr.2: 201872403-201872403 on Build GRCh38
rs904382093 Chr.2: 201872608-201872608 on Build GRCh38
rs201605804 Chr.2: 201872293-201872293 on Build GRCh38
rs201971784 Chr.2: 201872353-201872360 on Build GRCh38
rs376643967 Chr.2: 201872259-201872259 on Build GRCh38
rs751966765 Chr.2: 201872282-201872282 on Build GRCh38
rs201917344 Chr.2: 201872237-201872237 on Build GRCh38
rs370819607 Chr.2: 201872280-201872280 on Build GRCh38
rs775468611 Chr.2: 201872337-201872337 on Build GRCh38
rs199955601 Chr.2: 201872289-201872289 on Build GRCh38
rs1045072714 Chr.2: 201872470-201872470 on Build GRCh38
rs760254108 Chr.2: 201872244-201872244 on Build GRCh38
rs866789442 Chr.2: 201872508-201872508 on Build GRCh38
rs571966218 Chr.2: 201872302-201872302 on Build GRCh38
rs759178832 Chr.2: 201872367-201872367 on Build GRCh38
rs549609984 Chr.2: 201872535-201872535 on Build GRCh38
rs758451719 Chr.2: 201872303-201872303 on Build GRCh38
rs112648341 Chr.2: 201872479-201872479 on Build GRCh38
rs887483704 Chr.2: 201872449-201872449 on Build GRCh38
rs764774758 Chr.2: 201872372-201872372 on Build GRCh38
rs1056690517 Chr.2: 201872534-201872534 on Build GRCh38
rs377418969 Chr.2: 201872264-201872264 on Build GRCh38
rs570560555 Chr.2: 201872208-201872208 on Build GRCh38
rs777690518 Chr.2: 201872305-201872305 on Build GRCh38
rs923355091 Chr.2: 201872431-201872431 on Build GRCh38
rs763752662 Chr.2: 201872292-201872292 on Build GRCh38
rs188954760 Chr.2: 201872336-201872336 on Build GRCh38

View Full Product Details