Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 202464554-202465065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMPR2
Entrez Gene ID
659
Gene Name
bone morphogenetic protein receptor type 2
Gene Aliases
BMPR-II, BMPR3, BMR2, BRK-3, POVD1, PPH1, T-ALK
UniGene
Hs.471119
Gene Chromosome Location
Chr.2: 202376310-202567751 on Build GRCh38
Transcripts
NM_001204.6 XM_011511687.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6156563 c.181T>A p.S61T Chr.2: 202464913-202464913 on Build GRCh38
4188939 c.193delA p.T65fs*13 Chr.2: 202464925-202464925 on Build GRCh38
6612043 c.152_153delTC p.H53fs*1 Chr.2: 202464884-202464885 on Build GRCh38
6612045 c.246A>G p.Q82Q Chr.2: 202464978-202464978 on Build GRCh38
4400424 c.224G>A p.G75E Chr.2: 202464956-202464956 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1016065850 Chr.2: 202464970-202464970 on Build GRCh38
rs137852755 Chr.2: 202464852-202464852 on Build GRCh38
rs557567077 Chr.2: 202464656-202464656 on Build GRCh38
rs775715377 Chr.2: 202465018-202465018 on Build GRCh38
rs144389360 Chr.2: 202464962-202464962 on Build GRCh38
rs763770030 Chr.2: 202465010-202465011 on Build GRCh38
rs752139986 Chr.2: 202464951-202464951 on Build GRCh38
rs1027583541 Chr.2: 202464601-202464601 on Build GRCh38
rs200976092 Chr.2: 202464897-202464897 on Build GRCh38
rs757855256 Chr.2: 202464952-202464952 on Build GRCh38
rs992934350 Chr.2: 202464720-202464720 on Build GRCh38
rs777451350 Chr.2: 202464956-202464956 on Build GRCh38
rs767827692 Chr.2: 202464919-202464919 on Build GRCh38
rs759804753 Chr.2: 202464868-202464868 on Build GRCh38
rs753499293 Chr.2: 202465011-202465011 on Build GRCh38
rs760290506 Chr.2: 202464613-202464615 on Build GRCh38
rs770804811 Chr.2: 202464848-202464848 on Build GRCh38
rs764721647 Chr.2: 202464779-202464779 on Build GRCh38
rs754688935 Chr.2: 202464816-202464816 on Build GRCh38
rs745717516 Chr.2: 202464987-202464987 on Build GRCh38
rs768669785 Chr.2: 202464596-202464596 on Build GRCh38
rs147207234 Chr.2: 202464813-202464813 on Build GRCh38
rs762330274 Chr.2: 202464915-202464915 on Build GRCh38
rs770128128 Chr.2: 202464836-202464836 on Build GRCh38
rs757551343 Chr.2: 202464795-202464795 on Build GRCh38
rs769905466 Chr.2: 202464994-202464994 on Build GRCh38
rs763097991 Chr.2: 202465024-202465024 on Build GRCh38
rs969758661 Chr.2: 202464654-202464654 on Build GRCh38
rs750825305 Chr.2: 202464924-202464924 on Build GRCh38
rs112862820 Chr.2: 202464818-202464818 on Build GRCh38
rs543261516 Chr.2: 202464708-202464708 on Build GRCh38
rs137852742 Chr.2: 202464950-202464950 on Build GRCh38
rs201759998 Chr.2: 202464882-202464882 on Build GRCh38
rs752864146 Chr.2: 202464765-202464765 on Build GRCh38
rs746943205 Chr.2: 202464837-202464837 on Build GRCh38
rs376957186 Chr.2: 202464770-202464770 on Build GRCh38
rs771035488 Chr.2: 202464967-202464967 on Build GRCh38
rs963593230 Chr.2: 202464581-202464581 on Build GRCh38
rs980048183 Chr.2: 202464661-202464661 on Build GRCh38
rs747740309 Chr.2: 202464821-202464821 on Build GRCh38
rs886039223 Chr.2: 202464907-202464908 on Build GRCh38
rs754561255 Chr.2: 202464934-202464934 on Build GRCh38
rs573463511 Chr.2: 202464812-202464812 on Build GRCh38
rs1051805297 Chr.2: 202464887-202464887 on Build GRCh38
rs752005716 Chr.2: 202464906-202464906 on Build GRCh38
rs368430622 Chr.2: 202464827-202464827 on Build GRCh38
rs988297260 Chr.2: 202464733-202464733 on Build GRCh38
rs926929921 Chr.2: 202464680-202464680 on Build GRCh38
rs778682407 Chr.2: 202464944-202464944 on Build GRCh38
rs912963620 Chr.2: 202464751-202464751 on Build GRCh38
rs375690035 Chr.2: 202464757-202464757 on Build GRCh38
rs200036340 Chr.2: 202465011-202465011 on Build GRCh38
rs863223423 Chr.2: 202464918-202464919 on Build GRCh38
rs751917686 Chr.2: 202464782-202464782 on Build GRCh38
rs776603012 Chr.2: 202464849-202464849 on Build GRCh38
rs781115210 Chr.2: 202464986-202464986 on Build GRCh38
rs561600960 Chr.2: 202464734-202464734 on Build GRCh38
rs575944935 Chr.2: 202464665-202464665 on Build GRCh38
rs945836014 Chr.2: 202464788-202464788 on Build GRCh38
rs771887212 Chr.2: 202464826-202464826 on Build GRCh38
rs150080314 Chr.2: 202464879-202464879 on Build GRCh38

View Full Product Details