Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 206162409-206162906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEF1B2
Entrez Gene ID
1933
Gene Name
eukaryotic translation elongation factor 1 beta 2
Gene Aliases
EEF1B, EEF1B1, EF1B
UniGene
Hs.421608
Gene Chromosome Location
Chr.2: 206159594-206162929 on Build GRCh38
Transcripts
NM_001037663.1 NM_001959.3 NM_021121.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1015306 c.468G>A p.A156A Chr.2: 206162559-206162559 on Build GRCh38
5777691 c.475G>A p.E159K Chr.2: 206162566-206162566 on Build GRCh38
1015307 c.498A>C p.Q166H Chr.2: 206162589-206162589 on Build GRCh38
7300330 c.626C>T p.A209V Chr.2: 206162831-206162831 on Build GRCh38
4885338 c.665T>A p.F222Y Chr.2: 206162870-206162870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1022744069 Chr.2: 206162765-206162765 on Build GRCh38
rs911730164 Chr.2: 206162587-206162587 on Build GRCh38
rs753393059 Chr.2: 206162604-206162604 on Build GRCh38
rs371039052 Chr.2: 206162739-206162739 on Build GRCh38
rs868231442 Chr.2: 206162684-206162684 on Build GRCh38
rs759689793 Chr.2: 206162561-206162561 on Build GRCh38
rs367579304 Chr.2: 206162873-206162873 on Build GRCh38
rs746171124 Chr.2: 206162845-206162845 on Build GRCh38
rs774742946 Chr.2: 206162441-206162442 on Build GRCh38
rs751944979 Chr.2: 206162584-206162584 on Build GRCh38
rs377381799 Chr.2: 206162590-206162590 on Build GRCh38
rs766564190 Chr.2: 206162558-206162558 on Build GRCh38
rs759926959 Chr.2: 206162513-206162514 on Build GRCh38
rs970207541 Chr.2: 206162717-206162717 on Build GRCh38
rs746878595 Chr.2: 206162809-206162809 on Build GRCh38
rs764623938 Chr.2: 206162582-206162582 on Build GRCh38
rs889571248 Chr.2: 206162492-206162492 on Build GRCh38
rs182342432 Chr.2: 206162748-206162748 on Build GRCh38
rs147791783 Chr.2: 206162559-206162559 on Build GRCh38
rs774378420 Chr.2: 206162499-206162499 on Build GRCh38
rs956192383 Chr.2: 206162669-206162669 on Build GRCh38
rs1016954212 Chr.2: 206162752-206162752 on Build GRCh38
rs371429685 Chr.2: 206162622-206162622 on Build GRCh38
rs199531068 Chr.2: 206162456-206162456 on Build GRCh38
rs746825673 Chr.2: 206162644-206162644 on Build GRCh38
rs760874509 Chr.2: 206162441-206162441 on Build GRCh38
rs753100752 Chr.2: 206162472-206162472 on Build GRCh38
rs745542472 Chr.2: 206162694-206162695 on Build GRCh38
rs748182302 Chr.2: 206162626-206162626 on Build GRCh38
rs778273035 Chr.2: 206162623-206162623 on Build GRCh38
rs201807235 Chr.2: 206162737-206162737 on Build GRCh38
rs767926956 Chr.2: 206162705-206162705 on Build GRCh38
rs748811604 Chr.2: 206162494-206162494 on Build GRCh38
rs755489934 Chr.2: 206162460-206162460 on Build GRCh38
rs78028628 Chr.2: 206162683-206162683 on Build GRCh38
rs778229183 Chr.2: 206162477-206162477 on Build GRCh38
rs191829310 Chr.2: 206162617-206162617 on Build GRCh38
rs181533336 Chr.2: 206162475-206162475 on Build GRCh38
rs759614861 Chr.2: 206162665-206162665 on Build GRCh38
rs564396403 Chr.2: 206162523-206162523 on Build GRCh38
rs780839827 Chr.2: 206162823-206162823 on Build GRCh38
rs375707487 Chr.2: 206162448-206162448 on Build GRCh38
rs200903337 Chr.2: 206162704-206162704 on Build GRCh38
rs756725932 Chr.2: 206162646-206162649 on Build GRCh38
rs1035207797 Chr.2: 206162437-206162437 on Build GRCh38
rs1056996351 Chr.2: 206162693-206162694 on Build GRCh38
rs531453424 Chr.2: 206162647-206162647 on Build GRCh38
rs765160465 Chr.2: 206162562-206162562 on Build GRCh38
rs568250532 Chr.2: 206162687-206162687 on Build GRCh38
rs760925547 Chr.2: 206162554-206162554 on Build GRCh38
rs779876137 Chr.2: 206162598-206162598 on Build GRCh38
rs753151319 Chr.2: 206162632-206162633 on Build GRCh38
rs758821800 Chr.2: 206162575-206162575 on Build GRCh38
rs898818496 Chr.2: 206162835-206162835 on Build GRCh38
rs762085449 Chr.2: 206162434-206162434 on Build GRCh38
rs200556695 Chr.2: 206162876-206162876 on Build GRCh38
rs757852075 Chr.2: 206162782-206162782 on Build GRCh38
rs771959213 Chr.2: 206162515-206162515 on Build GRCh38
rs767967699 Chr.2: 206162633-206162633 on Build GRCh38
rs35430253 Chr.2: 206162683-206162684 on Build GRCh38
rs374217450 Chr.2: 206162751-206162751 on Build GRCh38
rs143558392 Chr.2: 206162848-206162848 on Build GRCh38
rs201660860 Chr.2: 206162652-206162652 on Build GRCh38
rs745576823 Chr.2: 206162483-206162483 on Build GRCh38
rs918529933 Chr.2: 206162884-206162884 on Build GRCh38
rs752123768 Chr.2: 206162755-206162755 on Build GRCh38
rs200210606 Chr.2: 206162682-206162683 on Build GRCh38
rs764565826 Chr.2: 206162633-206162634 on Build GRCh38
rs2359156 Chr.2: 206162574-206162574 on Build GRCh38
rs753542531 Chr.2: 206162736-206162736 on Build GRCh38
rs974834268 Chr.2: 206162596-206162596 on Build GRCh38
rs771964313 Chr.2: 206162630-206162630 on Build GRCh38
rs199828966 Chr.2: 206162634-206162634 on Build GRCh38
rs779753319 Chr.2: 206162493-206162493 on Build GRCh38
rs747977038 Chr.2: 206162500-206162500 on Build GRCh38
rs775431732 Chr.2: 206162686-206162686 on Build GRCh38
rs773436421 Chr.2: 206162440-206162440 on Build GRCh38
rs374002370 Chr.2: 206162854-206162854 on Build GRCh38
rs145497999 Chr.2: 206162684-206162685 on Build GRCh38
rs769978460 Chr.2: 206162682-206162682 on Build GRCh38
rs777715702 Chr.2: 206162808-206162808 on Build GRCh38
rs962590472 Chr.2: 206162799-206162799 on Build GRCh38
rs200624747 Chr.2: 206162467-206162467 on Build GRCh38
rs755827613 Chr.2: 206162484-206162484 on Build GRCh38
rs770721506 Chr.2: 206162811-206162811 on Build GRCh38
rs763150217 Chr.2: 206162867-206162867 on Build GRCh38
rs752065837 Chr.2: 206162703-206162703 on Build GRCh38
rs768003462 Chr.2: 206162497-206162497 on Build GRCh38
rs767990866 Chr.2: 206162880-206162880 on Build GRCh38
rs200814967 Chr.2: 206162566-206162566 on Build GRCh38

View Full Product Details