Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 206765310-206765813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FASTKD2
Entrez Gene ID
22868
Gene Name
FAST kinase domains 2
Gene Aliases
KIAA0971
UniGene
Hs.5930
Gene Chromosome Location
Chr.2: 206765388-206868284 on Build GRCh38
Transcripts
NM_001136193.1 NM_001136194.1 NM_014929.3 NM_001039845.2 NM_001282940.1 NR_104261.1 XM_005246315.4 XM_005246316.4 XM_011510628.2 XM_017003373.1 XM_017003374.1

Gene Symbol
MDH1B
Entrez Gene ID
130752
Gene Name
malate dehydrogenase 1B
Gene Aliases
RP11-95H11
UniGene
Hs.147816
Gene Chromosome Location
Chr.2: 206737763-206765549 on Build GRCh38
Transcripts
NM_001136193.1 NM_001136194.1 NM_014929.3 NM_001039845.2 NM_001282940.1 NR_104261.1 XM_005246315.4 XM_005246316.4 XM_011510628.2 XM_017003373.1 XM_017003374.1

SNP Detail

SNP ID Chromosome Location
rs533456275 Chr.2: 206765781-206765781 on Build GRCh38
rs886055503 Chr.2: 206765578-206765578 on Build GRCh38
rs929485339 Chr.2: 206765524-206765524 on Build GRCh38
rs758262453 Chr.2: 206765342-206765342 on Build GRCh38
rs938045816 Chr.2: 206765453-206765453 on Build GRCh38
rs765644765 Chr.2: 206765412-206765412 on Build GRCh38
rs878909570 Chr.2: 206765585-206765585 on Build GRCh38
rs1021291949 Chr.2: 206765605-206765605 on Build GRCh38
rs913368429 Chr.2: 206765462-206765462 on Build GRCh38
rs537404291 Chr.2: 206765433-206765433 on Build GRCh38
rs572148469 Chr.2: 206765494-206765494 on Build GRCh38
rs897019860 Chr.2: 206765697-206765697 on Build GRCh38
rs905899164 Chr.2: 206765484-206765484 on Build GRCh38
rs577105880 Chr.2: 206765628-206765628 on Build GRCh38
rs778933831 Chr.2: 206765432-206765432 on Build GRCh38
rs952561480 Chr.2: 206765337-206765337 on Build GRCh38
rs944989447 Chr.2: 206765647-206765647 on Build GRCh38
rs912160047 Chr.2: 206765640-206765640 on Build GRCh38
rs1003483181 Chr.2: 206765729-206765729 on Build GRCh38
rs16838842 Chr.2: 206765636-206765636 on Build GRCh38
rs943725960 Chr.2: 206765539-206765539 on Build GRCh38
rs539716623 Chr.2: 206765527-206765527 on Build GRCh38
rs950334190 Chr.2: 206765471-206765471 on Build GRCh38
rs541828091 Chr.2: 206765674-206765674 on Build GRCh38
rs920967631 Chr.2: 206765654-206765654 on Build GRCh38
rs937351964 Chr.2: 206765659-206765659 on Build GRCh38
rs1009367736 Chr.2: 206765602-206765602 on Build GRCh38
rs554566682 Chr.2: 206765459-206765459 on Build GRCh38
rs770149498 Chr.2: 206765750-206765750 on Build GRCh38
rs373376547 Chr.2: 206765366-206765366 on Build GRCh38
rs546021759 Chr.2: 206765635-206765635 on Build GRCh38
rs886055504 Chr.2: 206765582-206765582 on Build GRCh38
rs562090897 Chr.2: 206765739-206765739 on Build GRCh38
rs936202010 Chr.2: 206765396-206765396 on Build GRCh38
rs758275649 Chr.2: 206765553-206765553 on Build GRCh38
rs3762567 Chr.2: 206765614-206765614 on Build GRCh38
rs1049102753 Chr.2: 206765665-206765665 on Build GRCh38
rs553890149 Chr.2: 206765446-206765446 on Build GRCh38
rs893663694 Chr.2: 206765582-206765582 on Build GRCh38
rs926711358 Chr.2: 206765394-206765394 on Build GRCh38
rs1053538219 Chr.2: 206765557-206765557 on Build GRCh38
rs973931889 Chr.2: 206765634-206765634 on Build GRCh38
rs16838839 Chr.2: 206765369-206765369 on Build GRCh38
rs998424298 Chr.2: 206765706-206765706 on Build GRCh38
rs982068225 Chr.2: 206765382-206765382 on Build GRCh38
rs970608568 Chr.2: 206765608-206765608 on Build GRCh38
rs369595078 Chr.2: 206765518-206765518 on Build GRCh38
rs889713413 Chr.2: 206765730-206765730 on Build GRCh38
rs899496110 Chr.2: 206765550-206765550 on Build GRCh38
rs1042192728 Chr.2: 206765693-206765693 on Build GRCh38
rs973497427 Chr.2: 206765477-206765477 on Build GRCh38
rs1029564199 Chr.2: 206765722-206765722 on Build GRCh38
rs116297724 Chr.2: 206765747-206765747 on Build GRCh38
rs41272657 Chr.2: 206765547-206765547 on Build GRCh38
rs778133012 Chr.2: 206765591-206765591 on Build GRCh38
rs1033590244 Chr.2: 206765745-206765745 on Build GRCh38
rs575976614 Chr.2: 206765638-206765638 on Build GRCh38
rs959359971 Chr.2: 206765626-206765626 on Build GRCh38
rs906348634 Chr.2: 206765703-206765703 on Build GRCh38
rs1039243027 Chr.2: 206765648-206765648 on Build GRCh38
rs1009830930 Chr.2: 206765689-206765689 on Build GRCh38
rs1036819915 Chr.2: 206765696-206765696 on Build GRCh38
rs181391209 Chr.2: 206765406-206765406 on Build GRCh38
rs543042440 Chr.2: 206765668-206765668 on Build GRCh38
rs912107621 Chr.2: 206765531-206765531 on Build GRCh38
rs547709931 Chr.2: 206765353-206765353 on Build GRCh38
rs866957815 Chr.2: 206765617-206765617 on Build GRCh38
rs932327028 Chr.2: 206765660-206765660 on Build GRCh38
rs576495313 Chr.2: 206765639-206765639 on Build GRCh38
rs957877731 Chr.2: 206765357-206765357 on Build GRCh38
rs1049195196 Chr.2: 206765513-206765513 on Build GRCh38
rs386654479 Chr.2: 206765366-206765369 on Build GRCh38
rs781167145 Chr.2: 206765686-206765686 on Build GRCh38
rs551853750 Chr.2: 206765378-206765378 on Build GRCh38
rs1015955024 Chr.2: 206765346-206765346 on Build GRCh38
rs1020115171 Chr.2: 206765574-206765574 on Build GRCh38
rs985209901 Chr.2: 206765376-206765376 on Build GRCh38

View Full Product Details