Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 206771018-206771500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FASTKD2
Entrez Gene ID
22868
Gene Name
FAST kinase domains 2
Gene Aliases
KIAA0971
UniGene
Hs.5930
Gene Chromosome Location
Chr.2: 206765388-206868284 on Build GRCh38
Transcripts
NM_001136193.1 NM_001136194.1 NM_014929.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6090380 c.913G>C p.E305Q Chr.2: 206771213-206771213 on Build GRCh38
6560413 c.892G>A p.D298N Chr.2: 206771192-206771192 on Build GRCh38
6543881 c.942G>T p.M314I Chr.2: 206771242-206771242 on Build GRCh38
2714324 c.981G>C p.R327S Chr.2: 206771281-206771281 on Build GRCh38
6660702 c.945G>A p.K315K Chr.2: 206771245-206771245 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767784703 Chr.2: 206771308-206771308 on Build GRCh38
rs780122031 Chr.2: 206771314-206771314 on Build GRCh38
rs954353909 Chr.2: 206771183-206771183 on Build GRCh38
rs78043427 Chr.2: 206771324-206771324 on Build GRCh38
rs779995188 Chr.2: 206771135-206771135 on Build GRCh38
rs760260746 Chr.2: 206771139-206771139 on Build GRCh38
rs774742926 Chr.2: 206771325-206771326 on Build GRCh38
rs751349864 Chr.2: 206771301-206771301 on Build GRCh38
rs182485101 Chr.2: 206771126-206771126 on Build GRCh38
rs756262320 Chr.2: 206771313-206771313 on Build GRCh38
rs560557343 Chr.2: 206771234-206771234 on Build GRCh38
rs983668248 Chr.2: 206771363-206771363 on Build GRCh38
rs144499152 Chr.2: 206771211-206771211 on Build GRCh38
rs768683396 Chr.2: 206771295-206771296 on Build GRCh38
rs779266866 Chr.2: 206771344-206771344 on Build GRCh38
rs577446214 Chr.2: 206771412-206771412 on Build GRCh38
rs908185876 Chr.2: 206771050-206771050 on Build GRCh38
rs536902815 Chr.2: 206771457-206771457 on Build GRCh38
rs373041286 Chr.2: 206771189-206771189 on Build GRCh38
rs751316870 Chr.2: 206771206-206771206 on Build GRCh38
rs748512485 Chr.2: 206771342-206771342 on Build GRCh38
rs1054936826 Chr.2: 206771397-206771397 on Build GRCh38
rs777438143 Chr.2: 206771199-206771199 on Build GRCh38
rs757447932 Chr.2: 206771207-206771207 on Build GRCh38
rs774852881 Chr.2: 206771407-206771407 on Build GRCh38
rs776404857 Chr.2: 206771278-206771278 on Build GRCh38
rs965383941 Chr.2: 206771390-206771390 on Build GRCh38
rs961023543 Chr.2: 206771451-206771452 on Build GRCh38
rs770533635 Chr.2: 206771266-206771266 on Build GRCh38
rs373736857 Chr.2: 206771068-206771068 on Build GRCh38
rs373864607 Chr.2: 206771146-206771146 on Build GRCh38
rs764049350 Chr.2: 206771299-206771299 on Build GRCh38
rs555291098 Chr.2: 206771467-206771467 on Build GRCh38
rs562868703 Chr.2: 206771464-206771464 on Build GRCh38
rs774603398 Chr.2: 206771247-206771247 on Build GRCh38
rs762817520 Chr.2: 206771298-206771298 on Build GRCh38
rs749341332 Chr.2: 206771318-206771318 on Build GRCh38
rs779446873 Chr.2: 206771332-206771332 on Build GRCh38
rs767682048 Chr.2: 206771181-206771181 on Build GRCh38
rs745995042 Chr.2: 206771224-206771224 on Build GRCh38
rs759087735 Chr.2: 206771159-206771159 on Build GRCh38
rs775078257 Chr.2: 206771291-206771291 on Build GRCh38
rs111452375 Chr.2: 206771057-206771057 on Build GRCh38
rs373954092 Chr.2: 206771368-206771368 on Build GRCh38
rs746901759 Chr.2: 206771179-206771180 on Build GRCh38
rs768043175 Chr.2: 206771245-206771245 on Build GRCh38
rs556911533 Chr.2: 206771450-206771450 on Build GRCh38
rs932435785 Chr.2: 206771072-206771072 on Build GRCh38
rs764593791 Chr.2: 206771066-206771066 on Build GRCh38
rs920987981 Chr.2: 206771418-206771418 on Build GRCh38
rs1027766099 Chr.2: 206771349-206771349 on Build GRCh38
rs1005387795 Chr.2: 206771419-206771419 on Build GRCh38
rs750639388 Chr.2: 206771310-206771310 on Build GRCh38
rs748507111 Chr.2: 206771265-206771265 on Build GRCh38
rs780603742 Chr.2: 206771235-206771235 on Build GRCh38
rs778000951 Chr.2: 206771298-206771299 on Build GRCh38
rs771207407 Chr.2: 206771300-206771301 on Build GRCh38
rs759179248 Chr.2: 206771279-206771279 on Build GRCh38
rs41272661 Chr.2: 206771179-206771179 on Build GRCh38
rs75607323 Chr.2: 206771046-206771046 on Build GRCh38
rs950991507 Chr.2: 206771351-206771351 on Build GRCh38
rs755471367 Chr.2: 206771319-206771319 on Build GRCh38
rs764840790 Chr.2: 206771288-206771288 on Build GRCh38
rs961115343 Chr.2: 206771075-206771075 on Build GRCh38
rs750849208 Chr.2: 206771447-206771447 on Build GRCh38
rs932484860 Chr.2: 206771437-206771437 on Build GRCh38
rs976533362 Chr.2: 206771392-206771392 on Build GRCh38
rs376799842 Chr.2: 206771148-206771148 on Build GRCh38
rs769850733 Chr.2: 206771230-206771230 on Build GRCh38
rs749764322 Chr.2: 206771244-206771244 on Build GRCh38

View Full Product Details