Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 207742184-207742722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNYL1
Entrez Gene ID
151195
Gene Name
cyclin Y like 1
Gene Aliases
-
UniGene
Hs.471234
Gene Chromosome Location
Chr.2: 207711540-207756174 on Build GRCh38
Transcripts
NM_001142300.1 NM_152523.2 XM_005246343.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6623132 c.376C>T p.R126C Chr.2: 207742289-207742289 on Build GRCh38
4090636 c.431T>C p.V144A Chr.2: 207742287-207742287 on Build GRCh38
6623131 c.433C>T p.R145C Chr.2: 207742289-207742289 on Build GRCh38
1659660 c.380C>T p.P127L Chr.2: 207742236-207742236 on Build GRCh38
4090637 c.374T>C p.V125A Chr.2: 207742287-207742287 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750163682 Chr.2: 207742384-207742384 on Build GRCh38
rs985112406 Chr.2: 207742584-207742584 on Build GRCh38
rs771879619 Chr.2: 207742290-207742290 on Build GRCh38
rs933277636 Chr.2: 207742610-207742610 on Build GRCh38
rs564772215 Chr.2: 207742421-207742421 on Build GRCh38
rs761112096 Chr.2: 207742328-207742328 on Build GRCh38
rs1016170648 Chr.2: 207742518-207742518 on Build GRCh38
rs1024704525 Chr.2: 207742481-207742481 on Build GRCh38
rs909485975 Chr.2: 207742515-207742515 on Build GRCh38
rs953687359 Chr.2: 207742558-207742558 on Build GRCh38
rs919272496 Chr.2: 207742587-207742587 on Build GRCh38
rs1005831880 Chr.2: 207742495-207742495 on Build GRCh38
rs763422093 Chr.2: 207742211-207742213 on Build GRCh38
rs371700957 Chr.2: 207742676-207742676 on Build GRCh38
rs567555056 Chr.2: 207742470-207742470 on Build GRCh38
rs781012653 Chr.2: 207742350-207742350 on Build GRCh38
rs760019377 Chr.2: 207742356-207742356 on Build GRCh38
rs190904936 Chr.2: 207742462-207742462 on Build GRCh38
rs1002212824 Chr.2: 207742485-207742485 on Build GRCh38
rs141187768 Chr.2: 207742480-207742480 on Build GRCh38
rs1029377689 Chr.2: 207742546-207742546 on Build GRCh38
rs925646780 Chr.2: 207742585-207742585 on Build GRCh38
rs186464054 Chr.2: 207742416-207742416 on Build GRCh38
rs769571440 Chr.2: 207742224-207742224 on Build GRCh38
rs148228669 Chr.2: 207742650-207742650 on Build GRCh38
rs745648520 Chr.2: 207742450-207742450 on Build GRCh38
rs766728221 Chr.2: 207742331-207742331 on Build GRCh38
rs779783067 Chr.2: 207742388-207742388 on Build GRCh38
rs773659703 Chr.2: 207742316-207742316 on Build GRCh38
rs745695508 Chr.2: 207742223-207742223 on Build GRCh38
rs913051737 Chr.2: 207742680-207742682 on Build GRCh38
rs182153988 Chr.2: 207742545-207742545 on Build GRCh38
rs201153262 Chr.2: 207742465-207742465 on Build GRCh38
rs532509984 Chr.2: 207742404-207742404 on Build GRCh38
rs779825791 Chr.2: 207742262-207742262 on Build GRCh38
rs1030665580 Chr.2: 207742429-207742429 on Build GRCh38
rs780056280 Chr.2: 207742249-207742249 on Build GRCh38
rs933412126 Chr.2: 207742240-207742240 on Build GRCh38
rs768211458 Chr.2: 207742269-207742269 on Build GRCh38
rs1055355761 Chr.2: 207742491-207742491 on Build GRCh38
rs566572557 Chr.2: 207742552-207742552 on Build GRCh38
rs756126256 Chr.2: 207742261-207742261 on Build GRCh38
rs935827252 Chr.2: 207742302-207742302 on Build GRCh38
rs969555140 Chr.2: 207742488-207742488 on Build GRCh38
rs942293868 Chr.2: 207742522-207742522 on Build GRCh38
rs777300209 Chr.2: 207742654-207742654 on Build GRCh38
rs765278741 Chr.2: 207742362-207742362 on Build GRCh38
rs1051817013 Chr.2: 207742631-207742631 on Build GRCh38
rs758580864 Chr.2: 207742380-207742380 on Build GRCh38
rs769787508 Chr.2: 207742617-207742617 on Build GRCh38
rs530748666 Chr.2: 207742458-207742458 on Build GRCh38
rs972507427 Chr.2: 207742525-207742525 on Build GRCh38
rs752815298 Chr.2: 207742363-207742363 on Build GRCh38
rs201020093 Chr.2: 207742216-207742216 on Build GRCh38
rs764323427 Chr.2: 207742383-207742383 on Build GRCh38
rs774067777 Chr.2: 207742276-207742276 on Build GRCh38
rs544900589 Chr.2: 207742437-207742439 on Build GRCh38
rs560172236 Chr.2: 207742403-207742403 on Build GRCh38
rs13397055 Chr.2: 207742285-207742285 on Build GRCh38
rs778790998 Chr.2: 207742492-207742492 on Build GRCh38

View Full Product Details