Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 208146020-208146515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYGB
Entrez Gene ID
1419
Gene Name
crystallin gamma B
Gene Aliases
CRYG2, CTRCT39
UniGene
Hs.248102
Gene Chromosome Location
Chr.2: 208142573-208146327 on Build GRCh38
Transcripts
NM_005210.3 XM_017003402.1 NR_038437.1

Gene Symbol
LOC100507443
Entrez Gene ID
100507443
Gene Name
uncharacterized LOC100507443
Gene Aliases
-
UniGene
Hs.570295
Gene Chromosome Location
Chr.2: 208119129-208156762 on Build GRCh38
Transcripts
NM_005210.3 XM_017003402.1 NR_038437.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3962261 c.9+4A>G p.? Chr.2: 208146108-208146108 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778414651 Chr.2: 208146094-208146094 on Build GRCh38
rs188633178 Chr.2: 208146100-208146100 on Build GRCh38
rs562868029 Chr.2: 208146302-208146302 on Build GRCh38
rs747376795 Chr.2: 208146049-208146050 on Build GRCh38
rs757347829 Chr.2: 208146097-208146097 on Build GRCh38
rs751613013 Chr.2: 208146096-208146096 on Build GRCh38
rs977086178 Chr.2: 208146272-208146272 on Build GRCh38
rs531884220 Chr.2: 208146329-208146329 on Build GRCh38
rs765527661 Chr.2: 208146061-208146061 on Build GRCh38
rs967019416 Chr.2: 208146383-208146383 on Build GRCh38
rs201396091 Chr.2: 208146059-208146059 on Build GRCh38
rs989873935 Chr.2: 208146374-208146374 on Build GRCh38
rs776359571 Chr.2: 208146158-208146158 on Build GRCh38
rs887082548 Chr.2: 208146263-208146263 on Build GRCh38
rs759278681 Chr.2: 208146161-208146161 on Build GRCh38
rs386654563 Chr.2: 208146099-208146101 on Build GRCh38
rs560639303 Chr.2: 208146240-208146240 on Build GRCh38
rs772502575 Chr.2: 208146351-208146351 on Build GRCh38
rs961292694 Chr.2: 208146262-208146262 on Build GRCh38
rs184696883 Chr.2: 208146183-208146183 on Build GRCh38
rs144701686 Chr.2: 208146172-208146172 on Build GRCh38
rs901715012 Chr.2: 208146328-208146328 on Build GRCh38
rs865959976 Chr.2: 208146339-208146339 on Build GRCh38
rs931358799 Chr.2: 208146179-208146179 on Build GRCh38
rs1015845949 Chr.2: 208146233-208146234 on Build GRCh38
rs748042997 Chr.2: 208146110-208146110 on Build GRCh38
rs765866161 Chr.2: 208146330-208146330 on Build GRCh38
rs759941329 Chr.2: 208146058-208146058 on Build GRCh38
rs920085142 Chr.2: 208146437-208146437 on Build GRCh38
rs1005307731 Chr.2: 208146222-208146222 on Build GRCh38
rs781349310 Chr.2: 208146098-208146098 on Build GRCh38
rs928875204 Chr.2: 208146404-208146404 on Build GRCh38
rs75781846 Chr.2: 208146148-208146148 on Build GRCh38
rs147937008 Chr.2: 208146408-208146408 on Build GRCh38
rs1004783980 Chr.2: 208146273-208146273 on Build GRCh38
rs773332685 Chr.2: 208146116-208146116 on Build GRCh38
rs2289917 Chr.2: 208146167-208146167 on Build GRCh38
rs192997184 Chr.2: 208146101-208146101 on Build GRCh38
rs985164976 Chr.2: 208146305-208146305 on Build GRCh38
rs769978238 Chr.2: 208146054-208146054 on Build GRCh38
rs931412733 Chr.2: 208146460-208146460 on Build GRCh38
rs532801298 Chr.2: 208146244-208146244 on Build GRCh38
rs188025418 Chr.2: 208146380-208146380 on Build GRCh38
rs755711555 Chr.2: 208146166-208146166 on Build GRCh38
rs567482992 Chr.2: 208146402-208146402 on Build GRCh38
rs796283 Chr.2: 208146403-208146403 on Build GRCh38
rs749917012 Chr.2: 208146165-208146165 on Build GRCh38
rs888489323 Chr.2: 208146381-208146381 on Build GRCh38
rs922277107 Chr.2: 208146095-208146095 on Build GRCh38
rs3214759 Chr.2: 208146054-208146054 on Build GRCh38
rs957088019 Chr.2: 208146476-208146476 on Build GRCh38
rs1006527556 Chr.2: 208146441-208146441 on Build GRCh38
rs771872699 Chr.2: 208146112-208146112 on Build GRCh38
rs938004188 Chr.2: 208146378-208146378 on Build GRCh38
rs937576657 Chr.2: 208146407-208146407 on Build GRCh38
rs540054442 Chr.2: 208146421-208146422 on Build GRCh38
rs6750890 Chr.2: 208146290-208146290 on Build GRCh38
rs773424443 Chr.2: 208146137-208146137 on Build GRCh38
rs538582541 Chr.2: 208146453-208146453 on Build GRCh38
rs925686232 Chr.2: 208146312-208146312 on Build GRCh38
rs888564018 Chr.2: 208146214-208146214 on Build GRCh38
rs566723377 Chr.2: 208146055-208146055 on Build GRCh38
rs1047069448 Chr.2: 208146246-208146246 on Build GRCh38
rs753192985 Chr.2: 208146068-208146068 on Build GRCh38
rs758821979 Chr.2: 208146082-208146082 on Build GRCh38
rs760303014 Chr.2: 208146051-208146051 on Build GRCh38
rs1015457328 Chr.2: 208146052-208146052 on Build GRCh38
rs566655544 Chr.2: 208146489-208146489 on Build GRCh38
rs1047075042 Chr.2: 208146371-208146371 on Build GRCh38
rs760215065 Chr.2: 208146136-208146136 on Build GRCh38
rs184047772 Chr.2: 208146099-208146099 on Build GRCh38

View Full Product Details