Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 208163112-208163611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYGA
Entrez Gene ID
1418
Gene Name
crystallin gamma A
Gene Aliases
CRY-g-A, CRYG1, CRYG5
UniGene
Hs.122566
Gene Chromosome Location
Chr.2: 208160740-208163573 on Build GRCh38
Transcripts
NM_014617.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2714676 c.121G>A p.G41S Chr.2: 208163335-208163335 on Build GRCh38
2714677 c.55T>A p.C19S Chr.2: 208163401-208163401 on Build GRCh38
476829 c.143G>A p.R48H Chr.2: 208163313-208163313 on Build GRCh38
1669700 c.226G>A p.V76I Chr.2: 208163230-208163230 on Build GRCh38
5634338 c.79C>T p.R27W Chr.2: 208163377-208163377 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772950141 Chr.2: 208163405-208163405 on Build GRCh38
rs139353014 Chr.2: 208163217-208163217 on Build GRCh38
rs774360381 Chr.2: 208163388-208163388 on Build GRCh38
rs772816574 Chr.2: 208163472-208163472 on Build GRCh38
rs201827303 Chr.2: 208163378-208163378 on Build GRCh38
rs796279 Chr.2: 208163147-208163147 on Build GRCh38
rs780928270 Chr.2: 208163334-208163334 on Build GRCh38
rs188362743 Chr.2: 208163205-208163205 on Build GRCh38
rs796280 Chr.2: 208163467-208163467 on Build GRCh38
rs758405015 Chr.2: 208163531-208163531 on Build GRCh38
rs759390704 Chr.2: 208163262-208163262 on Build GRCh38
rs770475698 Chr.2: 208163186-208163186 on Build GRCh38
rs749790903 Chr.2: 208163294-208163294 on Build GRCh38
rs751172036 Chr.2: 208163230-208163230 on Build GRCh38
rs750806292 Chr.2: 208163281-208163281 on Build GRCh38
rs902028178 Chr.2: 208163293-208163293 on Build GRCh38
rs763539665 Chr.2: 208163265-208163265 on Build GRCh38
rs202133087 Chr.2: 208163577-208163577 on Build GRCh38
rs754681469 Chr.2: 208163513-208163513 on Build GRCh38
rs374303156 Chr.2: 208163501-208163501 on Build GRCh38
rs777353293 Chr.2: 208163160-208163160 on Build GRCh38
rs745601613 Chr.2: 208163319-208163319 on Build GRCh38
rs759445810 Chr.2: 208163216-208163216 on Build GRCh38
rs201484025 Chr.2: 208163376-208163376 on Build GRCh38
rs749448001 Chr.2: 208163198-208163198 on Build GRCh38
rs893192027 Chr.2: 208163324-208163324 on Build GRCh38
rs200908113 Chr.2: 208163434-208163434 on Build GRCh38
rs375579616 Chr.2: 208163523-208163523 on Build GRCh38
rs764656418 Chr.2: 208163492-208163492 on Build GRCh38
rs867113070 Chr.2: 208163374-208163374 on Build GRCh38
rs768683406 Chr.2: 208163335-208163335 on Build GRCh38
rs898031476 Chr.2: 208163157-208163157 on Build GRCh38
rs780765789 Chr.2: 208163246-208163246 on Build GRCh38
rs772385132 Chr.2: 208163255-208163255 on Build GRCh38
rs776104508 Chr.2: 208163554-208163554 on Build GRCh38
rs766174390 Chr.2: 208163413-208163413 on Build GRCh38
rs752704157 Chr.2: 208163367-208163367 on Build GRCh38
rs531952695 Chr.2: 208163449-208163449 on Build GRCh38
rs371745828 Chr.2: 208163338-208163338 on Build GRCh38
rs377147315 Chr.2: 208163564-208163564 on Build GRCh38
rs999437796 Chr.2: 208163383-208163383 on Build GRCh38
rs781698377 Chr.2: 208163173-208163173 on Build GRCh38
rs61743752 Chr.2: 208163403-208163403 on Build GRCh38
rs761341888 Chr.2: 208163390-208163390 on Build GRCh38
rs754147939 Chr.2: 208163415-208163415 on Build GRCh38
rs960546250 Chr.2: 208163399-208163399 on Build GRCh38
rs916358912 Chr.2: 208163407-208163407 on Build GRCh38
rs1034955367 Chr.2: 208163569-208163569 on Build GRCh38
rs765347325 Chr.2: 208163503-208163503 on Build GRCh38
rs370522253 Chr.2: 208163208-208163208 on Build GRCh38
rs763741494 Chr.2: 208163228-208163228 on Build GRCh38
rs748294649 Chr.2: 208163552-208163552 on Build GRCh38
rs369908341 Chr.2: 208163497-208163497 on Build GRCh38
rs779686715 Chr.2: 208163377-208163377 on Build GRCh38
rs757469338 Chr.2: 208163446-208163446 on Build GRCh38
rs374282720 Chr.2: 208163296-208163296 on Build GRCh38
rs200475169 Chr.2: 208163264-208163264 on Build GRCh38
rs778835173 Chr.2: 208163313-208163313 on Build GRCh38
rs767347878 Chr.2: 208163280-208163280 on Build GRCh38
rs760001981 Chr.2: 208163423-208163423 on Build GRCh38
rs758035434 Chr.2: 208163158-208163158 on Build GRCh38
rs776502634 Chr.2: 208163210-208163210 on Build GRCh38
rs1012021158 Chr.2: 208163361-208163361 on Build GRCh38
rs932169794 Chr.2: 208163483-208163483 on Build GRCh38
rs562523650 Chr.2: 208163448-208163448 on Build GRCh38
rs560771595 Chr.2: 208163435-208163435 on Build GRCh38
rs757901754 Chr.2: 208163226-208163226 on Build GRCh38
rs760579805 Chr.2: 208163422-208163423 on Build GRCh38
rs768912260 Chr.2: 208163202-208163202 on Build GRCh38
rs766118742 Chr.2: 208163209-208163209 on Build GRCh38
rs778747832 Chr.2: 208163516-208163516 on Build GRCh38
rs981986667 Chr.2: 208163259-208163259 on Build GRCh38
rs766873432 Chr.2: 208163347-208163347 on Build GRCh38
rs143962326 Chr.2: 208163232-208163232 on Build GRCh38
rs116558662 Chr.2: 208163386-208163386 on Build GRCh38
rs941130517 Chr.2: 208163141-208163141 on Build GRCh38
rs369076657 Chr.2: 208163451-208163451 on Build GRCh38
rs775831512 Chr.2: 208163566-208163566 on Build GRCh38
rs376230114 Chr.2: 208163263-208163263 on Build GRCh38
rs77906750 Chr.2: 208163188-208163188 on Build GRCh38
rs148902669 Chr.2: 208163533-208163533 on Build GRCh38
rs376030660 Chr.2: 208163371-208163371 on Build GRCh38
rs146865595 Chr.2: 208163428-208163428 on Build GRCh38
rs746403438 Chr.2: 208163177-208163177 on Build GRCh38
rs750368223 Chr.2: 208163314-208163314 on Build GRCh38
rs993706566 Chr.2: 208163159-208163159 on Build GRCh38
rs548789050 Chr.2: 208163458-208163458 on Build GRCh38
rs763424012 Chr.2: 208163491-208163491 on Build GRCh38
rs773733603 Chr.2: 208163336-208163336 on Build GRCh38
rs752649458 Chr.2: 208163212-208163212 on Build GRCh38
rs879786486 Chr.2: 208163391-208163391 on Build GRCh38
rs752251453 Chr.2: 208163153-208163153 on Build GRCh38
rs958131696 Chr.2: 208163218-208163218 on Build GRCh38
rs907533627 Chr.2: 208163437-208163437 on Build GRCh38
rs116623945 Chr.2: 208163284-208163284 on Build GRCh38
rs763679430 Chr.2: 208163225-208163225 on Build GRCh38
rs749333355 Chr.2: 208163250-208163250 on Build GRCh38
rs746459739 Chr.2: 208163542-208163542 on Build GRCh38
rs372935883 Chr.2: 208163544-208163544 on Build GRCh38
rs114156183 Chr.2: 208163431-208163431 on Build GRCh38
rs146228453 Chr.2: 208163285-208163285 on Build GRCh38
rs772186337 Chr.2: 208163469-208163469 on Build GRCh38
rs779538919 Chr.2: 208163362-208163362 on Build GRCh38
rs943644658 Chr.2: 208163397-208163397 on Build GRCh38
rs770474616 Chr.2: 208163476-208163476 on Build GRCh38
rs755408221 Chr.2: 208163358-208163358 on Build GRCh38
rs532100898 Chr.2: 208163412-208163412 on Build GRCh38
rs747260199 Chr.2: 208163375-208163375 on Build GRCh38
rs745553455 Chr.2: 208163560-208163560 on Build GRCh38
rs765662559 Chr.2: 208163487-208163487 on Build GRCh38
rs767569752 Chr.2: 208163510-208163510 on Build GRCh38
rs200460837 Chr.2: 208163488-208163488 on Build GRCh38
rs769165374 Chr.2: 208163511-208163511 on Build GRCh38
rs149152810 Chr.2: 208163409-208163409 on Build GRCh38
rs115598490 Chr.2: 208163257-208163257 on Build GRCh38
rs370073865 Chr.2: 208163277-208163277 on Build GRCh38
rs137892904 Chr.2: 208163231-208163231 on Build GRCh38
rs774592838 Chr.2: 208163251-208163251 on Build GRCh38
rs144143988 Chr.2: 208163346-208163346 on Build GRCh38
rs148628204 Chr.2: 208163270-208163270 on Build GRCh38
rs778898033 Chr.2: 208163525-208163525 on Build GRCh38
rs765603148 Chr.2: 208163427-208163427 on Build GRCh38
rs370025596 Chr.2: 208163165-208163165 on Build GRCh38
rs775363182 Chr.2: 208163323-208163323 on Build GRCh38
rs764529623 Chr.2: 208163581-208163581 on Build GRCh38
rs776358584 Chr.2: 208163484-208163484 on Build GRCh38
rs534202025 Chr.2: 208163474-208163474 on Build GRCh38
rs199835340 Chr.2: 208163315-208163315 on Build GRCh38
rs930316395 Chr.2: 208163254-208163254 on Build GRCh38
rs756792544 Chr.2: 208163236-208163236 on Build GRCh38
rs777902010 Chr.2: 208163372-208163372 on Build GRCh38
rs374581135 Chr.2: 208163237-208163237 on Build GRCh38
rs747383459 Chr.2: 208163164-208163164 on Build GRCh38

View Full Product Details