Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 208165290-208165788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf80
Entrez Gene ID
389073
Gene Name
chromosome 2 open reading frame 80
Gene Aliases
GONDA1
UniGene
Hs.198416
Gene Chromosome Location
Chr.2: 208165347-208190258 on Build GRCh38
Transcripts
NM_001099334.2 XM_017004074.1 XM_017004075.1 XM_017004076.1 XM_017004077.1 XM_017004078.1

SNP Detail

SNP ID Chromosome Location
rs200790495 Chr.2: 208165620-208165620 on Build GRCh38
rs545261066 Chr.2: 208165551-208165551 on Build GRCh38
rs961473553 Chr.2: 208165441-208165441 on Build GRCh38
rs758279577 Chr.2: 208165731-208165731 on Build GRCh38
rs10178748 Chr.2: 208165344-208165344 on Build GRCh38
rs539727681 Chr.2: 208165514-208165514 on Build GRCh38
rs902915326 Chr.2: 208165474-208165474 on Build GRCh38
rs925722411 Chr.2: 208165658-208165658 on Build GRCh38
rs951454380 Chr.2: 208165660-208165660 on Build GRCh38
rs760493916 Chr.2: 208165332-208165332 on Build GRCh38
rs949664611 Chr.2: 208165718-208165718 on Build GRCh38
rs972745476 Chr.2: 208165531-208165531 on Build GRCh38
rs943216078 Chr.2: 208165586-208165586 on Build GRCh38
rs1039626611 Chr.2: 208165322-208165322 on Build GRCh38
rs556460938 Chr.2: 208165515-208165515 on Build GRCh38
rs1052757439 Chr.2: 208165323-208165323 on Build GRCh38
rs367917289 Chr.2: 208165684-208165684 on Build GRCh38
rs756328795 Chr.2: 208165701-208165701 on Build GRCh38
rs774741859 Chr.2: 208165618-208165619 on Build GRCh38
rs1029616790 Chr.2: 208165541-208165541 on Build GRCh38
rs998631702 Chr.2: 208165491-208165491 on Build GRCh38
rs969767018 Chr.2: 208165587-208165587 on Build GRCh38
rs547568116 Chr.2: 208165416-208165416 on Build GRCh38
rs556084696 Chr.2: 208165388-208165388 on Build GRCh38
rs758884880 Chr.2: 208165756-208165756 on Build GRCh38
rs76084234 Chr.2: 208165619-208165619 on Build GRCh38
rs766575352 Chr.2: 208165715-208165715 on Build GRCh38
rs777784582 Chr.2: 208165737-208165737 on Build GRCh38
rs921209080 Chr.2: 208165729-208165729 on Build GRCh38
rs906088598 Chr.2: 208165733-208165733 on Build GRCh38
rs752527569 Chr.2: 208165726-208165726 on Build GRCh38
rs896693242 Chr.2: 208165550-208165550 on Build GRCh38
rs563362783 Chr.2: 208165571-208165571 on Build GRCh38
rs397755731 Chr.2: 208165617-208165618 on Build GRCh38
rs10191088 Chr.2: 208165326-208165326 on Build GRCh38
rs1023182662 Chr.2: 208165349-208165349 on Build GRCh38
rs995485638 Chr.2: 208165592-208165592 on Build GRCh38
rs73986528 Chr.2: 208165676-208165676 on Build GRCh38
rs965248802 Chr.2: 208165685-208165685 on Build GRCh38
rs895485581 Chr.2: 208165689-208165689 on Build GRCh38
rs1026967302 Chr.2: 208165659-208165659 on Build GRCh38
rs750518378 Chr.2: 208165694-208165694 on Build GRCh38
rs978600789 Chr.2: 208165613-208165613 on Build GRCh38
rs955338048 Chr.2: 208165545-208165545 on Build GRCh38
rs542864793 Chr.2: 208165386-208165386 on Build GRCh38
rs34111482 Chr.2: 208165606-208165607 on Build GRCh38
rs1045054697 Chr.2: 208165723-208165723 on Build GRCh38
rs574292522 Chr.2: 208165751-208165751 on Build GRCh38
rs1017451235 Chr.2: 208165418-208165418 on Build GRCh38
rs755317888 Chr.2: 208165577-208165577 on Build GRCh38
rs570680853 Chr.2: 208165471-208165471 on Build GRCh38

View Full Product Details