Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 210188944-210189451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACADL
Entrez Gene ID
33
Gene Name
acyl-CoA dehydrogenase, long chain
Gene Aliases
ACAD4, LCAD
UniGene
Hs.471277
Gene Chromosome Location
Chr.2: 210187990-210225491 on Build GRCh38
Transcripts
NM_001608.3 XM_005246517.4 XM_017003955.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1015834 c.1268C>T p.A423V Chr.2: 210188986-210188986 on Build GRCh38
5984706 c.1281C>G p.V427V Chr.2: 210188973-210188973 on Build GRCh38
5647351 c.1246G>T p.E416* Chr.2: 210189008-210189008 on Build GRCh38
1721424 c.1254G>A p.M418I Chr.2: 210189000-210189000 on Build GRCh38
6591669 c.1292A>C p.*431S Chr.2: 210188962-210188962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs397687662 Chr.2: 210189231-210189232 on Build GRCh38
rs1011394065 Chr.2: 210189362-210189362 on Build GRCh38
rs751128035 Chr.2: 210188980-210188980 on Build GRCh38
rs779939047 Chr.2: 210189093-210189094 on Build GRCh38
rs756382365 Chr.2: 210189033-210189033 on Build GRCh38
rs34169168 Chr.2: 210189364-210189364 on Build GRCh38
rs751519042 Chr.2: 210189105-210189105 on Build GRCh38
rs775497317 Chr.2: 210189026-210189026 on Build GRCh38
rs11403691 Chr.2: 210189223-210189224 on Build GRCh38
rs200346431 Chr.2: 210188977-210188977 on Build GRCh38
rs771868717 Chr.2: 210189066-210189067 on Build GRCh38
rs780641408 Chr.2: 210189020-210189020 on Build GRCh38
rs1028922906 Chr.2: 210189198-210189198 on Build GRCh38
rs994651259 Chr.2: 210189128-210189128 on Build GRCh38
rs748113818 Chr.2: 210189066-210189066 on Build GRCh38
rs773259187 Chr.2: 210189089-210189089 on Build GRCh38
rs541119070 Chr.2: 210189219-210189219 on Build GRCh38
rs532732605 Chr.2: 210189253-210189253 on Build GRCh38
rs967518727 Chr.2: 210189405-210189405 on Build GRCh38
rs747028647 Chr.2: 210189086-210189086 on Build GRCh38
rs6757575 Chr.2: 210189141-210189141 on Build GRCh38
rs572592795 Chr.2: 210189197-210189197 on Build GRCh38
rs540253402 Chr.2: 210189229-210189230 on Build GRCh38
rs370174318 Chr.2: 210188981-210188981 on Build GRCh38
rs78091981 Chr.2: 210189417-210189417 on Build GRCh38
rs368601761 Chr.2: 210189423-210189423 on Build GRCh38
rs889162974 Chr.2: 210189195-210189195 on Build GRCh38
rs1018950283 Chr.2: 210189392-210189392 on Build GRCh38
rs763588960 Chr.2: 210188970-210188970 on Build GRCh38
rs899011907 Chr.2: 210189076-210189076 on Build GRCh38
rs761007593 Chr.2: 210189090-210189090 on Build GRCh38
rs763960636 Chr.2: 210189096-210189096 on Build GRCh38
rs562293303 Chr.2: 210189094-210189094 on Build GRCh38
rs866655470 Chr.2: 210189376-210189376 on Build GRCh38
rs764988272 Chr.2: 210189091-210189091 on Build GRCh38
rs1008941897 Chr.2: 210189306-210189306 on Build GRCh38
rs745563670 Chr.2: 210189021-210189021 on Build GRCh38
rs763794276 Chr.2: 210189302-210189304 on Build GRCh38
rs1020122383 Chr.2: 210189373-210189373 on Build GRCh38
rs769348791 Chr.2: 210189048-210189048 on Build GRCh38
rs888996769 Chr.2: 210189242-210189242 on Build GRCh38
rs756803658 Chr.2: 210188988-210188988 on Build GRCh38
rs892958742 Chr.2: 210189328-210189328 on Build GRCh38
rs780610346 Chr.2: 210189040-210189040 on Build GRCh38
rs1052160005 Chr.2: 210189122-210189122 on Build GRCh38
rs202156594 Chr.2: 210189232-210189232 on Build GRCh38
rs909370695 Chr.2: 210188979-210188979 on Build GRCh38
rs775016696 Chr.2: 210189058-210189058 on Build GRCh38
rs564035894 Chr.2: 210189221-210189221 on Build GRCh38
rs772228742 Chr.2: 210189084-210189084 on Build GRCh38
rs749778106 Chr.2: 210189046-210189046 on Build GRCh38

View Full Product Details