Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 210590722-210591223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPS1
Entrez Gene ID
1373
Gene Name
carbamoyl-phosphate synthase 1
Gene Aliases
CPSASE1, PHN
UniGene
Hs.149252
Gene Chromosome Location
Chr.2: 210477682-210679107 on Build GRCh38
Transcripts
NM_001122633.2 NM_001875.4 XM_011510640.1 XM_011510641.2 XM_011510642.2 XM_011510643.2 XM_011510644.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3695230 c.958G>A p.A320T Chr.2: 210590899-210590899 on Build GRCh38
6876358 c.905G>A p.G302E Chr.2: 210590846-210590846 on Build GRCh38
6156814 c.951G>A p.M317I Chr.2: 210590892-210590892 on Build GRCh38
6156816 c.925G>T p.A309S Chr.2: 210590866-210590866 on Build GRCh38
6876359 c.887G>A p.G296E Chr.2: 210590846-210590846 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs865881540 Chr.2: 210590846-210590846 on Build GRCh38
rs775920437 Chr.2: 210590870-210590870 on Build GRCh38
rs1043653086 Chr.2: 210591041-210591042 on Build GRCh38
rs369197253 Chr.2: 210590839-210590839 on Build GRCh38
rs750254669 Chr.2: 210590869-210590869 on Build GRCh38
rs368879550 Chr.2: 210590766-210590766 on Build GRCh38
rs1038817529 Chr.2: 210590999-210590999 on Build GRCh38
rs774427324 Chr.2: 210590901-210590901 on Build GRCh38
rs549657614 Chr.2: 210591055-210591055 on Build GRCh38
rs547804162 Chr.2: 210591049-210591050 on Build GRCh38
rs569726440 Chr.2: 210591095-210591095 on Build GRCh38
rs754897201 Chr.2: 210590793-210590793 on Build GRCh38
rs749366764 Chr.2: 210590883-210590883 on Build GRCh38
rs946590771 Chr.2: 210591034-210591034 on Build GRCh38
rs761974037 Chr.2: 210590904-210590904 on Build GRCh38
rs998891944 Chr.2: 210591051-210591053 on Build GRCh38
rs905926423 Chr.2: 210591188-210591188 on Build GRCh38
rs146733657 Chr.2: 210590824-210590824 on Build GRCh38
rs901764396 Chr.2: 210591051-210591051 on Build GRCh38
rs759296092 Chr.2: 210590920-210590920 on Build GRCh38
rs983156096 Chr.2: 210590967-210590967 on Build GRCh38
rs900344612 Chr.2: 210591019-210591019 on Build GRCh38
rs763652819 Chr.2: 210590938-210590938 on Build GRCh38
rs780759800 Chr.2: 210590950-210590950 on Build GRCh38
rs367569543 Chr.2: 210591165-210591165 on Build GRCh38
rs756452498 Chr.2: 210591042-210591045 on Build GRCh38
rs974059313 Chr.2: 210591184-210591184 on Build GRCh38
rs751836081 Chr.2: 210590771-210590771 on Build GRCh38
rs781583758 Chr.2: 210590841-210590841 on Build GRCh38
rs770315452 Chr.2: 210590865-210590865 on Build GRCh38
rs373078786 Chr.2: 210590791-210590791 on Build GRCh38
rs777559659 Chr.2: 210590829-210590829 on Build GRCh38
rs751150083 Chr.2: 210590945-210590945 on Build GRCh38
rs376173850 Chr.2: 210590952-210590952 on Build GRCh38
rs148654695 Chr.2: 210590816-210590816 on Build GRCh38
rs762035533 Chr.2: 210590782-210590782 on Build GRCh38
rs1052980713 Chr.2: 210591068-210591068 on Build GRCh38
rs904521305 Chr.2: 210591041-210591041 on Build GRCh38
rs549170365 Chr.2: 210591046-210591049 on Build GRCh38
rs943047651 Chr.2: 210590958-210590958 on Build GRCh38
rs186874606 Chr.2: 210591162-210591162 on Build GRCh38
rs928901389 Chr.2: 210590823-210590823 on Build GRCh38
rs143608594 Chr.2: 210590842-210590842 on Build GRCh38
rs749736947 Chr.2: 210590957-210590957 on Build GRCh38
rs891628083 Chr.2: 210591081-210591081 on Build GRCh38
rs75431694 Chr.2: 210591195-210591195 on Build GRCh38
rs200407408 Chr.2: 210590948-210590948 on Build GRCh38
rs1014367104 Chr.2: 210591105-210591106 on Build GRCh38
rs746589282 Chr.2: 210590834-210590834 on Build GRCh38
rs933212302 Chr.2: 210591065-210591065 on Build GRCh38
rs766131653 Chr.2: 210590790-210590790 on Build GRCh38
rs758410860 Chr.2: 210590930-210590930 on Build GRCh38
rs911671535 Chr.2: 210590982-210590982 on Build GRCh38
rs78693997 Chr.2: 210590990-210590990 on Build GRCh38
rs973321068 Chr.2: 210590861-210590861 on Build GRCh38
rs200664811 Chr.2: 210590882-210590882 on Build GRCh38
rs147030565 Chr.2: 210590847-210590847 on Build GRCh38
rs745402454 Chr.2: 210590792-210590792 on Build GRCh38
rs1023112136 Chr.2: 210591126-210591126 on Build GRCh38
rs865937266 Chr.2: 210590900-210590900 on Build GRCh38
rs587780323 Chr.2: 210590896-210590896 on Build GRCh38
rs767663476 Chr.2: 210590911-210590911 on Build GRCh38
rs201276852 Chr.2: 210590925-210590925 on Build GRCh38
rs763059352 Chr.2: 210590750-210590750 on Build GRCh38
rs1011697559 Chr.2: 210591106-210591106 on Build GRCh38
rs544805806 Chr.2: 210591042-210591049 on Build GRCh38
rs527950704 Chr.2: 210590927-210590927 on Build GRCh38
rs753607947 Chr.2: 210590792-210590792 on Build GRCh38
rs780472297 Chr.2: 210590956-210590956 on Build GRCh38
rs920214738 Chr.2: 210590931-210590931 on Build GRCh38
rs1031749759 Chr.2: 210591076-210591076 on Build GRCh38
rs756996491 Chr.2: 210590836-210590836 on Build GRCh38
rs895927648 Chr.2: 210591104-210591104 on Build GRCh38
rs779222831 Chr.2: 210591045-210591046 on Build GRCh38
rs776454212 Chr.2: 210590913-210590913 on Build GRCh38
rs202130487 Chr.2: 210590815-210590815 on Build GRCh38

View Full Product Details