Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 214728941-214729483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BARD1
Entrez Gene ID
580
Gene Name
BRCA1 associated RING domain 1
Gene Aliases
-
UniGene
Hs.591642
Gene Chromosome Location
Chr.2: 214725645-214809711 on Build GRCh38
Transcripts
NM_000465.3 NM_001282543.1 NM_001282545.1 NM_001282548.1 NM_001282549.1 NR_104212.1 NR_104215.1 NR_104216.1 XM_017004613.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6915084 c.2027A>T p.Y676F Chr.2: 214728983-214728983 on Build GRCh38
4090994 c.2025C>T p.C675C Chr.2: 214728985-214728985 on Build GRCh38
6963610 c.2056delC p.H686fs*28 Chr.2: 214728954-214728954 on Build GRCh38
324796 c.2021G>T p.G674V Chr.2: 214728989-214728989 on Build GRCh38
6360509 c.2020G>T p.G674* Chr.2: 214728990-214728990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371282361 Chr.2: 214729058-214729058 on Build GRCh38
rs759021562 Chr.2: 214728997-214728997 on Build GRCh38
rs939762530 Chr.2: 214729243-214729243 on Build GRCh38
rs768178334 Chr.2: 214729028-214729028 on Build GRCh38
rs564142780 Chr.2: 214729436-214729436 on Build GRCh38
rs972499168 Chr.2: 214729418-214729418 on Build GRCh38
rs756177960 Chr.2: 214729031-214729031 on Build GRCh38
rs937744017 Chr.2: 214729398-214729398 on Build GRCh38
rs901867423 Chr.2: 214729339-214729339 on Build GRCh38
rs913708216 Chr.2: 214729208-214729208 on Build GRCh38
rs544192939 Chr.2: 214729290-214729290 on Build GRCh38
rs80353631 Chr.2: 214729257-214729257 on Build GRCh38
rs746790711 Chr.2: 214728981-214728981 on Build GRCh38
rs2075623 Chr.2: 214729127-214729127 on Build GRCh38
rs939708904 Chr.2: 214729417-214729417 on Build GRCh38
rs998937157 Chr.2: 214729346-214729346 on Build GRCh38
rs876658260 Chr.2: 214729010-214729010 on Build GRCh38
rs986125202 Chr.2: 214729144-214729144 on Build GRCh38
rs952226595 Chr.2: 214729287-214729287 on Build GRCh38
rs970254352 Chr.2: 214729391-214729391 on Build GRCh38
rs759922505 Chr.2: 214729212-214729212 on Build GRCh38
rs749485640 Chr.2: 214728966-214728966 on Build GRCh38
rs919707876 Chr.2: 214729439-214729439 on Build GRCh38
rs528400881 Chr.2: 214729171-214729171 on Build GRCh38
rs374748665 Chr.2: 214729143-214729143 on Build GRCh38
rs986239771 Chr.2: 214729296-214729296 on Build GRCh38
rs141614490 Chr.2: 214729173-214729173 on Build GRCh38
rs557138715 Chr.2: 214729201-214729201 on Build GRCh38
rs140586144 Chr.2: 214729329-214729329 on Build GRCh38
rs777147991 Chr.2: 214729077-214729077 on Build GRCh38
rs977930219 Chr.2: 214729189-214729189 on Build GRCh38
rs535394326 Chr.2: 214729181-214729181 on Build GRCh38
rs990354027 Chr.2: 214729214-214729214 on Build GRCh38
rs898312636 Chr.2: 214729258-214729258 on Build GRCh38
rs530775291 Chr.2: 214729128-214729128 on Build GRCh38
rs770917842 Chr.2: 214728983-214728983 on Build GRCh38
rs546872335 Chr.2: 214729389-214729389 on Build GRCh38
rs786203617 Chr.2: 214728995-214728995 on Build GRCh38
rs775039040 Chr.2: 214729002-214729002 on Build GRCh38
rs566787065 Chr.2: 214729441-214729441 on Build GRCh38
rs924788210 Chr.2: 214729210-214729210 on Build GRCh38
rs771766430 Chr.2: 214728977-214728977 on Build GRCh38
rs2075622 Chr.2: 214729147-214729147 on Build GRCh38
rs774052575 Chr.2: 214729325-214729325 on Build GRCh38
rs761788396 Chr.2: 214729372-214729372 on Build GRCh38
rs573443019 Chr.2: 214728979-214728979 on Build GRCh38
rs187240320 Chr.2: 214729019-214729019 on Build GRCh38
rs748347675 Chr.2: 214729028-214729028 on Build GRCh38
rs949072465 Chr.2: 214729421-214729421 on Build GRCh38
rs587780027 Chr.2: 214729020-214729020 on Build GRCh38
rs869144116 Chr.2: 214729121-214729121 on Build GRCh38
rs763465706 Chr.2: 214729411-214729412 on Build GRCh38
rs376886445 Chr.2: 214729043-214729043 on Build GRCh38
rs79130403 Chr.2: 214729271-214729271 on Build GRCh38
rs982118419 Chr.2: 214729392-214729392 on Build GRCh38
rs150593152 Chr.2: 214729265-214729265 on Build GRCh38
rs777178471 Chr.2: 214729209-214729209 on Build GRCh38
rs568849245 Chr.2: 214729224-214729224 on Build GRCh38
rs1027859083 Chr.2: 214729283-214729283 on Build GRCh38
rs957533271 Chr.2: 214729363-214729363 on Build GRCh38
rs976026657 Chr.2: 214729204-214729204 on Build GRCh38
rs769302736 Chr.2: 214729001-214729001 on Build GRCh38
rs185881889 Chr.2: 214729347-214729347 on Build GRCh38
rs1028170410 Chr.2: 214729132-214729132 on Build GRCh38
rs568072727 Chr.2: 214729180-214729180 on Build GRCh38
rs546919839 Chr.2: 214729453-214729453 on Build GRCh38
rs529213347 Chr.2: 214729030-214729030 on Build GRCh38
rs754017565 Chr.2: 214729036-214729036 on Build GRCh38
rs928301405 Chr.2: 214729237-214729237 on Build GRCh38
rs945195376 Chr.2: 214729356-214729356 on Build GRCh38
rs540058098 Chr.2: 214729227-214729227 on Build GRCh38
rs1014498719 Chr.2: 214729067-214729067 on Build GRCh38
rs867573161 Chr.2: 214729337-214729337 on Build GRCh38
rs869312497 Chr.2: 214729073-214729073 on Build GRCh38
rs528255964 Chr.2: 214729452-214729452 on Build GRCh38
rs776515437 Chr.2: 214728992-214728992 on Build GRCh38
rs1017604842 Chr.2: 214729305-214729305 on Build GRCh38
rs755282725 Chr.2: 214729051-214729051 on Build GRCh38
rs889674884 Chr.2: 214729320-214729320 on Build GRCh38
rs780112770 Chr.2: 214729035-214729035 on Build GRCh38
rs147982569 Chr.2: 214729122-214729122 on Build GRCh38
rs142839313 Chr.2: 214729413-214729413 on Build GRCh38
rs762601855 Chr.2: 214729009-214729009 on Build GRCh38
rs199758343 Chr.2: 214729018-214729018 on Build GRCh38
rs192064905 Chr.2: 214729241-214729241 on Build GRCh38
rs567516524 Chr.2: 214729236-214729236 on Build GRCh38

View Full Product Details