Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 214730319-214730817 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BARD1
Entrez Gene ID
580
Gene Name
BRCA1 associated RING domain 1
Gene Aliases
-
UniGene
Hs.591642
Gene Chromosome Location
Chr.2: 214725645-214809711 on Build GRCh38
Transcripts
NM_000465.3 NM_001282543.1 NM_001282545.1 NM_001282548.1 NM_001282549.1 NR_104212.1 NR_104215.1 NR_104216.1 XM_017004613.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6439061 c.1969C>T p.P657S Chr.2: 214730443-214730443 on Build GRCh38
5904142 c.1970C>T p.P657L Chr.2: 214730442-214730442 on Build GRCh38
1016061 c.1922G>A p.R641Q Chr.2: 214730490-214730490 on Build GRCh38
1245956 c.2001G>T p.L667L Chr.2: 214730411-214730411 on Build GRCh38
243167 c.1995A>G p.E665E Chr.2: 214730417-214730417 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886039589 Chr.2: 214730458-214730477 on Build GRCh38
rs12614960 Chr.2: 214730788-214730788 on Build GRCh38
rs587781376 Chr.2: 214730497-214730497 on Build GRCh38
rs3738886 Chr.2: 214730721-214730721 on Build GRCh38
rs768808840 Chr.2: 214730346-214730346 on Build GRCh38
rs864622634 Chr.2: 214730514-214730514 on Build GRCh38
rs587780024 Chr.2: 214730457-214730458 on Build GRCh38
rs876659021 Chr.2: 214730463-214730463 on Build GRCh38
rs876659046 Chr.2: 214730466-214730467 on Build GRCh38
rs147215925 Chr.2: 214730435-214730435 on Build GRCh38
rs1018054664 Chr.2: 214730719-214730719 on Build GRCh38
rs924129828 Chr.2: 214730569-214730569 on Build GRCh38
rs765487816 Chr.2: 214730488-214730488 on Build GRCh38
rs587780026 Chr.2: 214730418-214730418 on Build GRCh38
rs4986840 Chr.2: 214730575-214730575 on Build GRCh38
rs561178343 Chr.2: 214730757-214730757 on Build GRCh38
rs760750600 Chr.2: 214730692-214730692 on Build GRCh38
rs776976800 Chr.2: 214730547-214730547 on Build GRCh38
rs876658741 Chr.2: 214730452-214730452 on Build GRCh38
rs786202232 Chr.2: 214730472-214730472 on Build GRCh38
rs556775078 Chr.2: 214730427-214730427 on Build GRCh38
rs752870879 Chr.2: 214730490-214730490 on Build GRCh38
rs1048597173 Chr.2: 214730709-214730709 on Build GRCh38
rs876660360 Chr.2: 214730511-214730511 on Build GRCh38
rs587782767 Chr.2: 214730423-214730423 on Build GRCh38
rs573845527 Chr.2: 214730557-214730557 on Build GRCh38
rs778163347 Chr.2: 214730403-214730403 on Build GRCh38
rs879254244 Chr.2: 214730443-214730443 on Build GRCh38
rs876659560 Chr.2: 214730510-214730510 on Build GRCh38
rs780901872 Chr.2: 214730492-214730492 on Build GRCh38
rs781133235 Chr.2: 214730390-214730390 on Build GRCh38
rs876658395 Chr.2: 214730457-214730457 on Build GRCh38
rs527409471 Chr.2: 214730649-214730649 on Build GRCh38
rs864622239 Chr.2: 214730510-214730510 on Build GRCh38
rs3738887 Chr.2: 214730716-214730716 on Build GRCh38
rs786203772 Chr.2: 214730449-214730449 on Build GRCh38
rs1046874140 Chr.2: 214730705-214730705 on Build GRCh38
rs587782534 Chr.2: 214730437-214730437 on Build GRCh38
rs587782504 Chr.2: 214730479-214730480 on Build GRCh38
rs866207461 Chr.2: 214730601-214730601 on Build GRCh38
rs75237746 Chr.2: 214730345-214730345 on Build GRCh38
rs538389082 Chr.2: 214730738-214730738 on Build GRCh38
rs142778447 Chr.2: 214730614-214730614 on Build GRCh38
rs587781948 Chr.2: 214730491-214730491 on Build GRCh38
rs876658544 Chr.2: 214730419-214730419 on Build GRCh38
rs730881422 Chr.2: 214730416-214730416 on Build GRCh38
rs897309316 Chr.2: 214730639-214730639 on Build GRCh38
rs765959857 Chr.2: 214730698-214730698 on Build GRCh38
rs371902585 Chr.2: 214730715-214730715 on Build GRCh38
rs774986157 Chr.2: 214730538-214730539 on Build GRCh38
rs2228456 Chr.2: 214730479-214730479 on Build GRCh38
rs375745253 Chr.2: 214730544-214730544 on Build GRCh38
rs770348371 Chr.2: 214730762-214730762 on Build GRCh38
rs876658489 Chr.2: 214730498-214730498 on Build GRCh38
rs876658774 Chr.2: 214730433-214730433 on Build GRCh38
rs754012979 Chr.2: 214730465-214730465 on Build GRCh38
rs916559975 Chr.2: 214730596-214730596 on Build GRCh38
rs774178253 Chr.2: 214730520-214730520 on Build GRCh38
rs766781362 Chr.2: 214730446-214730446 on Build GRCh38
rs532833921 Chr.2: 214730604-214730604 on Build GRCh38
rs373636345 Chr.2: 214730540-214730540 on Build GRCh38
rs994024069 Chr.2: 214730411-214730411 on Build GRCh38
rs900859585 Chr.2: 214730751-214730751 on Build GRCh38
rs769934362 Chr.2: 214730361-214730361 on Build GRCh38
rs772577316 Chr.2: 214730524-214730524 on Build GRCh38
rs567493750 Chr.2: 214730409-214730409 on Build GRCh38
rs997857004 Chr.2: 214730733-214730733 on Build GRCh38
rs372186770 Chr.2: 214730366-214730366 on Build GRCh38
rs562753068 Chr.2: 214730603-214730603 on Build GRCh38
rs1053814359 Chr.2: 214730589-214730592 on Build GRCh38
rs187590361 Chr.2: 214730424-214730424 on Build GRCh38
rs923517227 Chr.2: 214730529-214730529 on Build GRCh38
rs778601623 Chr.2: 214730355-214730355 on Build GRCh38
rs572554455 Chr.2: 214730445-214730445 on Build GRCh38
rs745780723 Chr.2: 214730393-214730393 on Build GRCh38
rs771472958 Chr.2: 214730509-214730510 on Build GRCh38
rs756136465 Chr.2: 214730500-214730500 on Build GRCh38
rs770694880 Chr.2: 214730379-214730379 on Build GRCh38
rs773223671 Chr.2: 214730507-214730507 on Build GRCh38
rs374808704 Chr.2: 214730559-214730559 on Build GRCh38
rs559845412 Chr.2: 214730623-214730623 on Build GRCh38
rs906443784 Chr.2: 214730693-214730693 on Build GRCh38
rs2228455 Chr.2: 214730455-214730455 on Build GRCh38
rs948059310 Chr.2: 214730677-214730677 on Build GRCh38
rs550255355 Chr.2: 214730759-214730759 on Build GRCh38
rs2228456 Chr.2: 214730479-214730479 on Build GRCh38
rs747735614 Chr.2: 214730523-214730523 on Build GRCh38
rs587780025 Chr.2: 214730451-214730451 on Build GRCh38
rs370366588 Chr.2: 214730553-214730553 on Build GRCh38
rs377227840 Chr.2: 214730439-214730439 on Build GRCh38
rs769194998 Chr.2: 214730401-214730401 on Build GRCh38
rs376639978 Chr.2: 214730513-214730513 on Build GRCh38
rs775009517 Chr.2: 214730405-214730405 on Build GRCh38
rs786202462 Chr.2: 214730420-214730420 on Build GRCh38
rs786203739 Chr.2: 214730442-214730442 on Build GRCh38
rs773682954 Chr.2: 214730533-214730533 on Build GRCh38
rs768259863 Chr.2: 214730518-214730518 on Build GRCh38
rs768490891 Chr.2: 214730410-214730410 on Build GRCh38
rs3738888 Chr.2: 214730440-214730440 on Build GRCh38

View Full Product Details