Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 215318067-215318578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATIC
Entrez Gene ID
471
Gene Name
5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase
Gene Aliases
AICAR, AICARFT, HEL-S-70p, IMPCHASE, PURH
UniGene
Hs.90280
Gene Chromosome Location
Chr.2: 215311956-215359745 on Build GRCh38
Transcripts
NM_004044.6 XM_017004187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6156675 c.183G>T p.L61F Chr.2: 215318193-215318193 on Build GRCh38
5687793 c.188G>A p.G63E Chr.2: 215318198-215318198 on Build GRCh38
4091079 c.207T>C p.H69H Chr.2: 215318217-215318217 on Build GRCh38
4188593 c.149delA p.D50fs*5 Chr.2: 215318159-215318159 on Build GRCh38
7292804 c.190C>T p.R64C Chr.2: 215318200-215318200 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs948036748 Chr.2: 215318417-215318417 on Build GRCh38
rs966386481 Chr.2: 215318340-215318340 on Build GRCh38
rs746342557 Chr.2: 215318279-215318279 on Build GRCh38
rs771410955 Chr.2: 215318194-215318194 on Build GRCh38
rs760330806 Chr.2: 215318141-215318141 on Build GRCh38
rs764278248 Chr.2: 215318222-215318222 on Build GRCh38
rs1032290638 Chr.2: 215318343-215318343 on Build GRCh38
rs778335377 Chr.2: 215318192-215318192 on Build GRCh38
rs762719666 Chr.2: 215318527-215318527 on Build GRCh38
rs754454433 Chr.2: 215318175-215318175 on Build GRCh38
rs757062929 Chr.2: 215318149-215318149 on Build GRCh38
rs747764194 Chr.2: 215318185-215318185 on Build GRCh38
rs780025938 Chr.2: 215318161-215318161 on Build GRCh38
rs920215626 Chr.2: 215318484-215318484 on Build GRCh38
rs762539306 Chr.2: 215318093-215318093 on Build GRCh38
rs1031345029 Chr.2: 215318357-215318357 on Build GRCh38
rs941656788 Chr.2: 215318528-215318528 on Build GRCh38
rs557185991 Chr.2: 215318142-215318142 on Build GRCh38
rs750274350 Chr.2: 215318227-215318227 on Build GRCh38
rs200243425 Chr.2: 215318246-215318246 on Build GRCh38
rs747368066 Chr.2: 215318193-215318193 on Build GRCh38
rs559539947 Chr.2: 215318299-215318299 on Build GRCh38
rs78709969 Chr.2: 215318354-215318354 on Build GRCh38
rs142625411 Chr.2: 215318384-215318384 on Build GRCh38
rs546892581 Chr.2: 215318530-215318530 on Build GRCh38
rs766717765 Chr.2: 215318383-215318383 on Build GRCh38
rs766241704 Chr.2: 215318230-215318230 on Build GRCh38
rs113259114 Chr.2: 215318552-215318552 on Build GRCh38
rs753761573 Chr.2: 215318234-215318234 on Build GRCh38
rs778731660 Chr.2: 215318176-215318176 on Build GRCh38
rs771266190 Chr.2: 215318258-215318258 on Build GRCh38
rs1019744371 Chr.2: 215318326-215318326 on Build GRCh38
rs761634994 Chr.2: 215318239-215318239 on Build GRCh38
rs548232749 Chr.2: 215318342-215318342 on Build GRCh38
rs1050400723 Chr.2: 215318502-215318502 on Build GRCh38
rs766890677 Chr.2: 215318129-215318129 on Build GRCh38
rs752854432 Chr.2: 215318145-215318145 on Build GRCh38
rs928023829 Chr.2: 215318453-215318453 on Build GRCh38
rs776438273 Chr.2: 215318198-215318199 on Build GRCh38
rs761077005 Chr.2: 215318370-215318370 on Build GRCh38
rs530506345 Chr.2: 215318367-215318367 on Build GRCh38
rs763201142 Chr.2: 215318212-215318212 on Build GRCh38
rs776634842 Chr.2: 215318133-215318133 on Build GRCh38
rs763823990 Chr.2: 215318167-215318168 on Build GRCh38
rs1036974015 Chr.2: 215318423-215318423 on Build GRCh38
rs765062751 Chr.2: 215318263-215318266 on Build GRCh38
rs754477465 Chr.2: 215318245-215318245 on Build GRCh38
rs748753649 Chr.2: 215318174-215318174 on Build GRCh38
rs777126703 Chr.2: 215318196-215318196 on Build GRCh38
rs575560797 Chr.2: 215318159-215318159 on Build GRCh38
rs577250930 Chr.2: 215318288-215318288 on Build GRCh38
rs957922745 Chr.2: 215318344-215318344 on Build GRCh38
rs547917143 Chr.2: 215318515-215318515 on Build GRCh38
rs768319628 Chr.2: 215318247-215318247 on Build GRCh38
rs375380564 Chr.2: 215318138-215318138 on Build GRCh38
rs760513075 Chr.2: 215318228-215318228 on Build GRCh38
rs777565849 Chr.2: 215318248-215318248 on Build GRCh38
rs775716964 Chr.2: 215318208-215318208 on Build GRCh38
rs1015971848 Chr.2: 215318298-215318298 on Build GRCh38
rs761307452 Chr.2: 215318110-215318110 on Build GRCh38
rs928692805 Chr.2: 215318392-215318392 on Build GRCh38
rs781568379 Chr.2: 215318263-215318263 on Build GRCh38
rs191838418 Chr.2: 215318249-215318249 on Build GRCh38
rs771822727 Chr.2: 215318191-215318191 on Build GRCh38
rs760020376 Chr.2: 215318485-215318485 on Build GRCh38
rs368883626 Chr.2: 215318201-215318201 on Build GRCh38
rs568600704 Chr.2: 215318539-215318539 on Build GRCh38
rs535034335 Chr.2: 215318512-215318512 on Build GRCh38
rs931571844 Chr.2: 215318499-215318499 on Build GRCh38
rs145427771 Chr.2: 215318296-215318296 on Build GRCh38
rs940052887 Chr.2: 215318394-215318394 on Build GRCh38
rs775451236 Chr.2: 215318419-215318419 on Build GRCh38
rs756052816 Chr.2: 215318160-215318160 on Build GRCh38
rs770253516 Chr.2: 215318285-215318285 on Build GRCh38
rs759545338 Chr.2: 215318200-215318200 on Build GRCh38
rs774936617 Chr.2: 215318126-215318127 on Build GRCh38
rs954548278 Chr.2: 215318352-215318352 on Build GRCh38
rs182367530 Chr.2: 215318323-215318323 on Build GRCh38
rs570803253 Chr.2: 215318475-215318475 on Build GRCh38

View Full Product Details