Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 215325127-215325527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATIC
Entrez Gene ID
471
Gene Name
5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase
Gene Aliases
AICAR, AICARFT, HEL-S-70p, IMPCHASE, PURH
UniGene
Hs.90280
Gene Chromosome Location
Chr.2: 215311956-215359745 on Build GRCh38
Transcripts
NM_004044.6 XM_017004187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6363942 c.314C>A p.P105H Chr.2: 215325264-215325264 on Build GRCh38
7207044 c.305A>G p.N102S Chr.2: 215325255-215325255 on Build GRCh38
127378 c.324G>A p.K108K Chr.2: 215325274-215325274 on Build GRCh38
3757936 c.347C>G p.T116S Chr.2: 215325297-215325297 on Build GRCh38
3909559 c.378T>A p.I126I Chr.2: 215325328-215325328 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757749957 Chr.2: 215325229-215325229 on Build GRCh38
rs771242358 Chr.2: 215325237-215325237 on Build GRCh38
rs372823382 Chr.2: 215325261-215325261 on Build GRCh38
rs763913204 Chr.2: 215325198-215325198 on Build GRCh38
rs992092808 Chr.2: 215325421-215325424 on Build GRCh38
rs749985833 Chr.2: 215325210-215325210 on Build GRCh38
rs745674921 Chr.2: 215325192-215325192 on Build GRCh38
rs150904300 Chr.2: 215325455-215325455 on Build GRCh38
rs1026401298 Chr.2: 215325473-215325473 on Build GRCh38
rs755538098 Chr.2: 215325221-215325221 on Build GRCh38
rs753100430 Chr.2: 215325291-215325291 on Build GRCh38
rs772024484 Chr.2: 215325344-215325344 on Build GRCh38
rs754271666 Chr.2: 215325265-215325265 on Build GRCh38
rs896222998 Chr.2: 215325443-215325443 on Build GRCh38
rs774089396 Chr.2: 215325250-215325250 on Build GRCh38
rs756503280 Chr.2: 215325211-215325211 on Build GRCh38
rs374349232 Chr.2: 215325208-215325208 on Build GRCh38
rs917799306 Chr.2: 215325430-215325430 on Build GRCh38
rs779287107 Chr.2: 215325225-215325225 on Build GRCh38
rs867575051 Chr.2: 215325377-215325377 on Build GRCh38
rs765791354 Chr.2: 215325287-215325287 on Build GRCh38
rs760175417 Chr.2: 215325356-215325356 on Build GRCh38
rs753599464 Chr.2: 215325167-215325168 on Build GRCh38
rs772768704 Chr.2: 215325346-215325346 on Build GRCh38
rs972800050 Chr.2: 215325447-215325447 on Build GRCh38
rs931446368 Chr.2: 215325471-215325471 on Build GRCh38
rs370646845 Chr.2: 215325205-215325205 on Build GRCh38
rs749096659 Chr.2: 215325321-215325321 on Build GRCh38
rs1021031556 Chr.2: 215325322-215325322 on Build GRCh38
rs748077057 Chr.2: 215325343-215325343 on Build GRCh38
rs759971633 Chr.2: 215325368-215325368 on Build GRCh38
rs768277320 Chr.2: 215325329-215325329 on Build GRCh38
rs772678539 Chr.2: 215325259-215325259 on Build GRCh38
rs746457093 Chr.2: 215325362-215325363 on Build GRCh38
rs766770737 Chr.2: 215325219-215325219 on Build GRCh38
rs757608964 Chr.2: 215325305-215325305 on Build GRCh38
rs141553318 Chr.2: 215325226-215325226 on Build GRCh38
rs781558741 Chr.2: 215325316-215325316 on Build GRCh38
rs771443605 Chr.2: 215325254-215325254 on Build GRCh38
rs768703956 Chr.2: 215325249-215325249 on Build GRCh38
rs770491729 Chr.2: 215325195-215325196 on Build GRCh38
rs779277115 Chr.2: 215325234-215325235 on Build GRCh38
rs568677758 Chr.2: 215325242-215325242 on Build GRCh38
rs550473309 Chr.2: 215325236-215325236 on Build GRCh38
rs78089903 Chr.2: 215325204-215325204 on Build GRCh38
rs770578299 Chr.2: 215325359-215325359 on Build GRCh38
rs572919021 Chr.2: 215325328-215325328 on Build GRCh38
rs373947561 Chr.2: 215325196-215325196 on Build GRCh38
rs1050244445 Chr.2: 215325494-215325494 on Build GRCh38
rs150994921 Chr.2: 215325330-215325330 on Build GRCh38
rs375997993 Chr.2: 215325300-215325300 on Build GRCh38
rs759755383 Chr.2: 215325285-215325285 on Build GRCh38
rs368525228 Chr.2: 215325239-215325239 on Build GRCh38
rs35373619 Chr.2: 215325453-215325454 on Build GRCh38
rs754181820 Chr.2: 215325220-215325220 on Build GRCh38
rs766003571 Chr.2: 215325263-215325263 on Build GRCh38
rs758534337 Chr.2: 215325293-215325293 on Build GRCh38
rs78530224 Chr.2: 215325195-215325195 on Build GRCh38
rs186070291 Chr.2: 215325418-215325418 on Build GRCh38
rs2372536 Chr.2: 215325297-215325297 on Build GRCh38
rs1019801127 Chr.2: 215325277-215325277 on Build GRCh38
rs964508906 Chr.2: 215325231-215325231 on Build GRCh38
rs1008447373 Chr.2: 215325267-215325267 on Build GRCh38
rs1001005285 Chr.2: 215325358-215325358 on Build GRCh38
rs1056242332 Chr.2: 215325434-215325434 on Build GRCh38
rs533547933 Chr.2: 215325421-215325421 on Build GRCh38
rs370059601 Chr.2: 215325363-215325363 on Build GRCh38

View Full Product Details