Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 215332315-215332815 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATIC
Entrez Gene ID
471
Gene Name
5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase
Gene Aliases
AICAR, AICARFT, HEL-S-70p, IMPCHASE, PURH
UniGene
Hs.90280
Gene Chromosome Location
Chr.2: 215311956-215359745 on Build GRCh38
Transcripts
NM_004044.6 XM_017004187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3577494 c.731T>C p.L244S Chr.2: 215332424-215332424 on Build GRCh38
6578012 c.804C>G p.V268V Chr.2: 215332497-215332497 on Build GRCh38
442284 c.707_708insA p.F237fs*45 Chr.2: 215332400-215332401 on Build GRCh38
3577495 c.779C>T p.P260L Chr.2: 215332472-215332472 on Build GRCh38
4991698 c.792delT p.F265fs*77 Chr.2: 215332485-215332485 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762550408 Chr.2: 215332550-215332550 on Build GRCh38
rs909704443 Chr.2: 215332724-215332724 on Build GRCh38
rs750765780 Chr.2: 215332472-215332472 on Build GRCh38
rs776539487 Chr.2: 215332429-215332429 on Build GRCh38
rs763816500 Chr.2: 215332466-215332466 on Build GRCh38
rs1023320804 Chr.2: 215332667-215332668 on Build GRCh38
rs556427086 Chr.2: 215332428-215332428 on Build GRCh38
rs752235664 Chr.2: 215332353-215332353 on Build GRCh38
rs967274518 Chr.2: 215332592-215332592 on Build GRCh38
rs752105264 Chr.2: 215332609-215332609 on Build GRCh38
rs553998884 Chr.2: 215332666-215332667 on Build GRCh38
rs531345076 Chr.2: 215332694-215332694 on Build GRCh38
rs990886523 Chr.2: 215332672-215332672 on Build GRCh38
rs767907801 Chr.2: 215332471-215332471 on Build GRCh38
rs149050827 Chr.2: 215332777-215332777 on Build GRCh38
rs200997625 Chr.2: 215332441-215332441 on Build GRCh38
rs772398085 Chr.2: 215332529-215332529 on Build GRCh38
rs180904761 Chr.2: 215332679-215332679 on Build GRCh38
rs202217105 Chr.2: 215332362-215332362 on Build GRCh38
rs753965140 Chr.2: 215332484-215332484 on Build GRCh38
rs756581030 Chr.2: 215332401-215332401 on Build GRCh38
rs886645279 Chr.2: 215332425-215332425 on Build GRCh38
rs933951153 Chr.2: 215332772-215332772 on Build GRCh38
rs764747617 Chr.2: 215332357-215332357 on Build GRCh38
rs374163623 Chr.2: 215332385-215332385 on Build GRCh38
rs1020149119 Chr.2: 215332577-215332577 on Build GRCh38
rs918425350 Chr.2: 215332341-215332341 on Build GRCh38
rs751120987 Chr.2: 215332468-215332468 on Build GRCh38
rs934109007 Chr.2: 215332494-215332494 on Build GRCh38
rs55726617 Chr.2: 215332395-215332395 on Build GRCh38
rs759406879 Chr.2: 215332343-215332343 on Build GRCh38
rs764393346 Chr.2: 215332478-215332479 on Build GRCh38
rs951129669 Chr.2: 215332695-215332695 on Build GRCh38
rs267599202 Chr.2: 215332500-215332500 on Build GRCh38
rs769863387 Chr.2: 215332531-215332531 on Build GRCh38
rs942593171 Chr.2: 215332396-215332396 on Build GRCh38
rs995853353 Chr.2: 215332541-215332541 on Build GRCh38
rs375890690 Chr.2: 215332340-215332340 on Build GRCh38
rs113047875 Chr.2: 215332366-215332366 on Build GRCh38
rs527460867 Chr.2: 215332565-215332565 on Build GRCh38
rs200251432 Chr.2: 215332516-215332516 on Build GRCh38
rs780155225 Chr.2: 215332405-215332405 on Build GRCh38
rs143393493 Chr.2: 215332416-215332416 on Build GRCh38
rs79918378 Chr.2: 215332697-215332697 on Build GRCh38
rs759599403 Chr.2: 215332431-215332431 on Build GRCh38
rs779225067 Chr.2: 215332505-215332505 on Build GRCh38
rs898916901 Chr.2: 215332490-215332490 on Build GRCh38
rs560930353 Chr.2: 215332681-215332681 on Build GRCh38
rs76012683 Chr.2: 215332338-215332338 on Build GRCh38
rs112449215 Chr.2: 215332345-215332345 on Build GRCh38
rs746845244 Chr.2: 215332420-215332420 on Build GRCh38
rs949640050 Chr.2: 215332389-215332389 on Build GRCh38
rs373852805 Chr.2: 215332545-215332545 on Build GRCh38
rs762266604 Chr.2: 215332470-215332470 on Build GRCh38
rs759150834 Chr.2: 215332348-215332348 on Build GRCh38
rs777338179 Chr.2: 215332735-215332735 on Build GRCh38
rs140409106 Chr.2: 215332417-215332417 on Build GRCh38
rs749313836 Chr.2: 215332410-215332410 on Build GRCh38
rs372671136 Chr.2: 215332664-215332664 on Build GRCh38
rs565432639 Chr.2: 215332667-215332670 on Build GRCh38
rs762299898 Chr.2: 215332459-215332459 on Build GRCh38
rs151259995 Chr.2: 215332476-215332476 on Build GRCh38
rs754173354 Chr.2: 215332485-215332485 on Build GRCh38
rs144904859 Chr.2: 215332764-215332764 on Build GRCh38
rs925781777 Chr.2: 215332645-215332645 on Build GRCh38
rs150438200 Chr.2: 215332477-215332477 on Build GRCh38
rs764883597 Chr.2: 215332352-215332352 on Build GRCh38
rs879174265 Chr.2: 215332738-215332738 on Build GRCh38
rs894757440 Chr.2: 215332648-215332648 on Build GRCh38
rs545890470 Chr.2: 215332680-215332680 on Build GRCh38
rs958383767 Chr.2: 215332668-215332668 on Build GRCh38
rs775211176 Chr.2: 215332458-215332458 on Build GRCh38
rs190484695 Chr.2: 215332556-215332556 on Build GRCh38
rs370954832 Chr.2: 215332518-215332518 on Build GRCh38
rs754962743 Chr.2: 215332488-215332488 on Build GRCh38
rs768068470 Chr.2: 215332670-215332671 on Build GRCh38
rs1054610978 Chr.2: 215332591-215332591 on Build GRCh38
rs781187321 Chr.2: 215332530-215332530 on Build GRCh38
rs184328612 Chr.2: 215332756-215332756 on Build GRCh38
rs576046697 Chr.2: 215332380-215332380 on Build GRCh38

View Full Product Details