Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 215361884-215362383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FN1
Entrez Gene ID
2335
Gene Name
fibronectin 1
Gene Aliases
CIG, ED-B, FINC, FN, FNZ, GFND, GFND2, LETS, MSF
UniGene
Hs.203717
Gene Chromosome Location
Chr.2: 215360440-215436167 on Build GRCh38
Transcripts
NM_001306129.1 NM_001306130.1 NM_001306131.1 NM_001306132.1 NM_002026.3 NM_212474.2 NM_212476.2 NM_212478.2 NM_212482.2 XM_005246397.1 XM_005246398.1 XM_005246399.1 XM_005246401.1 XM_005246402.1 XM_005246403.1 XM_005246404.1 XM_005246405.1 XM_005246406.1 XM_005246407.1 XM_005246408.1 XM_005246409.1 XM_005246410.1 XM_005246411.1 XM_005246412.1 XM_005246414.1 XM_005246416.1 XM_017003692.1 XM_017003693.1 XM_017003694.1 XM_017003695.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4831290 c.6978C>T p.Y2326Y Chr.2: 215361987-215361987 on Build GRCh38
5712140 c.7346A>G p.H2449R Chr.2: 215361985-215361985 on Build GRCh38
6368237 c.7349A>G p.Q2450R Chr.2: 215361982-215361982 on Build GRCh38
7368575 c.7295G>C p.S2432T Chr.2: 215362036-215362036 on Build GRCh38
5058642 c.6907C>T p.R2303C Chr.2: 215362058-215362058 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750752854 Chr.2: 215362045-215362045 on Build GRCh38
rs1049995824 Chr.2: 215361915-215361915 on Build GRCh38
rs763422179 Chr.2: 215362113-215362113 on Build GRCh38
rs550137802 Chr.2: 215362143-215362143 on Build GRCh38
rs761562863 Chr.2: 215362094-215362094 on Build GRCh38
rs780439441 Chr.2: 215362124-215362124 on Build GRCh38
rs370833185 Chr.2: 215362031-215362031 on Build GRCh38
rs150636748 Chr.2: 215362065-215362065 on Build GRCh38
rs549165594 Chr.2: 215362107-215362107 on Build GRCh38
rs770422507 Chr.2: 215361926-215361926 on Build GRCh38
rs1003487753 Chr.2: 215362092-215362092 on Build GRCh38
rs902464947 Chr.2: 215361969-215361969 on Build GRCh38
rs1040926799 Chr.2: 215362282-215362282 on Build GRCh38
rs200155993 Chr.2: 215362079-215362079 on Build GRCh38
rs751303604 Chr.2: 215362021-215362021 on Build GRCh38
rs766212576 Chr.2: 215361962-215361962 on Build GRCh38
rs962839589 Chr.2: 215361952-215361952 on Build GRCh38
rs893475921 Chr.2: 215362345-215362345 on Build GRCh38
rs931435023 Chr.2: 215361912-215361912 on Build GRCh38
rs777473071 Chr.2: 215362110-215362111 on Build GRCh38
rs965853274 Chr.2: 215362108-215362108 on Build GRCh38
rs1040978896 Chr.2: 215361960-215361960 on Build GRCh38
rs767223039 Chr.2: 215362100-215362100 on Build GRCh38
rs147460086 Chr.2: 215362036-215362036 on Build GRCh38
rs755629195 Chr.2: 215362108-215362110 on Build GRCh38
rs766977112 Chr.2: 215361970-215361971 on Build GRCh38
rs1002133370 Chr.2: 215361910-215361910 on Build GRCh38
rs759273898 Chr.2: 215362073-215362073 on Build GRCh38
rs754356447 Chr.2: 215362131-215362131 on Build GRCh38
rs137921535 Chr.2: 215361916-215361916 on Build GRCh38
rs752145367 Chr.2: 215362289-215362289 on Build GRCh38
rs148505961 Chr.2: 215362057-215362057 on Build GRCh38
rs531653996 Chr.2: 215362134-215362134 on Build GRCh38
rs762328949 Chr.2: 215361996-215361996 on Build GRCh38
rs756955589 Chr.2: 215362029-215362029 on Build GRCh38
rs776403679 Chr.2: 215361972-215361972 on Build GRCh38
rs780377696 Chr.2: 215362048-215362048 on Build GRCh38
rs543224167 Chr.2: 215362294-215362294 on Build GRCh38
rs186473709 Chr.2: 215362348-215362348 on Build GRCh38
rs376141269 Chr.2: 215362110-215362110 on Build GRCh38
rs149830916 Chr.2: 215362068-215362068 on Build GRCh38
rs756702314 Chr.2: 215362046-215362046 on Build GRCh38
rs376532901 Chr.2: 215361947-215361947 on Build GRCh38
rs911333765 Chr.2: 215362250-215362250 on Build GRCh38
rs551626688 Chr.2: 215362255-215362255 on Build GRCh38
rs190187140 Chr.2: 215362260-215362260 on Build GRCh38
rs988262455 Chr.2: 215362109-215362109 on Build GRCh38
rs140721264 Chr.2: 215362077-215362077 on Build GRCh38
rs761996366 Chr.2: 215361961-215361961 on Build GRCh38
rs147396555 Chr.2: 215361977-215361977 on Build GRCh38
rs1028475224 Chr.2: 215362316-215362317 on Build GRCh38
rs913623581 Chr.2: 215362349-215362349 on Build GRCh38
rs1028557072 Chr.2: 215362032-215362032 on Build GRCh38
rs995762610 Chr.2: 215361983-215361983 on Build GRCh38
rs923834582 Chr.2: 215362283-215362283 on Build GRCh38
rs954314599 Chr.2: 215362318-215362318 on Build GRCh38
rs184857180 Chr.2: 215362123-215362123 on Build GRCh38
rs181619033 Chr.2: 215362347-215362347 on Build GRCh38
rs567352724 Chr.2: 215362317-215362317 on Build GRCh38
rs73987859 Chr.2: 215362230-215362230 on Build GRCh38
rs759470429 Chr.2: 215361985-215361985 on Build GRCh38
rs750840778 Chr.2: 215362101-215362101 on Build GRCh38
rs774400320 Chr.2: 215361950-215361950 on Build GRCh38
rs1056508322 Chr.2: 215362248-215362248 on Build GRCh38
rs574355866 Chr.2: 215362293-215362293 on Build GRCh38
rs1037485970 Chr.2: 215362292-215362292 on Build GRCh38
rs752220359 Chr.2: 215362016-215362017 on Build GRCh38
rs1008885616 Chr.2: 215362326-215362326 on Build GRCh38
rs35525385 Chr.2: 215362286-215362286 on Build GRCh38
rs372963822 Chr.2: 215362072-215362072 on Build GRCh38
rs762925169 Chr.2: 215362089-215362089 on Build GRCh38
rs569093565 Chr.2: 215362147-215362147 on Build GRCh38
rs995702119 Chr.2: 215362302-215362302 on Build GRCh38
rs763627375 Chr.2: 215362015-215362015 on Build GRCh38
rs1018620600 Chr.2: 215362341-215362341 on Build GRCh38
rs371108916 Chr.2: 215362063-215362063 on Build GRCh38
rs765023530 Chr.2: 215361989-215361989 on Build GRCh38
rs772213289 Chr.2: 215362071-215362071 on Build GRCh38
rs768945288 Chr.2: 215361949-215361949 on Build GRCh38
rs766121465 Chr.2: 215362017-215362017 on Build GRCh38
rs751109054 Chr.2: 215362093-215362093 on Build GRCh38
rs13394662 Chr.2: 215361919-215361919 on Build GRCh38
rs752269352 Chr.2: 215361994-215361994 on Build GRCh38
rs926571992 Chr.2: 215362146-215362146 on Build GRCh38
rs775329459 Chr.2: 215362084-215362084 on Build GRCh38
rs775645872 Chr.2: 215361929-215361929 on Build GRCh38

View Full Product Details