Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218401486-218401981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTDSP1
Entrez Gene ID
58190
Gene Name
CTD small phosphatase 1
Gene Aliases
NIF3, NLI-IF, NLIIF, SCP1
UniGene
Hs.444468
Gene Chromosome Location
Chr.2: 218398338-218405941 on Build GRCh38
Transcripts
NM_001206878.1 NM_021198.2 NM_182642.2 XM_011511569.2 XM_011511570.2 XM_017004615.1 XM_017004616.1 XM_017004617.1 XM_017004618.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3577691 c.211C>T p.P71S Chr.2: 218401707-218401707 on Build GRCh38
5465414 c.71A>G p.D24G Chr.2: 218401567-218401567 on Build GRCh38
82092 c.111C>T p.G37G Chr.2: 218401607-218401607 on Build GRCh38
3766541 c.204C>A p.G68G Chr.2: 218401700-218401700 on Build GRCh38
3714122 c.160C>A p.L54M Chr.2: 218401656-218401656 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758353883 Chr.2: 218401678-218401678 on Build GRCh38
rs750946268 Chr.2: 218401628-218401628 on Build GRCh38
rs767066083 Chr.2: 218401669-218401669 on Build GRCh38
rs1020728731 Chr.2: 218401891-218401891 on Build GRCh38
rs2227249 Chr.2: 218401662-218401662 on Build GRCh38
rs761244574 Chr.2: 218401667-218401667 on Build GRCh38
rs753541890 Chr.2: 218401520-218401520 on Build GRCh38
rs560807301 Chr.2: 218401518-218401518 on Build GRCh38
rs371016771 Chr.2: 218401742-218401742 on Build GRCh38
rs543297376 Chr.2: 218401910-218401910 on Build GRCh38
rs763554630 Chr.2: 218401603-218401603 on Build GRCh38
rs1000081277 Chr.2: 218401932-218401932 on Build GRCh38
rs766894908 Chr.2: 218401724-218401724 on Build GRCh38
rs748228855 Chr.2: 218401641-218401641 on Build GRCh38
rs554190636 Chr.2: 218401734-218401734 on Build GRCh38
rs376602431 Chr.2: 218401703-218401703 on Build GRCh38
rs762028780 Chr.2: 218401564-218401564 on Build GRCh38
rs768811153 Chr.2: 218401561-218401561 on Build GRCh38
rs770262431 Chr.2: 218401554-218401554 on Build GRCh38
rs745753612 Chr.2: 218401695-218401695 on Build GRCh38
rs2227252 Chr.2: 218401718-218401718 on Build GRCh38
rs761474879 Chr.2: 218401723-218401723 on Build GRCh38
rs755455799 Chr.2: 218401676-218401676 on Build GRCh38
rs1047813043 Chr.2: 218401779-218401779 on Build GRCh38
rs757792576 Chr.2: 218401544-218401544 on Build GRCh38
rs759244769 Chr.2: 218401570-218401570 on Build GRCh38
rs141917217 Chr.2: 218401665-218401665 on Build GRCh38
rs773638043 Chr.2: 218401534-218401534 on Build GRCh38
rs78148168 Chr.2: 218401919-218401919 on Build GRCh38
rs1014395417 Chr.2: 218401824-218401824 on Build GRCh38
rs61542239 Chr.2: 218401917-218401917 on Build GRCh38
rs200409368 Chr.2: 218401555-218401555 on Build GRCh38
rs920574526 Chr.2: 218401950-218401950 on Build GRCh38
rs931553998 Chr.2: 218401858-218401858 on Build GRCh38
rs761541525 Chr.2: 218401865-218401865 on Build GRCh38
rs777898337 Chr.2: 218401745-218401745 on Build GRCh38
rs1039983524 Chr.2: 218401836-218401836 on Build GRCh38
rs776367664 Chr.2: 218401647-218401647 on Build GRCh38
rs2227253 Chr.2: 218401737-218401737 on Build GRCh38
rs138499758 Chr.2: 218401663-218401663 on Build GRCh38
rs964697838 Chr.2: 218401916-218401916 on Build GRCh38
rs576966757 Chr.2: 218401959-218401959 on Build GRCh38
rs752920374 Chr.2: 218401739-218401739 on Build GRCh38
rs766831695 Chr.2: 218401671-218401671 on Build GRCh38
rs561357395 Chr.2: 218401913-218401913 on Build GRCh38
rs1052445551 Chr.2: 218401565-218401565 on Build GRCh38
rs772347987 Chr.2: 218401642-218401642 on Build GRCh38
rs879869219 Chr.2: 218401883-218401883 on Build GRCh38
rs945314999 Chr.2: 218401550-218401550 on Build GRCh38
rs2607727 Chr.2: 218401909-218401909 on Build GRCh38
rs746206635 Chr.2: 218401551-218401551 on Build GRCh38
rs752012691 Chr.2: 218401541-218401541 on Build GRCh38
rs867598212 Chr.2: 218401887-218401887 on Build GRCh38
rs759532806 Chr.2: 218401656-218401656 on Build GRCh38
rs756378172 Chr.2: 218401635-218401635 on Build GRCh38
rs144048283 Chr.2: 218401682-218401682 on Build GRCh38
rs532240927 Chr.2: 218401604-218401604 on Build GRCh38
rs754847996 Chr.2: 218401523-218401523 on Build GRCh38
rs906142413 Chr.2: 218401878-218401878 on Build GRCh38
rs762615334 Chr.2: 218401597-218401597 on Build GRCh38
rs371256121 Chr.2: 218401576-218401576 on Build GRCh38
rs565070665 Chr.2: 218401568-218401568 on Build GRCh38
rs867022177 Chr.2: 218401838-218401838 on Build GRCh38
rs770966169 Chr.2: 218401566-218401566 on Build GRCh38
rs373019518 Chr.2: 218401625-218401625 on Build GRCh38
rs943352145 Chr.2: 218401751-218401751 on Build GRCh38
rs764732826 Chr.2: 218401885-218401885 on Build GRCh38
rs554566053 Chr.2: 218401782-218401782 on Build GRCh38
rs748388380 Chr.2: 218401677-218401677 on Build GRCh38
rs956782202 Chr.2: 218401756-218401756 on Build GRCh38
rs543185144 Chr.2: 218401557-218401557 on Build GRCh38
rs930661084 Chr.2: 218401952-218401952 on Build GRCh38
rs930690520 Chr.2: 218401770-218401770 on Build GRCh38
rs979570684 Chr.2: 218401538-218401538 on Build GRCh38
rs538907354 Chr.2: 218401733-218401733 on Build GRCh38
rs986315237 Chr.2: 218401654-218401677 on Build GRCh38
rs2227250 Chr.2: 218401670-218401670 on Build GRCh38
rs753376280 Chr.2: 218401753-218401754 on Build GRCh38
rs147667569 Chr.2: 218401640-218401640 on Build GRCh38
rs756819498 Chr.2: 218401611-218401611 on Build GRCh38
rs777653516 Chr.2: 218401844-218401844 on Build GRCh38
rs944011565 Chr.2: 218401807-218401807 on Build GRCh38
rs528129011 Chr.2: 218401847-218401848 on Build GRCh38
rs968950551 Chr.2: 218401533-218401533 on Build GRCh38
rs549688651 Chr.2: 218401738-218401738 on Build GRCh38
rs752459193 Chr.2: 218401535-218401535 on Build GRCh38
rs761604173 Chr.2: 218401826-218401826 on Build GRCh38
rs1046384186 Chr.2: 218401856-218401856 on Build GRCh38
rs376873930 Chr.2: 218401758-218401758 on Build GRCh38
rs2227251 Chr.2: 218401700-218401700 on Build GRCh38
rs781700464 Chr.2: 218401545-218401545 on Build GRCh38
rs911830142 Chr.2: 218401749-218401749 on Build GRCh38
rs768972814 Chr.2: 218401639-218401639 on Build GRCh38
rs920673130 Chr.2: 218401761-218401761 on Build GRCh38
rs912605891 Chr.2: 218401763-218401763 on Build GRCh38
rs779899761 Chr.2: 218401760-218401760 on Build GRCh38
rs112758322 Chr.2: 218401910-218401910 on Build GRCh38
rs1056802588 Chr.2: 218401888-218401888 on Build GRCh38
rs915787208 Chr.2: 218401522-218401522 on Build GRCh38
rs372485935 Chr.2: 218401717-218401717 on Build GRCh38
rs749236212 Chr.2: 218401764-218401764 on Build GRCh38
rs547460809 Chr.2: 218401638-218401638 on Build GRCh38
rs529670147 Chr.2: 218401659-218401659 on Build GRCh38
rs763631594 Chr.2: 218401747-218401750 on Build GRCh38
rs772743618 Chr.2: 218401729-218401729 on Build GRCh38
rs542614834 Chr.2: 218401507-218401507 on Build GRCh38
rs2227248 Chr.2: 218401537-218401537 on Build GRCh38
rs765083410 Chr.2: 218401574-218401574 on Build GRCh38
rs753239606 Chr.2: 218401790-218401790 on Build GRCh38
rs749077426 Chr.2: 218401706-218401706 on Build GRCh38
rs754228370 Chr.2: 218401675-218401675 on Build GRCh38

View Full Product Details