Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218402212-218402717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTDSP1
Entrez Gene ID
58190
Gene Name
CTD small phosphatase 1
Gene Aliases
NIF3, NLI-IF, NLIIF, SCP1
UniGene
Hs.444468
Gene Chromosome Location
Chr.2: 218398338-218405941 on Build GRCh38
Transcripts
NM_001206878.1 NM_021198.2 NM_182642.2 XM_011511569.2 XM_011511570.2 XM_017004615.1 XM_017004616.1 XM_017004617.1 XM_017004618.1 NR_029500.1

Gene Symbol
MIR26B
Entrez Gene ID
407017
Gene Name
microRNA 26b
Gene Aliases
MIRN26B, hsa-mir-26b, miR-26b
UniGene
Gene Chromosome Location
Chr.2: 218402646-218402722 on Build GRCh38
Transcripts
NM_001206878.1 NM_021198.2 NM_182642.2 XM_011511569.2 XM_011511570.2 XM_017004615.1 XM_017004616.1 XM_017004617.1 XM_017004618.1 NR_029500.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7363196 c.342C>A p.F114L Chr.2: 218402369-218402369 on Build GRCh38
6894962 c.367G>A p.V123M Chr.2: 218402394-218402394 on Build GRCh38
5880398 c.364G>T p.G122W Chr.2: 218402391-218402391 on Build GRCh38
6894963 c.366G>A p.G122G Chr.2: 218402393-218402393 on Build GRCh38
5631296 c.328A>G p.N110D Chr.2: 218402355-218402355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1053046633 Chr.2: 218402478-218402478 on Build GRCh38
rs201501044 Chr.2: 218402629-218402629 on Build GRCh38
rs774816654 Chr.2: 218402486-218402486 on Build GRCh38
rs554678491 Chr.2: 218402277-218402277 on Build GRCh38
rs760684567 Chr.2: 218402267-218402267 on Build GRCh38
rs760143675 Chr.2: 218402493-218402493 on Build GRCh38
rs774977260 Chr.2: 218402340-218402340 on Build GRCh38
rs772118973 Chr.2: 218402645-218402645 on Build GRCh38
rs748671313 Chr.2: 218402396-218402396 on Build GRCh38
rs375845906 Chr.2: 218402250-218402250 on Build GRCh38
rs755635864 Chr.2: 218402319-218402319 on Build GRCh38
rs764954295 Chr.2: 218402334-218402335 on Build GRCh38
rs981520577 Chr.2: 218402385-218402385 on Build GRCh38
rs992998663 Chr.2: 218402615-218402615 on Build GRCh38
rs550380352 Chr.2: 218402445-218402445 on Build GRCh38
rs778510838 Chr.2: 218402611-218402611 on Build GRCh38
rs1048919338 Chr.2: 218402501-218402501 on Build GRCh38
rs763172887 Chr.2: 218402639-218402639 on Build GRCh38
rs996013987 Chr.2: 218402496-218402496 on Build GRCh38
rs935666147 Chr.2: 218402472-218402472 on Build GRCh38
rs758302524 Chr.2: 218402302-218402302 on Build GRCh38
rs377203158 Chr.2: 218402451-218402451 on Build GRCh38
rs747417711 Chr.2: 218402412-218402412 on Build GRCh38
rs763927426 Chr.2: 218402357-218402357 on Build GRCh38
rs548273517 Chr.2: 218402679-218402679 on Build GRCh38
rs891731002 Chr.2: 218402480-218402480 on Build GRCh38
rs550684286 Chr.2: 218402648-218402648 on Build GRCh38
rs772381836 Chr.2: 218402397-218402397 on Build GRCh38
rs149582075 Chr.2: 218402471-218402471 on Build GRCh38
rs779459596 Chr.2: 218402320-218402320 on Build GRCh38
rs368390101 Chr.2: 218402619-218402619 on Build GRCh38
rs561130226 Chr.2: 218402413-218402413 on Build GRCh38
rs766444859 Chr.2: 218402345-218402345 on Build GRCh38
rs529627330 Chr.2: 218402584-218402584 on Build GRCh38
rs754578666 Chr.2: 218402378-218402378 on Build GRCh38
rs767441828 Chr.2: 218402362-218402362 on Build GRCh38
rs997375985 Chr.2: 218402529-218402529 on Build GRCh38
rs563418366 Chr.2: 218402585-218402585 on Build GRCh38
rs1014806411 Chr.2: 218402525-218402525 on Build GRCh38
rs766465594 Chr.2: 218402298-218402298 on Build GRCh38
rs771399614 Chr.2: 218402453-218402453 on Build GRCh38
rs759268777 Chr.2: 218402681-218402681 on Build GRCh38
rs370760593 Chr.2: 218402343-218402343 on Build GRCh38
rs188612260 Chr.2: 218402684-218402684 on Build GRCh38
rs754385144 Chr.2: 218402600-218402600 on Build GRCh38
rs1034425890 Chr.2: 218402543-218402543 on Build GRCh38
rs775574642 Chr.2: 218402612-218402612 on Build GRCh38
rs1015518181 Chr.2: 218402586-218402586 on Build GRCh38
rs79126749 Chr.2: 218402633-218402633 on Build GRCh38
rs759712299 Chr.2: 218402354-218402354 on Build GRCh38
rs866326766 Chr.2: 218402481-218402481 on Build GRCh38
rs769721321 Chr.2: 218402635-218402635 on Build GRCh38
rs780988397 Chr.2: 218402313-218402313 on Build GRCh38
rs541188783 Chr.2: 218402476-218402476 on Build GRCh38
rs372400662 Chr.2: 218402303-218402303 on Build GRCh38
rs767601296 Chr.2: 218402438-218402438 on Build GRCh38
rs776007664 Chr.2: 218402243-218402243 on Build GRCh38
rs776544845 Chr.2: 218402351-218402351 on Build GRCh38
rs565919718 Chr.2: 218402647-218402647 on Build GRCh38
rs992968925 Chr.2: 218402433-218402433 on Build GRCh38
rs138242622 Chr.2: 218402308-218402308 on Build GRCh38
rs751581087 Chr.2: 218402360-218402360 on Build GRCh38
rs769923230 Chr.2: 218402426-218402426 on Build GRCh38
rs200929399 Chr.2: 218402301-218402301 on Build GRCh38
rs758272601 Chr.2: 218402599-218402601 on Build GRCh38
rs900280914 Chr.2: 218402494-218402495 on Build GRCh38
rs927388342 Chr.2: 218402386-218402386 on Build GRCh38
rs990236769 Chr.2: 218402569-218402570 on Build GRCh38
rs745358670 Chr.2: 218402235-218402235 on Build GRCh38
rs779217801 Chr.2: 218402379-218402379 on Build GRCh38
rs912863478 Chr.2: 218402692-218402692 on Build GRCh38
rs775679854 Chr.2: 218402636-218402636 on Build GRCh38
rs778143891 Chr.2: 218402400-218402400 on Build GRCh38
rs779802832 Chr.2: 218402373-218402373 on Build GRCh38
rs756810983 Chr.2: 218402250-218402251 on Build GRCh38
rs887613483 Chr.2: 218402504-218402504 on Build GRCh38
rs935725604 Chr.2: 218402650-218402650 on Build GRCh38
rs753166998 Chr.2: 218402618-218402618 on Build GRCh38
rs367622179 Chr.2: 218402341-218402341 on Build GRCh38
rs773485560 Chr.2: 218402256-218402256 on Build GRCh38
rs530468664 Chr.2: 218402613-218402613 on Build GRCh38
rs771395138 Chr.2: 218402630-218402630 on Build GRCh38
rs371311622 Chr.2: 218402366-218402366 on Build GRCh38
rs944390574 Chr.2: 218402485-218402485 on Build GRCh38
rs748732877 Chr.2: 218402377-218402377 on Build GRCh38
rs574716484 Chr.2: 218402526-218402526 on Build GRCh38
rs531789720 Chr.2: 218402634-218402634 on Build GRCh38
rs766205860 Chr.2: 218402672-218402672 on Build GRCh38
rs769019081 Chr.2: 218402338-218402338 on Build GRCh38
rs952847530 Chr.2: 218402577-218402577 on Build GRCh38
rs975485291 Chr.2: 218402329-218402329 on Build GRCh38
rs372354754 Chr.2: 218402624-218402624 on Build GRCh38
rs745529377 Chr.2: 218402316-218402316 on Build GRCh38
rs1048804001 Chr.2: 218402688-218402688 on Build GRCh38
rs959082307 Chr.2: 218402546-218402546 on Build GRCh38
rs759530944 Chr.2: 218402422-218402422 on Build GRCh38
rs199738523 Chr.2: 218402244-218402244 on Build GRCh38
rs760572605 Chr.2: 218402450-218402450 on Build GRCh38
rs1008614748 Chr.2: 218402265-218402265 on Build GRCh38
rs144071139 Chr.2: 218402511-218402511 on Build GRCh38
rs769496049 Chr.2: 218402236-218402236 on Build GRCh38
rs750118204 Chr.2: 218402339-218402339 on Build GRCh38
rs902184043 Chr.2: 218402264-218402264 on Build GRCh38
rs750056056 Chr.2: 218402363-218402363 on Build GRCh38
rs370798418 Chr.2: 218402342-218402342 on Build GRCh38
rs990752581 Chr.2: 218402449-218402449 on Build GRCh38
rs914711293 Chr.2: 218402570-218402571 on Build GRCh38
rs761873445 Chr.2: 218402434-218402434 on Build GRCh38
rs776971622 Chr.2: 218402414-218402414 on Build GRCh38
rs750219570 Chr.2: 218402448-218402448 on Build GRCh38
rs1028397265 Chr.2: 218402561-218402561 on Build GRCh38
rs370091958 Chr.2: 218402251-218402251 on Build GRCh38
rs372797160 Chr.2: 218402330-218402330 on Build GRCh38

View Full Product Details