Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218402929-218403426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTDSP1
Entrez Gene ID
58190
Gene Name
CTD small phosphatase 1
Gene Aliases
NIF3, NLI-IF, NLIIF, SCP1
UniGene
Hs.444468
Gene Chromosome Location
Chr.2: 218398338-218405941 on Build GRCh38
Transcripts
NM_001206878.1 NM_021198.2 NM_182642.2 XM_011511569.2 XM_011511570.2 XM_017004615.1 XM_017004616.1 XM_017004617.1 XM_017004618.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5015972 c.493_495delCTG p.L166delL Chr.2: 218403253-218403255 on Build GRCh38
1016340 c.598C>T p.R200W Chr.2: 218403358-218403358 on Build GRCh38
1166047 c.559A>T p.N187Y Chr.2: 218403319-218403319 on Build GRCh38
5021359 c.458C>T p.A153V Chr.2: 218403114-218403114 on Build GRCh38
1133003 c.605T>G p.V202G Chr.2: 218403365-218403365 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201362148 Chr.2: 218403216-218403216 on Build GRCh38
rs767602823 Chr.2: 218403358-218403358 on Build GRCh38
rs928429276 Chr.2: 218403194-218403194 on Build GRCh38
rs751623474 Chr.2: 218403262-218403262 on Build GRCh38
rs760313508 Chr.2: 218403215-218403215 on Build GRCh38
rs367625093 Chr.2: 218403010-218403010 on Build GRCh38
rs763118113 Chr.2: 218403253-218403253 on Build GRCh38
rs2227259 Chr.2: 218403234-218403234 on Build GRCh38
rs371780293 Chr.2: 218403015-218403015 on Build GRCh38
rs764860528 Chr.2: 218402964-218402964 on Build GRCh38
rs771568703 Chr.2: 218403205-218403205 on Build GRCh38
rs777504608 Chr.2: 218403378-218403378 on Build GRCh38
rs975788806 Chr.2: 218403064-218403064 on Build GRCh38
rs952999080 Chr.2: 218403362-218403362 on Build GRCh38
rs764216314 Chr.2: 218403343-218403343 on Build GRCh38
rs761822791 Chr.2: 218403355-218403355 on Build GRCh38
rs751309985 Chr.2: 218403157-218403157 on Build GRCh38
rs183416148 Chr.2: 218403165-218403165 on Build GRCh38
rs778600045 Chr.2: 218403278-218403278 on Build GRCh38
rs373050159 Chr.2: 218403318-218403318 on Build GRCh38
rs770995590 Chr.2: 218403237-218403238 on Build GRCh38
rs371853283 Chr.2: 218403026-218403026 on Build GRCh38
rs750784843 Chr.2: 218402980-218402980 on Build GRCh38
rs778567598 Chr.2: 218403200-218403200 on Build GRCh38
rs779857064 Chr.2: 218403149-218403149 on Build GRCh38
rs772562703 Chr.2: 218403210-218403210 on Build GRCh38
rs754070826 Chr.2: 218403041-218403041 on Build GRCh38
rs752812937 Chr.2: 218403076-218403076 on Build GRCh38
rs2227255 Chr.2: 218403058-218403058 on Build GRCh38
rs750323073 Chr.2: 218403185-218403185 on Build GRCh38
rs1036186554 Chr.2: 218403010-218403011 on Build GRCh38
rs145171368 Chr.2: 218403284-218403284 on Build GRCh38
rs780354648 Chr.2: 218402985-218402985 on Build GRCh38
rs931919077 Chr.2: 218403100-218403100 on Build GRCh38
rs749531944 Chr.2: 218402988-218402988 on Build GRCh38
rs375530018 Chr.2: 218403148-218403148 on Build GRCh38
rs530186884 Chr.2: 218403014-218403014 on Build GRCh38
rs780119781 Chr.2: 218402984-218403010 on Build GRCh38
rs754234738 Chr.2: 218403170-218403170 on Build GRCh38
rs541993910 Chr.2: 218403372-218403372 on Build GRCh38
rs1001828632 Chr.2: 218403196-218403196 on Build GRCh38
rs767036952 Chr.2: 218403168-218403168 on Build GRCh38
rs765533971 Chr.2: 218403227-218403227 on Build GRCh38
rs759916914 Chr.2: 218403222-218403222 on Build GRCh38
rs2227257 Chr.2: 218403178-218403178 on Build GRCh38
rs1002155078 Chr.2: 218403017-218403017 on Build GRCh38
rs756022832 Chr.2: 218403189-218403189 on Build GRCh38
rs988588684 Chr.2: 218403037-218403037 on Build GRCh38
rs193256483 Chr.2: 218403029-218403029 on Build GRCh38
rs765496351 Chr.2: 218403173-218403173 on Build GRCh38
rs200471933 Chr.2: 218403151-218403151 on Build GRCh38
rs756612276 Chr.2: 218402981-218402981 on Build GRCh38
rs766196233 Chr.2: 218403152-218403152 on Build GRCh38
rs754837135 Chr.2: 218403364-218403364 on Build GRCh38
rs757967575 Chr.2: 218403285-218403285 on Build GRCh38
rs764418232 Chr.2: 218403258-218403258 on Build GRCh38
rs530933090 Chr.2: 218403032-218403032 on Build GRCh38
rs755057198 Chr.2: 218402996-218402996 on Build GRCh38
rs368165173 Chr.2: 218403019-218403019 on Build GRCh38
rs999022783 Chr.2: 218403112-218403112 on Build GRCh38
rs909114293 Chr.2: 218403377-218403377 on Build GRCh38
rs139465180 Chr.2: 218403160-218403160 on Build GRCh38
rs893335202 Chr.2: 218403218-218403218 on Build GRCh38
rs752867676 Chr.2: 218403176-218403176 on Build GRCh38
rs1019708438 Chr.2: 218403287-218403287 on Build GRCh38
rs2227258 Chr.2: 218403190-218403190 on Build GRCh38
rs147970292 Chr.2: 218403341-218403341 on Build GRCh38
rs775191368 Chr.2: 218402965-218402965 on Build GRCh38
rs202177697 Chr.2: 218403310-218403310 on Build GRCh38
rs777349013 Chr.2: 218403217-218403217 on Build GRCh38
rs765265559 Chr.2: 218403365-218403365 on Build GRCh38
rs767958426 Chr.2: 218402979-218402979 on Build GRCh38
rs760039671 Chr.2: 218403171-218403171 on Build GRCh38
rs779169110 Chr.2: 218403000-218403000 on Build GRCh38
rs752424152 Chr.2: 218403367-218403367 on Build GRCh38
rs35551716 Chr.2: 218403399-218403399 on Build GRCh38
rs763607465 Chr.2: 218402969-218402969 on Build GRCh38
rs142262587 Chr.2: 218403121-218403121 on Build GRCh38
rs773887679 Chr.2: 218403025-218403025 on Build GRCh38
rs745672299 Chr.2: 218403009-218403009 on Build GRCh38
rs763774922 Chr.2: 218403023-218403023 on Build GRCh38
rs953030217 Chr.2: 218403172-218403172 on Build GRCh38
rs570745354 Chr.2: 218403070-218403070 on Build GRCh38
rs750895624 Chr.2: 218403030-218403030 on Build GRCh38
rs777954344 Chr.2: 218403026-218403027 on Build GRCh38
rs903304393 Chr.2: 218403071-218403071 on Build GRCh38
rs374746834 Chr.2: 218403007-218403007 on Build GRCh38
rs2227254 Chr.2: 218402967-218402967 on Build GRCh38
rs971884192 Chr.2: 218403397-218403397 on Build GRCh38
rs908957688 Chr.2: 218403180-218403180 on Build GRCh38
rs754998014 Chr.2: 218403016-218403016 on Build GRCh38
rs781043176 Chr.2: 218403167-218403167 on Build GRCh38
rs774559740 Chr.2: 218403348-218403348 on Build GRCh38
rs754925266 Chr.2: 218403161-218403161 on Build GRCh38
rs757504481 Chr.2: 218403304-218403304 on Build GRCh38
rs746677198 Chr.2: 218403393-218403393 on Build GRCh38
rs112546602 Chr.2: 218402951-218402951 on Build GRCh38
rs777629081 Chr.2: 218403288-218403288 on Build GRCh38
rs749396916 Chr.2: 218403139-218403140 on Build GRCh38
rs746384440 Chr.2: 218403312-218403312 on Build GRCh38
rs761318742 Chr.2: 218403162-218403162 on Build GRCh38
rs2227256 Chr.2: 218403147-218403147 on Build GRCh38
rs748219897 Chr.2: 218403006-218403006 on Build GRCh38
rs1048893436 Chr.2: 218402956-218402956 on Build GRCh38
rs777740284 Chr.2: 218403179-218403179 on Build GRCh38
rs751962266 Chr.2: 218402975-218402975 on Build GRCh38
rs541352179 Chr.2: 218402978-218402978 on Build GRCh38
rs569783106 Chr.2: 218403031-218403031 on Build GRCh38
rs775687462 Chr.2: 218403324-218403324 on Build GRCh38
rs146608193 Chr.2: 218403276-218403276 on Build GRCh38
rs371054585 Chr.2: 218403359-218403359 on Build GRCh38
rs1010430010 Chr.2: 218403219-218403219 on Build GRCh38
rs539235122 Chr.2: 218403083-218403083 on Build GRCh38
rs766237091 Chr.2: 218403271-218403271 on Build GRCh38
rs535608825 Chr.2: 218403164-218403164 on Build GRCh38
rs149257024 Chr.2: 218403294-218403294 on Build GRCh38
rs910841273 Chr.2: 218402998-218403016 on Build GRCh38
rs759301870 Chr.2: 218402962-218402962 on Build GRCh38
rs779055787 Chr.2: 218403061-218403061 on Build GRCh38
rs370393498 Chr.2: 218403199-218403199 on Build GRCh38
rs746983928 Chr.2: 218402995-218403013 on Build GRCh38
rs781118277 Chr.2: 218403137-218403137 on Build GRCh38
rs1056099853 Chr.2: 218403212-218403213 on Build GRCh38

View Full Product Details