Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218404596-218405102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTDSP1
Entrez Gene ID
58190
Gene Name
CTD small phosphatase 1
Gene Aliases
NIF3, NLI-IF, NLIIF, SCP1
UniGene
Hs.444468
Gene Chromosome Location
Chr.2: 218398338-218405941 on Build GRCh38
Transcripts
NM_001206878.1 NM_021198.2 NM_182642.2 XM_011511569.2 XM_011511570.2

SNP Detail

SNP ID Chromosome Location
rs933699200 Chr.2: 218404795-218404795 on Build GRCh38
rs969558083 Chr.2: 218404713-218404713 on Build GRCh38
rs979656916 Chr.2: 218404715-218404715 on Build GRCh38
rs951006939 Chr.2: 218404853-218404853 on Build GRCh38
rs1032591776 Chr.2: 218404972-218404972 on Build GRCh38
rs979630549 Chr.2: 218404937-218404938 on Build GRCh38
rs911001883 Chr.2: 218405057-218405057 on Build GRCh38
rs540092670 Chr.2: 218404815-218404815 on Build GRCh38
rs925554825 Chr.2: 218404729-218404729 on Build GRCh38
rs950874351 Chr.2: 218404692-218404692 on Build GRCh38
rs1030398883 Chr.2: 218404625-218404625 on Build GRCh38
rs1039083821 Chr.2: 218405065-218405065 on Build GRCh38
rs1007573194 Chr.2: 218404630-218404630 on Build GRCh38
rs957136822 Chr.2: 218404973-218404973 on Build GRCh38
rs998048654 Chr.2: 218404816-218404816 on Build GRCh38
rs551780824 Chr.2: 218404626-218404626 on Build GRCh38
rs1014059136 Chr.2: 218404873-218404873 on Build GRCh38
rs112815236 Chr.2: 218404774-218404774 on Build GRCh38
rs747085491 Chr.2: 218404830-218404830 on Build GRCh38
rs146891368 Chr.2: 218404832-218404832 on Build GRCh38
rs768827995 Chr.2: 218404866-218404866 on Build GRCh38
rs933869145 Chr.2: 218405026-218405026 on Build GRCh38
rs889626520 Chr.2: 218404622-218404622 on Build GRCh38
rs140714484 Chr.2: 218404991-218404991 on Build GRCh38
rs764024694 Chr.2: 218405066-218405066 on Build GRCh38
rs935645407 Chr.2: 218404740-218404740 on Build GRCh38
rs969532425 Chr.2: 218404886-218404886 on Build GRCh38
rs189039843 Chr.2: 218404757-218404757 on Build GRCh38
rs770639533 Chr.2: 218404808-218404808 on Build GRCh38
rs567065861 Chr.2: 218404650-218404650 on Build GRCh38
rs916715805 Chr.2: 218404677-218404677 on Build GRCh38
rs537434161 Chr.2: 218404660-218404660 on Build GRCh38
rs992257328 Chr.2: 218404668-218404669 on Build GRCh38
rs889616446 Chr.2: 218405051-218405051 on Build GRCh38
rs919463044 Chr.2: 218404667-218404667 on Build GRCh38
rs548667506 Chr.2: 218404649-218404649 on Build GRCh38
rs537563550 Chr.2: 218405017-218405017 on Build GRCh38
rs914870186 Chr.2: 218404758-218404758 on Build GRCh38
rs896495238 Chr.2: 218404855-218404855 on Build GRCh38
rs757396180 Chr.2: 218405068-218405068 on Build GRCh38
rs879204867 Chr.2: 218404912-218404912 on Build GRCh38
rs754036246 Chr.2: 218405067-218405067 on Build GRCh38
rs573427977 Chr.2: 218404836-218404836 on Build GRCh38
rs540875603 Chr.2: 218404847-218404847 on Build GRCh38
rs1026028365 Chr.2: 218404850-218404850 on Build GRCh38
rs990972020 Chr.2: 218404949-218404949 on Build GRCh38
rs780128794 Chr.2: 218404823-218404823 on Build GRCh38
rs760542708 Chr.2: 218405053-218405053 on Build GRCh38
rs1035562368 Chr.2: 218404915-218404915 on Build GRCh38
rs962665367 Chr.2: 218404643-218404643 on Build GRCh38
rs946654283 Chr.2: 218404763-218404763 on Build GRCh38
rs982314106 Chr.2: 218404876-218404876 on Build GRCh38
rs914943830 Chr.2: 218404969-218404969 on Build GRCh38
rs1026503465 Chr.2: 218404848-218404848 on Build GRCh38
rs759376293 Chr.2: 218404936-218404936 on Build GRCh38
rs927751856 Chr.2: 218404732-218404732 on Build GRCh38
rs995081715 Chr.2: 218404840-218404840 on Build GRCh38
rs1050876970 Chr.2: 218404809-218404809 on Build GRCh38
rs1050951211 Chr.2: 218405043-218405043 on Build GRCh38
rs758275905 Chr.2: 218404743-218404743 on Build GRCh38
rs555887719 Chr.2: 218404902-218404902 on Build GRCh38
rs946351719 Chr.2: 218405005-218405005 on Build GRCh38
rs3770211 Chr.2: 218405006-218405006 on Build GRCh38
rs982290512 Chr.2: 218404712-218404712 on Build GRCh38
rs1017990226 Chr.2: 218404641-218404641 on Build GRCh38
rs369176318 Chr.2: 218404708-218404708 on Build GRCh38
rs963342962 Chr.2: 218404837-218404837 on Build GRCh38
rs889590684 Chr.2: 218404818-218404818 on Build GRCh38
rs937727542 Chr.2: 218404756-218404756 on Build GRCh38

View Full Product Details