Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218659938-218660434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCS1L
Entrez Gene ID
617
Gene Name
BCS1 homolog, ubiquinol-cytochrome c reductase complex chaperone
Gene Aliases
BCS, BCS1, BJS, FLNMS, GRACILE, Hs.6719, MC3DN1, PTD, h-BCS, h-BCS1
UniGene
Hs.471401
Gene Chromosome Location
Chr.2: 218659656-218663443 on Build GRCh38
Transcripts
NM_001079866.1 NM_001257342.1 NM_001257343.1 NM_001257344.1 NM_001318836.1 NM_001320717.1 NM_004328.4 XM_005246748.2 XM_005246749.3 XM_006712678.1 XM_017004631.1 XM_017004632.1 XM_017004633.1

SNP Detail

SNP ID Chromosome Location
rs1000352446 Chr.2: 218659981-218659981 on Build GRCh38
rs898301590 Chr.2: 218660148-218660148 on Build GRCh38
rs768686955 Chr.2: 218660391-218660391 on Build GRCh38
rs77487601 Chr.2: 218660344-218660344 on Build GRCh38
rs777598708 Chr.2: 218659960-218659960 on Build GRCh38
rs954512080 Chr.2: 218660385-218660385 on Build GRCh38
rs3806556 Chr.2: 218660284-218660284 on Build GRCh38
rs973184953 Chr.2: 218660179-218660179 on Build GRCh38
rs745861723 Chr.2: 218660069-218660069 on Build GRCh38
rs1044431632 Chr.2: 218660344-218660345 on Build GRCh38
rs886055625 Chr.2: 218660168-218660168 on Build GRCh38
rs564613166 Chr.2: 218660030-218660030 on Build GRCh38
rs748920339 Chr.2: 218659965-218659965 on Build GRCh38
rs3821045 Chr.2: 218660375-218660375 on Build GRCh38
rs1042636432 Chr.2: 218660024-218660024 on Build GRCh38
rs537510938 Chr.2: 218660122-218660122 on Build GRCh38
rs985847027 Chr.2: 218660144-218660144 on Build GRCh38
rs993997377 Chr.2: 218660168-218660168 on Build GRCh38
rs1029971116 Chr.2: 218660064-218660064 on Build GRCh38
rs985815476 Chr.2: 218660332-218660332 on Build GRCh38
rs77595651 Chr.2: 218660306-218660306 on Build GRCh38
rs1053253813 Chr.2: 218660365-218660365 on Build GRCh38
rs774164629 Chr.2: 218660025-218660025 on Build GRCh38
rs1031867129 Chr.2: 218660021-218660021 on Build GRCh38
rs138294740 Chr.2: 218660298-218660298 on Build GRCh38
rs963347534 Chr.2: 218660161-218660161 on Build GRCh38
rs548027338 Chr.2: 218659971-218659971 on Build GRCh38
rs1009334495 Chr.2: 218660048-218660048 on Build GRCh38
rs1038421920 Chr.2: 218660143-218660143 on Build GRCh38
rs115162602 Chr.2: 218660409-218660409 on Build GRCh38
rs1017286562 Chr.2: 218660395-218660395 on Build GRCh38
rs910445833 Chr.2: 218660160-218660160 on Build GRCh38
rs889798623 Chr.2: 218660123-218660123 on Build GRCh38
rs1025560137 Chr.2: 218660185-218660185 on Build GRCh38
rs182280364 Chr.2: 218660353-218660353 on Build GRCh38
rs1023637711 Chr.2: 218659969-218659969 on Build GRCh38
rs1021417953 Chr.2: 218660354-218660354 on Build GRCh38
rs188224298 Chr.2: 218660201-218660201 on Build GRCh38
rs567040894 Chr.2: 218660018-218660018 on Build GRCh38
rs1006907254 Chr.2: 218660131-218660131 on Build GRCh38
rs548744217 Chr.2: 218660001-218660001 on Build GRCh38
rs386833855 Chr.2: 218660400-218660400 on Build GRCh38
rs978688978 Chr.2: 218660379-218660379 on Build GRCh38

View Full Product Details