Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218661225-218661729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCS1L
Entrez Gene ID
617
Gene Name
BCS1 homolog, ubiquinol-cytochrome c reductase complex chaperone
Gene Aliases
BCS, BCS1, BJS, FLNMS, GRACILE, Hs.6719, MC3DN1, PTD, h-BCS, h-BCS1
UniGene
Hs.471401
Gene Chromosome Location
Chr.2: 218659656-218663443 on Build GRCh38
Transcripts
NM_001079866.1 NM_001257342.1 NM_001257343.1 NM_001257344.1 NM_001318836.1 NM_001320717.1 NM_004328.4 XM_005246748.2 XM_005246749.3 XM_006712678.1 XM_017004631.1 XM_017004632.1 XM_017004633.1 XM_017004634.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5058668 c.288A>T p.E96D Chr.2: 218661275-218661275 on Build GRCh38
6587856 c.321-3C>G p.? Chr.2: 218661403-218661403 on Build GRCh38
4985570 c.301C>T p.P101S Chr.2: 218661288-218661288 on Build GRCh38
7266888 c.460+7G>A p.? Chr.2: 218661552-218661552 on Build GRCh38
4570501 c.279T>C p.T93T Chr.2: 218661266-218661266 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1001355643 Chr.2: 218661677-218661677 on Build GRCh38
rs957463359 Chr.2: 218661393-218661393 on Build GRCh38
rs762566470 Chr.2: 218661641-218661641 on Build GRCh38
rs756708393 Chr.2: 218661555-218661555 on Build GRCh38
rs1032863083 Chr.2: 218661697-218661697 on Build GRCh38
rs748561443 Chr.2: 218661485-218661486 on Build GRCh38
rs1027271702 Chr.2: 218661469-218661469 on Build GRCh38
rs376836925 Chr.2: 218661296-218661296 on Build GRCh38
rs1057517412 Chr.2: 218661503-218661503 on Build GRCh38
rs755347184 Chr.2: 218661374-218661377 on Build GRCh38
rs199851412 Chr.2: 218661499-218661499 on Build GRCh38
rs746640681 Chr.2: 218661435-218661435 on Build GRCh38
rs543586069 Chr.2: 218661703-218661703 on Build GRCh38
rs750506359 Chr.2: 218661416-218661416 on Build GRCh38
rs141257714 Chr.2: 218661410-218661410 on Build GRCh38
rs775911546 Chr.2: 218661381-218661381 on Build GRCh38
rs776363896 Chr.2: 218661394-218661394 on Build GRCh38
rs752688266 Chr.2: 218661426-218661426 on Build GRCh38
rs775793638 Chr.2: 218661498-218661498 on Build GRCh38
rs375934184 Chr.2: 218661320-218661320 on Build GRCh38
rs149382836 Chr.2: 218661411-218661411 on Build GRCh38
rs1057516954 Chr.2: 218661547-218661547 on Build GRCh38
rs369691608 Chr.2: 218661255-218661255 on Build GRCh38
rs776055199 Chr.2: 218661302-218661302 on Build GRCh38
rs777735526 Chr.2: 218661434-218661434 on Build GRCh38
rs879132075 Chr.2: 218661646-218661646 on Build GRCh38
rs765732284 Chr.2: 218661340-218661340 on Build GRCh38
rs780752138 Chr.2: 218661468-218661468 on Build GRCh38
rs531442370 Chr.2: 218661656-218661656 on Build GRCh38
rs772937517 Chr.2: 218661267-218661267 on Build GRCh38
rs930414096 Chr.2: 218661611-218661611 on Build GRCh38
rs772010441 Chr.2: 218661263-218661263 on Build GRCh38
rs982826850 Chr.2: 218661603-218661603 on Build GRCh38
rs760608304 Chr.2: 218661550-218661550 on Build GRCh38
rs542467112 Chr.2: 218661636-218661636 on Build GRCh38
rs767694386 Chr.2: 218661531-218661531 on Build GRCh38
rs572015036 Chr.2: 218661530-218661530 on Build GRCh38
rs747956412 Chr.2: 218661256-218661256 on Build GRCh38
rs377365292 Chr.2: 218661537-218661537 on Build GRCh38
rs386833856 Chr.2: 218661308-218661308 on Build GRCh38
rs1047358116 Chr.2: 218661612-218661612 on Build GRCh38
rs769358314 Chr.2: 218661494-218661494 on Build GRCh38
rs751739625 Chr.2: 218661405-218661405 on Build GRCh38
rs905811405 Chr.2: 218661660-218661661 on Build GRCh38
rs758462006 Chr.2: 218661591-218661591 on Build GRCh38
rs121908572 Chr.2: 218661283-218661283 on Build GRCh38
rs766516213 Chr.2: 218661574-218661574 on Build GRCh38
rs757128513 Chr.2: 218661647-218661647 on Build GRCh38
rs746693506 Chr.2: 218661274-218661274 on Build GRCh38
rs768958849 Chr.2: 218661505-218661505 on Build GRCh38
rs988786862 Chr.2: 218661395-218661395 on Build GRCh38
rs764447616 Chr.2: 218661399-218661399 on Build GRCh38
rs754934987 Chr.2: 218661421-218661421 on Build GRCh38
rs751484879 Chr.2: 218661483-218661483 on Build GRCh38
rs879252201 Chr.2: 218661624-218661624 on Build GRCh38
rs778769841 Chr.2: 218661425-218661425 on Build GRCh38
rs750499133 Chr.2: 218661535-218661535 on Build GRCh38
rs752635954 Chr.2: 218661579-218661579 on Build GRCh38
rs386833857 Chr.2: 218661516-218661516 on Build GRCh38
rs770749420 Chr.2: 218661440-218661440 on Build GRCh38
rs745428773 Chr.2: 218661477-218661477 on Build GRCh38
rs138756092 Chr.2: 218661290-218661290 on Build GRCh38
rs560845438 Chr.2: 218661640-218661640 on Build GRCh38
rs866209645 Chr.2: 218661281-218661281 on Build GRCh38
rs1033088624 Chr.2: 218661361-218661361 on Build GRCh38

View Full Product Details