Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218814466-218814966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP27A1
Entrez Gene ID
1593
Gene Name
cytochrome P450 family 27 subfamily A member 1
Gene Aliases
CP27, CTX, CYP27
UniGene
Hs.516700
Gene Chromosome Location
Chr.2: 218781006-218815293 on Build GRCh38
Transcripts
NM_000784.3 XM_017003488.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3044092 c.1471G>A p.A491T Chr.2: 218814752-218814752 on Build GRCh38
5809608 c.1346G>T p.W449L Chr.2: 218814627-218814627 on Build GRCh38
4527884 c.1497G>A p.V499V Chr.2: 218814931-218814931 on Build GRCh38
1016509 c.1343G>A p.R448H Chr.2: 218814624-218814624 on Build GRCh38
5736423 c.1321C>T p.P441S Chr.2: 218814602-218814602 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751264658 Chr.2: 218814845-218814845 on Build GRCh38
rs760024535 Chr.2: 218814613-218814613 on Build GRCh38
rs757381848 Chr.2: 218814554-218814554 on Build GRCh38
rs199638075 Chr.2: 218814717-218814717 on Build GRCh38
rs919410960 Chr.2: 218814510-218814510 on Build GRCh38
rs587778787 Chr.2: 218814683-218814683 on Build GRCh38
rs72551322 Chr.2: 218814716-218814716 on Build GRCh38
rs577558935 Chr.2: 218814943-218814943 on Build GRCh38
rs370868184 Chr.2: 218814624-218814624 on Build GRCh38
rs755665162 Chr.2: 218814907-218814907 on Build GRCh38
rs774127681 Chr.2: 218814567-218814567 on Build GRCh38
rs562478738 Chr.2: 218814502-218814502 on Build GRCh38
rs763270290 Chr.2: 218814489-218814498 on Build GRCh38
rs200883871 Chr.2: 218814696-218814696 on Build GRCh38
rs765660985 Chr.2: 218814615-218814615 on Build GRCh38
rs770435184 Chr.2: 218814691-218814691 on Build GRCh38
rs879088060 Chr.2: 218814656-218814659 on Build GRCh38
rs778132806 Chr.2: 218814518-218814518 on Build GRCh38
rs78938783 Chr.2: 218814908-218814908 on Build GRCh38
rs754657308 Chr.2: 218814862-218814862 on Build GRCh38
rs373520260 Chr.2: 218814492-218814492 on Build GRCh38
rs1051581449 Chr.2: 218814874-218814874 on Build GRCh38
rs1008075672 Chr.2: 218814706-218814706 on Build GRCh38
rs146577883 Chr.2: 218814854-218814854 on Build GRCh38
rs373870129 Chr.2: 218814650-218814650 on Build GRCh38
rs751408247 Chr.2: 218814873-218814873 on Build GRCh38
rs184930780 Chr.2: 218814809-218814809 on Build GRCh38
rs760161031 Chr.2: 218814499-218814499 on Build GRCh38
rs773501493 Chr.2: 218814938-218814938 on Build GRCh38
rs573781554 Chr.2: 218814839-218814839 on Build GRCh38
rs890298373 Chr.2: 218814906-218814906 on Build GRCh38
rs763497885 Chr.2: 218814782-218814782 on Build GRCh38
rs139279260 Chr.2: 218814715-218814715 on Build GRCh38
rs55998945 Chr.2: 218814655-218814655 on Build GRCh38
rs549032526 Chr.2: 218814694-218814694 on Build GRCh38
rs768346937 Chr.2: 218814566-218814566 on Build GRCh38
rs112509525 Chr.2: 218814491-218814491 on Build GRCh38
rs377290147 Chr.2: 218814928-218814928 on Build GRCh38
rs200106364 Chr.2: 218814741-218814741 on Build GRCh38
rs770631423 Chr.2: 218814744-218814744 on Build GRCh38
rs774920989 Chr.2: 218814792-218814792 on Build GRCh38
rs769144632 Chr.2: 218814785-218814785 on Build GRCh38
rs777141854 Chr.2: 218814609-218814609 on Build GRCh38
rs755434730 Chr.2: 218814792-218814792 on Build GRCh38
rs72551323 Chr.2: 218814752-218814752 on Build GRCh38
rs527395716 Chr.2: 218814637-218814637 on Build GRCh38
rs776087607 Chr.2: 218814775-218814775 on Build GRCh38
rs1038115413 Chr.2: 218814837-218814837 on Build GRCh38
rs753166698 Chr.2: 218814617-218814617 on Build GRCh38
rs769146248 Chr.2: 218814771-218814771 on Build GRCh38
rs201107032 Chr.2: 218814579-218814579 on Build GRCh38
rs926265857 Chr.2: 218814827-218814827 on Build GRCh38
rs986444449 Chr.2: 218814824-218814824 on Build GRCh38
rs730882199 Chr.2: 218814623-218814623 on Build GRCh38
rs977947148 Chr.2: 218814763-218814763 on Build GRCh38
rs748563770 Chr.2: 218814923-218814923 on Build GRCh38
rs1024898619 Chr.2: 218814753-218814753 on Build GRCh38
rs776752717 Chr.2: 218814942-218814942 on Build GRCh38
rs777432192 Chr.2: 218814675-218814675 on Build GRCh38
rs587778785 Chr.2: 218814544-218814544 on Build GRCh38
rs776275363 Chr.2: 218814693-218814693 on Build GRCh38
rs371493850 Chr.2: 218814507-218814507 on Build GRCh38
rs748020709 Chr.2: 218814660-218814660 on Build GRCh38
rs774975996 Chr.2: 218814911-218814911 on Build GRCh38
rs138596741 Chr.2: 218814700-218814700 on Build GRCh38
rs765832998 Chr.2: 218814596-218814596 on Build GRCh38
rs372756101 Chr.2: 218814901-218814901 on Build GRCh38
rs188379599 Chr.2: 218814515-218814515 on Build GRCh38
rs781360671 Chr.2: 218814654-218814654 on Build GRCh38
rs772825536 Chr.2: 218814581-218814581 on Build GRCh38
rs1039198612 Chr.2: 218814629-218814629 on Build GRCh38
rs150585977 Chr.2: 218814737-218814737 on Build GRCh38
rs371728112 Chr.2: 218814682-218814682 on Build GRCh38
rs757719396 Chr.2: 218814822-218814822 on Build GRCh38
rs752869724 Chr.2: 218814512-218814512 on Build GRCh38
rs121908098 Chr.2: 218814701-218814701 on Build GRCh38
rs933888044 Chr.2: 218814796-218814796 on Build GRCh38
rs780929951 Chr.2: 218814898-218814898 on Build GRCh38
rs762253767 Chr.2: 218814798-218814798 on Build GRCh38
rs121908097 Chr.2: 218814702-218814702 on Build GRCh38
rs140842830 Chr.2: 218814939-218814939 on Build GRCh38
rs368499036 Chr.2: 218814789-218814789 on Build GRCh38
rs752231649 Chr.2: 218814931-218814932 on Build GRCh38
rs764008118 Chr.2: 218814872-218814872 on Build GRCh38
rs758400055 Chr.2: 218814871-218814871 on Build GRCh38
rs781778588 Chr.2: 218814913-218814943 on Build GRCh38
rs886044581 Chr.2: 218814592-218814592 on Build GRCh38
rs200977221 Chr.2: 218814523-218814523 on Build GRCh38
rs749202662 Chr.2: 218814563-218814563 on Build GRCh38
rs766481873 Chr.2: 218814941-218814941 on Build GRCh38
rs143002163 Chr.2: 218814578-218814578 on Build GRCh38
rs116288589 Chr.2: 218814829-218814829 on Build GRCh38
rs755008787 Chr.2: 218814656-218814656 on Build GRCh38
rs771819245 Chr.2: 218814662-218814662 on Build GRCh38
rs200450104 Chr.2: 218814828-218814828 on Build GRCh38
rs367920782 Chr.2: 218814593-218814593 on Build GRCh38
rs751969035 Chr.2: 218814650-218814651 on Build GRCh38
rs767829467 Chr.2: 218814807-218814807 on Build GRCh38
rs1027016990 Chr.2: 218814695-218814695 on Build GRCh38
rs766603950 Chr.2: 218814509-218814510 on Build GRCh38
rs376750859 Chr.2: 218814506-218814506 on Build GRCh38
rs980944460 Chr.2: 218814812-218814812 on Build GRCh38
rs752690404 Chr.2: 218814865-218814865 on Build GRCh38
rs886703307 Chr.2: 218814893-218814893 on Build GRCh38
rs111570247 Chr.2: 218814909-218814909 on Build GRCh38
rs757019811 Chr.2: 218814891-218814891 on Build GRCh38
rs747135158 Chr.2: 218814520-218814520 on Build GRCh38
rs780172950 Chr.2: 218814556-218814556 on Build GRCh38

View Full Product Details