Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218990083-218990590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYBA2
Entrez Gene ID
1412
Gene Name
crystallin beta A2
Gene Aliases
CTRCT42
UniGene
Hs.415790
Gene Chromosome Location
Chr.2: 218990190-218993405 on Build GRCh38
Transcripts
NM_057093.1 NM_057094.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1016529 c.575T>C p.I192T Chr.2: 218990271-218990271 on Build GRCh38
6635840 c.504C>T p.D168D Chr.2: 218990342-218990342 on Build GRCh38
4977781 c.577C>T p.R193W Chr.2: 218990269-218990269 on Build GRCh38
1016530 c.513C>T p.S171S Chr.2: 218990333-218990333 on Build GRCh38
6635838 c.517G>T p.E173* Chr.2: 218990329-218990329 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747774568 Chr.2: 218990390-218990390 on Build GRCh38
rs1010828571 Chr.2: 218990296-218990296 on Build GRCh38
rs1045829512 Chr.2: 218990408-218990408 on Build GRCh38
rs1020230777 Chr.2: 218990444-218990444 on Build GRCh38
rs750405127 Chr.2: 218990371-218990371 on Build GRCh38
rs911559991 Chr.2: 218990108-218990108 on Build GRCh38
rs533278076 Chr.2: 218990133-218990133 on Build GRCh38
rs375813911 Chr.2: 218990322-218990322 on Build GRCh38
rs369927720 Chr.2: 218990443-218990443 on Build GRCh38
rs897936346 Chr.2: 218990103-218990103 on Build GRCh38
rs762255352 Chr.2: 218990261-218990261 on Build GRCh38
rs867811553 Chr.2: 218990195-218990195 on Build GRCh38
rs752848980 Chr.2: 218990358-218990358 on Build GRCh38
rs141747065 Chr.2: 218990306-218990306 on Build GRCh38
rs144661907 Chr.2: 218990275-218990275 on Build GRCh38
rs1010487031 Chr.2: 218990476-218990476 on Build GRCh38
rs373545183 Chr.2: 218990341-218990341 on Build GRCh38
rs893661334 Chr.2: 218990253-218990253 on Build GRCh38
rs754056321 Chr.2: 218990290-218990290 on Build GRCh38
rs778980754 Chr.2: 218990439-218990439 on Build GRCh38
rs373588968 Chr.2: 218990315-218990315 on Build GRCh38
rs772800048 Chr.2: 218990409-218990409 on Build GRCh38
rs902229113 Chr.2: 218990530-218990530 on Build GRCh38
rs1020819648 Chr.2: 218990474-218990474 on Build GRCh38
rs563054179 Chr.2: 218990132-218990132 on Build GRCh38
rs368299657 Chr.2: 218990440-218990440 on Build GRCh38
rs779872453 Chr.2: 218990319-218990319 on Build GRCh38
rs530965093 Chr.2: 218990332-218990332 on Build GRCh38
rs369876527 Chr.2: 218990208-218990208 on Build GRCh38
rs759684232 Chr.2: 218990346-218990346 on Build GRCh38
rs370422798 Chr.2: 218990336-218990336 on Build GRCh38
rs748194398 Chr.2: 218990305-218990305 on Build GRCh38
rs1042303489 Chr.2: 218990511-218990511 on Build GRCh38
rs375552220 Chr.2: 218990389-218990389 on Build GRCh38
rs974618256 Chr.2: 218990119-218990119 on Build GRCh38
rs566777591 Chr.2: 218990186-218990186 on Build GRCh38
rs777799274 Chr.2: 218990370-218990370 on Build GRCh38
rs771335800 Chr.2: 218990378-218990378 on Build GRCh38
rs927339879 Chr.2: 218990436-218990436 on Build GRCh38
rs749042754 Chr.2: 218990379-218990379 on Build GRCh38
rs770817564 Chr.2: 218990238-218990238 on Build GRCh38
rs966525239 Chr.2: 218990289-218990289 on Build GRCh38
rs776874617 Chr.2: 218990432-218990432 on Build GRCh38
rs544465368 Chr.2: 218990504-218990504 on Build GRCh38
rs771404920 Chr.2: 218990430-218990430 on Build GRCh38
rs907957193 Chr.2: 218990190-218990190 on Build GRCh38
rs567587351 Chr.2: 218990245-218990245 on Build GRCh38
rs964621510 Chr.2: 218990113-218990113 on Build GRCh38
rs773859071 Chr.2: 218990258-218990258 on Build GRCh38
rs893752799 Chr.2: 218990428-218990428 on Build GRCh38
rs976817170 Chr.2: 218990299-218990299 on Build GRCh38
rs201412404 Chr.2: 218990271-218990271 on Build GRCh38
rs771707941 Chr.2: 218990335-218990335 on Build GRCh38
rs773899013 Chr.2: 218990338-218990338 on Build GRCh38
rs755180140 Chr.2: 218990297-218990297 on Build GRCh38
rs1021105498 Chr.2: 218990287-218990287 on Build GRCh38
rs746009855 Chr.2: 218990445-218990445 on Build GRCh38
rs776413819 Chr.2: 218990243-218990243 on Build GRCh38
rs761296957 Chr.2: 218990412-218990412 on Build GRCh38
rs184853949 Chr.2: 218990244-218990244 on Build GRCh38
rs761469307 Chr.2: 218990333-218990333 on Build GRCh38
rs574001450 Chr.2: 218990468-218990468 on Build GRCh38
rs77427424 Chr.2: 218990340-218990340 on Build GRCh38
rs891280523 Chr.2: 218990529-218990529 on Build GRCh38
rs762632040 Chr.2: 218990248-218990248 on Build GRCh38
rs373198900 Chr.2: 218990239-218990239 on Build GRCh38
rs746944455 Chr.2: 218990226-218990226 on Build GRCh38
rs1046429112 Chr.2: 218990152-218990152 on Build GRCh38
rs758539282 Chr.2: 218990361-218990361 on Build GRCh38
rs149014666 Chr.2: 218990550-218990550 on Build GRCh38
rs920483418 Chr.2: 218990154-218990154 on Build GRCh38
rs771761247 Chr.2: 218990396-218990396 on Build GRCh38
rs748911121 Chr.2: 218990320-218990320 on Build GRCh38
rs997950654 Chr.2: 218990547-218990547 on Build GRCh38
rs377757379 Chr.2: 218990450-218990450 on Build GRCh38
rs113372763 Chr.2: 218990106-218990106 on Build GRCh38
rs781162289 Chr.2: 218990314-218990314 on Build GRCh38
rs150303676 Chr.2: 218990269-218990269 on Build GRCh38
rs534167945 Chr.2: 218990188-218990188 on Build GRCh38
rs1046794886 Chr.2: 218990373-218990373 on Build GRCh38
rs893428663 Chr.2: 218990462-218990462 on Build GRCh38
rs765440590 Chr.2: 218990347-218990347 on Build GRCh38
rs950200832 Chr.2: 218990402-218990402 on Build GRCh38
rs778743003 Chr.2: 218990554-218990554 on Build GRCh38
rs768324556 Chr.2: 218990386-218990386 on Build GRCh38
rs756015791 Chr.2: 218990418-218990418 on Build GRCh38
rs371047040 Chr.2: 218990255-218990255 on Build GRCh38
rs778892229 Chr.2: 218990303-218990303 on Build GRCh38
rs752630962 Chr.2: 218990392-218990395 on Build GRCh38
rs770881949 Chr.2: 218990398-218990398 on Build GRCh38
rs768082090 Chr.2: 218990267-218990267 on Build GRCh38
rs370006179 Chr.2: 218990182-218990182 on Build GRCh38

View Full Product Details