Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219209624-219210120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB6
Entrez Gene ID
10058
Gene Name
ATP binding cassette subfamily B member 6 (Langereis blood group)
Gene Aliases
ABC, ABC14, DUH3, LAN, MCOPCB7, MTABC3, PRP, PSHK2, umat
UniGene
Hs.107911
Gene Chromosome Location
Chr.2: 219209766-219218990 on Build GRCh38
Transcripts
NM_005689.2 NM_001270998.1 NM_001270999.1 NM_138802.2 XM_006712285.1 XM_006712286.3 XM_017003368.1

Gene Symbol
ZFAND2B
Entrez Gene ID
130617
Gene Name
zinc finger AN1-type containing 2B
Gene Aliases
AIRAPL
UniGene
Hs.534540
Gene Chromosome Location
Chr.2: 219206784-219209651 on Build GRCh38
Transcripts
NM_005689.2 NM_001270998.1 NM_001270999.1 NM_138802.2 XM_006712285.1 XM_006712286.3 XM_017003368.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6584311 c.2512C>G p.Q838E Chr.2: 219209955-219209955 on Build GRCh38
4658214 c.2483A>G p.Q828R Chr.2: 219209984-219209984 on Build GRCh38
1016602 c.2466G>A p.W822* Chr.2: 219210001-219210001 on Build GRCh38
5744761 c.2524C>T p.R842W Chr.2: 219209943-219209943 on Build GRCh38
1016604 c.2447G>A p.G816E Chr.2: 219210020-219210020 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761325184 Chr.2: 219210043-219210043 on Build GRCh38
rs763530773 Chr.2: 219210092-219210092 on Build GRCh38
rs373393094 Chr.2: 219210014-219210014 on Build GRCh38
rs139308368 Chr.2: 219210033-219210033 on Build GRCh38
rs553417258 Chr.2: 219209695-219209695 on Build GRCh38
rs763725814 Chr.2: 219209954-219209954 on Build GRCh38
rs878990668 Chr.2: 219210061-219210061 on Build GRCh38
rs756094735 Chr.2: 219210084-219210084 on Build GRCh38
rs944164593 Chr.2: 219209926-219209926 on Build GRCh38
rs959807531 Chr.2: 219209770-219209770 on Build GRCh38
rs750962689 Chr.2: 219209966-219209966 on Build GRCh38
rs988296057 Chr.2: 219209892-219209892 on Build GRCh38
rs766977785 Chr.2: 219210052-219210052 on Build GRCh38
rs780047348 Chr.2: 219209988-219209988 on Build GRCh38
rs778732570 Chr.2: 219209816-219209816 on Build GRCh38
rs202011349 Chr.2: 219210093-219210093 on Build GRCh38
rs752982431 Chr.2: 219210064-219210064 on Build GRCh38
rs387906910 Chr.2: 219210036-219210036 on Build GRCh38
rs545514319 Chr.2: 219209745-219209745 on Build GRCh38
rs756521360 Chr.2: 219209967-219209967 on Build GRCh38
rs765654061 Chr.2: 219210060-219210060 on Build GRCh38
rs922358325 Chr.2: 219209962-219209962 on Build GRCh38
rs767772482 Chr.2: 219209662-219209665 on Build GRCh38
rs755359852 Chr.2: 219210058-219210058 on Build GRCh38
rs543774852 Chr.2: 219210026-219210026 on Build GRCh38
rs147604992 Chr.2: 219209722-219209722 on Build GRCh38
rs559375312 Chr.2: 219209800-219209800 on Build GRCh38
rs377572280 Chr.2: 219210031-219210031 on Build GRCh38
rs1053289 Chr.2: 219209924-219209924 on Build GRCh38
rs755184076 Chr.2: 219209655-219209655 on Build GRCh38
rs1035547817 Chr.2: 219210019-219210019 on Build GRCh38
rs988348533 Chr.2: 219210096-219210096 on Build GRCh38
rs745749208 Chr.2: 219209925-219209925 on Build GRCh38
rs761310544 Chr.2: 219210049-219210049 on Build GRCh38
rs772531740 Chr.2: 219210048-219210048 on Build GRCh38
rs769650201 Chr.2: 219209935-219209935 on Build GRCh38
rs748245425 Chr.2: 219209896-219209896 on Build GRCh38
rs766853293 Chr.2: 219209997-219209997 on Build GRCh38
rs539822413 Chr.2: 219209964-219209964 on Build GRCh38
rs1004119780 Chr.2: 219210015-219210015 on Build GRCh38
rs575493086 Chr.2: 219209732-219209732 on Build GRCh38
rs1050392602 Chr.2: 219209670-219209670 on Build GRCh38
rs568168383 Chr.2: 219209893-219209893 on Build GRCh38
rs762664642 Chr.2: 219209942-219209942 on Build GRCh38
rs553140087 Chr.2: 219210018-219210018 on Build GRCh38
rs369553433 Chr.2: 219209980-219209980 on Build GRCh38
rs754240082 Chr.2: 219210054-219210056 on Build GRCh38
rs73993544 Chr.2: 219210071-219210071 on Build GRCh38
rs1023094614 Chr.2: 219209818-219209818 on Build GRCh38
rs770820009 Chr.2: 219209904-219209904 on Build GRCh38
rs756974513 Chr.2: 219209809-219209809 on Build GRCh38
rs745827559 Chr.2: 219210027-219210027 on Build GRCh38
rs925531635 Chr.2: 219209849-219209849 on Build GRCh38
rs1049959061 Chr.2: 219210017-219210017 on Build GRCh38
rs750635453 Chr.2: 219209846-219209846 on Build GRCh38
rs555725074 Chr.2: 219209728-219209728 on Build GRCh38
rs548354046 Chr.2: 219209841-219209841 on Build GRCh38
rs754250687 Chr.2: 219210002-219210002 on Build GRCh38
rs112949641 Chr.2: 219209666-219209666 on Build GRCh38
rs755306736 Chr.2: 219210004-219210004 on Build GRCh38
rs140892945 Chr.2: 219209736-219209736 on Build GRCh38
rs753026092 Chr.2: 219209933-219209934 on Build GRCh38
rs182422937 Chr.2: 219209737-219209737 on Build GRCh38
rs756425793 Chr.2: 219210005-219210006 on Build GRCh38
rs188740431 Chr.2: 219209742-219209742 on Build GRCh38
rs1005958470 Chr.2: 219209701-219209701 on Build GRCh38
rs199873911 Chr.2: 219209951-219209951 on Build GRCh38
rs17783626 Chr.2: 219209720-219209720 on Build GRCh38
rs11538185 Chr.2: 219209872-219209872 on Build GRCh38
rs372899405 Chr.2: 219209955-219209955 on Build GRCh38
rs980103316 Chr.2: 219209836-219209836 on Build GRCh38
rs1035118105 Chr.2: 219209740-219209740 on Build GRCh38
rs141144142 Chr.2: 219210038-219210038 on Build GRCh38
rs767517641 Chr.2: 219210048-219210049 on Build GRCh38
rs754302020 Chr.2: 219210057-219210057 on Build GRCh38
rs193300004 Chr.2: 219209912-219209912 on Build GRCh38
rs768341073 Chr.2: 219210042-219210042 on Build GRCh38
rs886995685 Chr.2: 219209983-219209983 on Build GRCh38
rs996987566 Chr.2: 219209669-219209669 on Build GRCh38
rs777660932 Chr.2: 219210024-219210024 on Build GRCh38
rs751581597 Chr.2: 219210081-219210081 on Build GRCh38
rs150160163 Chr.2: 219209825-219209825 on Build GRCh38
rs1022681189 Chr.2: 219210068-219210068 on Build GRCh38
rs375057553 Chr.2: 219209957-219209957 on Build GRCh38
rs1035452750 Chr.2: 219209756-219209756 on Build GRCh38
rs145339431 Chr.2: 219209936-219209936 on Build GRCh38

View Full Product Details