Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219213737-219214229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB6
Entrez Gene ID
10058
Gene Name
ATP binding cassette subfamily B member 6 (Langereis blood group)
Gene Aliases
ABC, ABC14, DUH3, LAN, MCOPCB7, MTABC3, PRP, PSHK2, umat
UniGene
Hs.107911
Gene Chromosome Location
Chr.2: 219209766-219218990 on Build GRCh38
Transcripts
NM_005689.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4986021 c.1435G>A p.A479T Chr.2: 219214138-219214138 on Build GRCh38
4435489 c.1404C>T p.A468A Chr.2: 219214169-219214169 on Build GRCh38
1016613 c.1469C>T p.S490L Chr.2: 219213935-219213935 on Build GRCh38
4658219 c.1424G>A p.R475H Chr.2: 219214149-219214149 on Build GRCh38
6590086 c.1429C>T p.R477* Chr.2: 219214144-219214144 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs903808859 Chr.2: 219214084-219214084 on Build GRCh38
rs780162701 Chr.2: 219214138-219214138 on Build GRCh38
rs1000591910 Chr.2: 219214040-219214050 on Build GRCh38
rs774476996 Chr.2: 219213826-219213826 on Build GRCh38
rs779076466 Chr.2: 219214115-219214115 on Build GRCh38
rs375304697 Chr.2: 219213953-219213953 on Build GRCh38
rs572277264 Chr.2: 219214077-219214077 on Build GRCh38
rs768793204 Chr.2: 219213821-219213821 on Build GRCh38
rs777482073 Chr.2: 219213853-219213853 on Build GRCh38
rs751006802 Chr.2: 219213908-219213908 on Build GRCh38
rs1009261984 Chr.2: 219214024-219214024 on Build GRCh38
rs779222692 Chr.2: 219213931-219213931 on Build GRCh38
rs760581554 Chr.2: 219213837-219213837 on Build GRCh38
rs372510577 Chr.2: 219213889-219213889 on Build GRCh38
rs939435403 Chr.2: 219213988-219213988 on Build GRCh38
rs758116183 Chr.2: 219213850-219213850 on Build GRCh38
rs763094760 Chr.2: 219213818-219213818 on Build GRCh38
rs772193917 Chr.2: 219213935-219213935 on Build GRCh38
rs930870288 Chr.2: 219213948-219213948 on Build GRCh38
rs755386922 Chr.2: 219214099-219214099 on Build GRCh38
rs17849341 Chr.2: 219213892-219213892 on Build GRCh38
rs756813864 Chr.2: 219213899-219213899 on Build GRCh38
rs749571222 Chr.2: 219213871-219213871 on Build GRCh38
rs777349105 Chr.2: 219213782-219213782 on Build GRCh38
rs757840060 Chr.2: 219213882-219213882 on Build GRCh38
rs771092973 Chr.2: 219214137-219214137 on Build GRCh38
rs2293080 Chr.2: 219214059-219214059 on Build GRCh38
rs753025645 Chr.2: 219214117-219214117 on Build GRCh38
rs535952795 Chr.2: 219214017-219214017 on Build GRCh38
rs756601629 Chr.2: 219213915-219213915 on Build GRCh38
rs778809252 Chr.2: 219214204-219214204 on Build GRCh38
rs747822533 Chr.2: 219214116-219214116 on Build GRCh38
rs746211931 Chr.2: 219213808-219213808 on Build GRCh38
rs750852272 Chr.2: 219213992-219213992 on Build GRCh38
rs772815355 Chr.2: 219214159-219214159 on Build GRCh38
rs111677240 Chr.2: 219214168-219214168 on Build GRCh38
rs777954726 Chr.2: 219214127-219214127 on Build GRCh38
rs762871169 Chr.2: 219213964-219213964 on Build GRCh38
rs79405980 Chr.2: 219213874-219213874 on Build GRCh38
rs747053444 Chr.2: 219214134-219214134 on Build GRCh38
rs575032343 Chr.2: 219213810-219213810 on Build GRCh38
rs777270402 Chr.2: 219214171-219214171 on Build GRCh38
rs190816106 Chr.2: 219214096-219214096 on Build GRCh38
rs748256942 Chr.2: 219213934-219213934 on Build GRCh38
rs140244896 Chr.2: 219214150-219214150 on Build GRCh38
rs752400873 Chr.2: 219213847-219213847 on Build GRCh38
rs527260048 Chr.2: 219213895-219213895 on Build GRCh38
rs141029409 Chr.2: 219214160-219214160 on Build GRCh38
rs567014317 Chr.2: 219213993-219213993 on Build GRCh38
rs971598448 Chr.2: 219214065-219214065 on Build GRCh38
rs765188273 Chr.2: 219213885-219213885 on Build GRCh38
rs558103501 Chr.2: 219214144-219214144 on Build GRCh38
rs772078524 Chr.2: 219213860-219213861 on Build GRCh38
rs149363094 Chr.2: 219213842-219213842 on Build GRCh38
rs757424774 Chr.2: 219214173-219214173 on Build GRCh38
rs796934839 Chr.2: 219213802-219213802 on Build GRCh38
rs766872448 Chr.2: 219213904-219213904 on Build GRCh38
rs914639932 Chr.2: 219214136-219214136 on Build GRCh38
rs200689831 Chr.2: 219213843-219213843 on Build GRCh38
rs532805022 Chr.2: 219213855-219213855 on Build GRCh38
rs781417749 Chr.2: 219214180-219214180 on Build GRCh38
rs964485285 Chr.2: 219213825-219213825 on Build GRCh38
rs990138760 Chr.2: 219214104-219214104 on Build GRCh38
rs778541482 Chr.2: 219214149-219214149 on Build GRCh38
rs201100181 Chr.2: 219214119-219214119 on Build GRCh38
rs775702828 Chr.2: 219213817-219213817 on Build GRCh38
rs749179479 Chr.2: 219214143-219214143 on Build GRCh38
rs144749234 Chr.2: 219214169-219214169 on Build GRCh38
rs768457293 Chr.2: 219214200-219214200 on Build GRCh38
rs900125722 Chr.2: 219214035-219214035 on Build GRCh38
rs763747092 Chr.2: 219213989-219213989 on Build GRCh38
rs754343480 Chr.2: 219214164-219214164 on Build GRCh38
rs148899972 Chr.2: 219214172-219214172 on Build GRCh38
rs370493038 Chr.2: 219213919-219213919 on Build GRCh38
rs368781340 Chr.2: 219214197-219214197 on Build GRCh38
rs747533202 Chr.2: 219213789-219213789 on Build GRCh38
rs1040298941 Chr.2: 219214025-219214025 on Build GRCh38
rs757993411 Chr.2: 219213779-219213779 on Build GRCh38
rs563880197 Chr.2: 219213844-219213844 on Build GRCh38
rs762741744 Chr.2: 219213890-219213890 on Build GRCh38
rs775551356 Chr.2: 219213983-219213984 on Build GRCh38
rs377028407 Chr.2: 219213870-219213870 on Build GRCh38
rs886609379 Chr.2: 219213975-219213975 on Build GRCh38
rs763768429 Chr.2: 219213893-219213893 on Build GRCh38
rs147445258 Chr.2: 219213930-219213930 on Build GRCh38
rs781458211 Chr.2: 219213800-219213800 on Build GRCh38
rs370349438 Chr.2: 219213883-219213883 on Build GRCh38
rs867934934 Chr.2: 219213876-219213876 on Build GRCh38
rs1047961287 Chr.2: 219213956-219213956 on Build GRCh38
rs749624340 Chr.2: 219213921-219213921 on Build GRCh38
rs771594650 Chr.2: 219214154-219214154 on Build GRCh38
rs749201224 Chr.2: 219213846-219213847 on Build GRCh38
rs769830815 Chr.2: 219213958-219213958 on Build GRCh38
rs760882591 Chr.2: 219214129-219214131 on Build GRCh38
rs201609106 Chr.2: 219213790-219213790 on Build GRCh38
rs202127374 Chr.2: 219214095-219214095 on Build GRCh38
rs368251947 Chr.2: 219213961-219213961 on Build GRCh38
rs569538868 Chr.2: 219214045-219214045 on Build GRCh38
rs140089441 Chr.2: 219213873-219213873 on Build GRCh38
rs771456013 Chr.2: 219213799-219213799 on Build GRCh38
rs574944957 Chr.2: 219214148-219214148 on Build GRCh38
rs920871888 Chr.2: 219213943-219213943 on Build GRCh38
rs538564058 Chr.2: 219214131-219214131 on Build GRCh38
rs2139823 Chr.2: 219213774-219213774 on Build GRCh38
rs887553295 Chr.2: 219214053-219214053 on Build GRCh38
rs367776479 Chr.2: 219213867-219213867 on Build GRCh38
rs549520020 Chr.2: 219214037-219214037 on Build GRCh38
rs143105148 Chr.2: 219213856-219213856 on Build GRCh38
rs1015137922 Chr.2: 219214060-219214060 on Build GRCh38
rs199839474 Chr.2: 219214075-219214075 on Build GRCh38
rs1042258296 Chr.2: 219213768-219213768 on Build GRCh38

View Full Product Details