Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219220060-219220569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG9A
Entrez Gene ID
79065
Gene Name
autophagy related 9A
Gene Aliases
APG9L1, MGD3208, mATG9
UniGene
Hs.323363
Gene Chromosome Location
Chr.2: 219219380-219229688 on Build GRCh38
Transcripts
NM_001077198.2 NM_024085.4 NR_104255.1

SNP Detail

SNP ID Chromosome Location
rs938898509 Chr.2: 219220526-219220526 on Build GRCh38
rs866369227 Chr.2: 219220131-219220131 on Build GRCh38
rs1005054272 Chr.2: 219220349-219220349 on Build GRCh38
rs979874783 Chr.2: 219220425-219220425 on Build GRCh38
rs1055991571 Chr.2: 219220530-219220530 on Build GRCh38
rs768835279 Chr.2: 219220475-219220475 on Build GRCh38
rs763824395 Chr.2: 219220413-219220413 on Build GRCh38
rs888067470 Chr.2: 219220342-219220342 on Build GRCh38
rs117189666 Chr.2: 219220387-219220387 on Build GRCh38
rs1039838251 Chr.2: 219220239-219220239 on Build GRCh38
rs756659338 Chr.2: 219220464-219220464 on Build GRCh38
rs960783502 Chr.2: 219220124-219220124 on Build GRCh38
rs767395834 Chr.2: 219220129-219220129 on Build GRCh38
rs529838997 Chr.2: 219220296-219220296 on Build GRCh38
rs978350704 Chr.2: 219220328-219220328 on Build GRCh38
rs537917341 Chr.2: 219220353-219220353 on Build GRCh38
rs200547303 Chr.2: 219220469-219220469 on Build GRCh38
rs3755047 Chr.2: 219220142-219220142 on Build GRCh38
rs1011636134 Chr.2: 219220302-219220302 on Build GRCh38
rs753991659 Chr.2: 219220254-219220254 on Build GRCh38
rs780669489 Chr.2: 219220466-219220466 on Build GRCh38
rs751110548 Chr.2: 219220418-219220418 on Build GRCh38
rs940883842 Chr.2: 219220222-219220222 on Build GRCh38
rs1013640016 Chr.2: 219220384-219220384 on Build GRCh38
rs1039527489 Chr.2: 219220518-219220518 on Build GRCh38
rs960830841 Chr.2: 219220370-219220370 on Build GRCh38
rs3755048 Chr.2: 219220180-219220180 on Build GRCh38
rs984544896 Chr.2: 219220388-219220388 on Build GRCh38
rs544946681 Chr.2: 219220188-219220188 on Build GRCh38
rs899563672 Chr.2: 219220243-219220243 on Build GRCh38
rs967891695 Chr.2: 219220320-219220320 on Build GRCh38
rs745992887 Chr.2: 219220496-219220496 on Build GRCh38
rs183165143 Chr.2: 219220369-219220369 on Build GRCh38
rs1022139985 Chr.2: 219220316-219220316 on Build GRCh38
rs921672417 Chr.2: 219220164-219220164 on Build GRCh38
rs372702555 Chr.2: 219220492-219220492 on Build GRCh38
rs756862988 Chr.2: 219220432-219220432 on Build GRCh38
rs547360316 Chr.2: 219220110-219220110 on Build GRCh38
rs956611044 Chr.2: 219220462-219220462 on Build GRCh38
rs907505790 Chr.2: 219220260-219220260 on Build GRCh38
rs374847915 Chr.2: 219220440-219220440 on Build GRCh38
rs549750187 Chr.2: 219220336-219220336 on Build GRCh38
rs746507601 Chr.2: 219220403-219220403 on Build GRCh38
rs757539098 Chr.2: 219220294-219220294 on Build GRCh38
rs746427852 Chr.2: 219220441-219220441 on Build GRCh38
rs1030919760 Chr.2: 219220351-219220351 on Build GRCh38
rs978004596 Chr.2: 219220148-219220148 on Build GRCh38
rs748305674 Chr.2: 219220482-219220482 on Build GRCh38
rs773700678 Chr.2: 219220482-219220483 on Build GRCh38
rs925304806 Chr.2: 219220456-219220456 on Build GRCh38
rs564002052 Chr.2: 219220169-219220169 on Build GRCh38
rs774701055 Chr.2: 219220481-219220481 on Build GRCh38
rs368646256 Chr.2: 219220471-219220471 on Build GRCh38
rs1034280210 Chr.2: 219220283-219220283 on Build GRCh38
rs943877388 Chr.2: 219220477-219220477 on Build GRCh38
rs12472988 Chr.2: 219220128-219220128 on Build GRCh38
rs979437891 Chr.2: 219220153-219220153 on Build GRCh38
rs147462116 Chr.2: 219220154-219220154 on Build GRCh38
rs187244984 Chr.2: 219220228-219220228 on Build GRCh38
rs975784594 Chr.2: 219220163-219220163 on Build GRCh38
rs370426449 Chr.2: 219220404-219220404 on Build GRCh38
rs190572698 Chr.2: 219220263-219220263 on Build GRCh38
rs944333396 Chr.2: 219220155-219220155 on Build GRCh38
rs1015045391 Chr.2: 219220119-219220119 on Build GRCh38
rs10799 Chr.2: 219220216-219220216 on Build GRCh38
rs567053473 Chr.2: 219220291-219220291 on Build GRCh38
rs921558788 Chr.2: 219220378-219220378 on Build GRCh38

View Full Product Details