Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219224017-219224512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG9A
Entrez Gene ID
79065
Gene Name
autophagy related 9A
Gene Aliases
APG9L1, MGD3208, mATG9
UniGene
Hs.323363
Gene Chromosome Location
Chr.2: 219219380-219229688 on Build GRCh38
Transcripts
NM_001077198.2 NM_024085.4 NR_104255.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6888420 c.1256C>T p.T419I Chr.2: 219224115-219224115 on Build GRCh38
5396263 c.1208C>T p.A403V Chr.2: 219224163-219224163 on Build GRCh38
6900340 c.1002C>T p.L334L Chr.2: 219224369-219224369 on Build GRCh38
6606752 c.1058G>A p.R353H Chr.2: 219224313-219224313 on Build GRCh38
3044651 c.866G>A p.R289H Chr.2: 219224505-219224505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754159145 Chr.2: 219224362-219224362 on Build GRCh38
rs757746159 Chr.2: 219224132-219224132 on Build GRCh38
rs531910156 Chr.2: 219224049-219224049 on Build GRCh38
rs749503489 Chr.2: 219224240-219224240 on Build GRCh38
rs369031181 Chr.2: 219224120-219224120 on Build GRCh38
rs779713032 Chr.2: 219224249-219224249 on Build GRCh38
rs753564253 Chr.2: 219224091-219224091 on Build GRCh38
rs766730899 Chr.2: 219224361-219224361 on Build GRCh38
rs768021337 Chr.2: 219224217-219224217 on Build GRCh38
rs767917505 Chr.2: 219224327-219224327 on Build GRCh38
rs931204688 Chr.2: 219224349-219224349 on Build GRCh38
rs745716494 Chr.2: 219224275-219224275 on Build GRCh38
rs773178345 Chr.2: 219224168-219224168 on Build GRCh38
rs760516947 Chr.2: 219224086-219224086 on Build GRCh38
rs992127535 Chr.2: 219224439-219224439 on Build GRCh38
rs886764360 Chr.2: 219224400-219224400 on Build GRCh38
rs779047144 Chr.2: 219224395-219224395 on Build GRCh38
rs1003891496 Chr.2: 219224197-219224197 on Build GRCh38
rs200537526 Chr.2: 219224441-219224441 on Build GRCh38
rs764800423 Chr.2: 219224181-219224181 on Build GRCh38
rs750939878 Chr.2: 219224339-219224339 on Build GRCh38
rs566492636 Chr.2: 219224345-219224345 on Build GRCh38
rs982172613 Chr.2: 219224056-219224056 on Build GRCh38
rs761352054 Chr.2: 219224326-219224326 on Build GRCh38
rs752153508 Chr.2: 219224204-219224204 on Build GRCh38
rs370223751 Chr.2: 219224350-219224350 on Build GRCh38
rs199726579 Chr.2: 219224090-219224090 on Build GRCh38
rs913912626 Chr.2: 219224041-219224041 on Build GRCh38
rs199914797 Chr.2: 219224261-219224261 on Build GRCh38
rs57664536 Chr.2: 219224464-219224464 on Build GRCh38
rs757871025 Chr.2: 219224212-219224212 on Build GRCh38
rs780268080 Chr.2: 219224153-219224153 on Build GRCh38
rs776309417 Chr.2: 219224176-219224176 on Build GRCh38
rs752160504 Chr.2: 219224114-219224114 on Build GRCh38
rs759315800 Chr.2: 219224177-219224177 on Build GRCh38
rs201218710 Chr.2: 219224250-219224250 on Build GRCh38
rs549042911 Chr.2: 219224485-219224485 on Build GRCh38
rs568908389 Chr.2: 219224486-219224486 on Build GRCh38
rs746205802 Chr.2: 219224140-219224140 on Build GRCh38
rs906317927 Chr.2: 219224442-219224442 on Build GRCh38
rs372960557 Chr.2: 219224123-219224123 on Build GRCh38
rs369827739 Chr.2: 219224229-219224229 on Build GRCh38
rs954840628 Chr.2: 219224355-219224355 on Build GRCh38
rs201871476 Chr.2: 219224075-219224075 on Build GRCh38
rs775464670 Chr.2: 219224043-219224043 on Build GRCh38
rs376440161 Chr.2: 219224221-219224221 on Build GRCh38
rs200848767 Chr.2: 219224113-219224113 on Build GRCh38
rs749336185 Chr.2: 219224156-219224156 on Build GRCh38
rs766136368 Chr.2: 219224087-219224087 on Build GRCh38
rs1024710617 Chr.2: 219224209-219224209 on Build GRCh38
rs769617716 Chr.2: 219224278-219224278 on Build GRCh38
rs778267948 Chr.2: 219224076-219224076 on Build GRCh38
rs994614002 Chr.2: 219224291-219224291 on Build GRCh38
rs770811574 Chr.2: 219224171-219224171 on Build GRCh38
rs763041796 Chr.2: 219224046-219224046 on Build GRCh38
rs754274617 Chr.2: 219224095-219224095 on Build GRCh38
rs866013931 Chr.2: 219224379-219224379 on Build GRCh38
rs376174649 Chr.2: 219224258-219224258 on Build GRCh38
rs755211778 Chr.2: 219224382-219224382 on Build GRCh38
rs368643354 Chr.2: 219224407-219224407 on Build GRCh38
rs570518317 Chr.2: 219224045-219224045 on Build GRCh38
rs375131249 Chr.2: 219224134-219224134 on Build GRCh38
rs17855976 Chr.2: 219224230-219224230 on Build GRCh38
rs752389669 Chr.2: 219224191-219224191 on Build GRCh38
rs989733874 Chr.2: 219224246-219224246 on Build GRCh38
rs775055046 Chr.2: 219224290-219224290 on Build GRCh38
rs564912563 Chr.2: 219224427-219224427 on Build GRCh38
rs757913888 Chr.2: 219224119-219224119 on Build GRCh38
rs769934429 Chr.2: 219224141-219224141 on Build GRCh38
rs781382230 Chr.2: 219224135-219224135 on Build GRCh38
rs759078193 Chr.2: 219224419-219224419 on Build GRCh38
rs762204626 Chr.2: 219224165-219224165 on Build GRCh38
rs535141552 Chr.2: 219224473-219224473 on Build GRCh38
rs774022899 Chr.2: 219224318-219224318 on Build GRCh38
rs374338738 Chr.2: 219224396-219224396 on Build GRCh38
rs761300429 Chr.2: 219224487-219224487 on Build GRCh38
rs763675302 Chr.2: 219224200-219224200 on Build GRCh38
rs377110782 Chr.2: 219224306-219224306 on Build GRCh38
rs1026050748 Chr.2: 219224307-219224307 on Build GRCh38
rs769517837 Chr.2: 219224429-219224429 on Build GRCh38
rs756437040 Chr.2: 219224222-219224222 on Build GRCh38
rs374035695 Chr.2: 219224286-219224286 on Build GRCh38
rs947564468 Chr.2: 219224489-219224489 on Build GRCh38
rs746660377 Chr.2: 219224476-219224476 on Build GRCh38
rs570750110 Chr.2: 219224304-219224304 on Build GRCh38
rs762609003 Chr.2: 219224192-219224192 on Build GRCh38

View Full Product Details