Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219282947-219283440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB2
Entrez Gene ID
3300
Gene Name
DnaJ heat shock protein family (Hsp40) member B2
Gene Aliases
CMT2T, DSMA5, HSJ-1, HSJ1, HSPF3
UniGene
Hs.77768
Gene Chromosome Location
Chr.2: 219279318-219286900 on Build GRCh38
Transcripts
NM_001039550.1 NM_006736.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7248498 c.446-8G>A p.? Chr.2: 219283125-219283125 on Build GRCh38
4841686 c.458C>A p.S153* Chr.2: 219283145-219283145 on Build GRCh38
5564753 c.530G>A p.R177H Chr.2: 219283217-219283217 on Build GRCh38
350571 c.533G>A p.R178H Chr.2: 219283220-219283220 on Build GRCh38
4583214 c.548G>A p.R183K Chr.2: 219283235-219283235 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775571150 Chr.2: 219283333-219283333 on Build GRCh38
rs757055788 Chr.2: 219283232-219283232 on Build GRCh38
rs896870289 Chr.2: 219283164-219283165 on Build GRCh38
rs541164513 Chr.2: 219283088-219283088 on Build GRCh38
rs1007135501 Chr.2: 219283366-219283366 on Build GRCh38
rs1006852724 Chr.2: 219283309-219283309 on Build GRCh38
rs755622620 Chr.2: 219283156-219283156 on Build GRCh38
rs140419734 Chr.2: 219283087-219283089 on Build GRCh38
rs998117764 Chr.2: 219283321-219283321 on Build GRCh38
rs1010859064 Chr.2: 219283037-219283037 on Build GRCh38
rs528627093 Chr.2: 219282968-219282968 on Build GRCh38
rs1016652527 Chr.2: 219283003-219283003 on Build GRCh38
rs780605624 Chr.2: 219283160-219283160 on Build GRCh38
rs377437714 Chr.2: 219283210-219283210 on Build GRCh38
rs763352607 Chr.2: 219283089-219283089 on Build GRCh38
rs902489268 Chr.2: 219283301-219283301 on Build GRCh38
rs576761656 Chr.2: 219283278-219283278 on Build GRCh38
rs376532188 Chr.2: 219283214-219283214 on Build GRCh38
rs769095774 Chr.2: 219283181-219283181 on Build GRCh38
rs3821038 Chr.2: 219283045-219283045 on Build GRCh38
rs774704720 Chr.2: 219283193-219283193 on Build GRCh38
rs558617819 Chr.2: 219283082-219283082 on Build GRCh38
rs549126522 Chr.2: 219283108-219283108 on Build GRCh38
rs369758068 Chr.2: 219283235-219283235 on Build GRCh38
rs780569796 Chr.2: 219282971-219282971 on Build GRCh38
rs946747306 Chr.2: 219283254-219283254 on Build GRCh38
rs2276639 Chr.2: 219283010-219283010 on Build GRCh38
rs767736834 Chr.2: 219283110-219283110 on Build GRCh38
rs563221082 Chr.2: 219283206-219283206 on Build GRCh38
rs761466124 Chr.2: 219283104-219283106 on Build GRCh38
rs769004015 Chr.2: 219283092-219283092 on Build GRCh38
rs776388749 Chr.2: 219283096-219283097 on Build GRCh38
rs142128824 Chr.2: 219283344-219283344 on Build GRCh38
rs373945499 Chr.2: 219283390-219283390 on Build GRCh38
rs889391320 Chr.2: 219283269-219283269 on Build GRCh38
rs748494804 Chr.2: 219283194-219283194 on Build GRCh38
rs777721164 Chr.2: 219283142-219283142 on Build GRCh38
rs377762531 Chr.2: 219283051-219283051 on Build GRCh38
rs201363015 Chr.2: 219283178-219283178 on Build GRCh38
rs376263765 Chr.2: 219283356-219283356 on Build GRCh38
rs764080481 Chr.2: 219283219-219283219 on Build GRCh38
rs780852315 Chr.2: 219283145-219283145 on Build GRCh38
rs1055176141 Chr.2: 219283231-219283231 on Build GRCh38
rs1035809428 Chr.2: 219283367-219283367 on Build GRCh38
rs762985004 Chr.2: 219283217-219283217 on Build GRCh38
rs569644807 Chr.2: 219283063-219283063 on Build GRCh38
rs750489959 Chr.2: 219283112-219283112 on Build GRCh38
rs1029960371 Chr.2: 219283340-219283340 on Build GRCh38
rs544935330 Chr.2: 219283180-219283180 on Build GRCh38
rs572034536 Chr.2: 219283083-219283083 on Build GRCh38
rs3821039 Chr.2: 219283125-219283125 on Build GRCh38
rs993546807 Chr.2: 219283039-219283039 on Build GRCh38
rs771207537 Chr.2: 219283403-219283403 on Build GRCh38
rs375051736 Chr.2: 219283124-219283124 on Build GRCh38
rs548673998 Chr.2: 219283415-219283415 on Build GRCh38
rs774614720 Chr.2: 219283106-219283106 on Build GRCh38
rs755679147 Chr.2: 219283279-219283279 on Build GRCh38
rs985236791 Chr.2: 219283079-219283079 on Build GRCh38
rs372032059 Chr.2: 219283199-219283199 on Build GRCh38
rs960567752 Chr.2: 219283050-219283050 on Build GRCh38
rs773359849 Chr.2: 219283397-219283397 on Build GRCh38
rs766549564 Chr.2: 219283209-219283209 on Build GRCh38
rs953975232 Chr.2: 219283072-219283072 on Build GRCh38
rs765110238 Chr.2: 219283112-219283112 on Build GRCh38
rs559746196 Chr.2: 219283345-219283345 on Build GRCh38
rs202123709 Chr.2: 219283220-219283220 on Build GRCh38
rs756892919 Chr.2: 219283143-219283143 on Build GRCh38
rs758322672 Chr.2: 219283131-219283131 on Build GRCh38
rs781621441 Chr.2: 219283300-219283300 on Build GRCh38
rs183788853 Chr.2: 219283117-219283117 on Build GRCh38
rs1055584387 Chr.2: 219282994-219282994 on Build GRCh38
rs550057743 Chr.2: 219283020-219283020 on Build GRCh38
rs538614292 Chr.2: 219283064-219283064 on Build GRCh38
rs748730652 Chr.2: 219283282-219283282 on Build GRCh38
rs371203834 Chr.2: 219283378-219283378 on Build GRCh38
rs938110004 Chr.2: 219282983-219282983 on Build GRCh38
rs148642155 Chr.2: 219283041-219283041 on Build GRCh38
rs772479925 Chr.2: 219283391-219283391 on Build GRCh38
rs893873016 Chr.2: 219283249-219283249 on Build GRCh38
rs773452073 Chr.2: 219283203-219283203 on Build GRCh38
rs1042383298 Chr.2: 219283259-219283259 on Build GRCh38
rs542308257 Chr.2: 219283410-219283410 on Build GRCh38
rs745422775 Chr.2: 219283146-219283146 on Build GRCh38
rs779554070 Chr.2: 219283281-219283281 on Build GRCh38
rs750259439 Chr.2: 219283383-219283384 on Build GRCh38
rs760786001 Chr.2: 219283116-219283116 on Build GRCh38
rs866864354 Chr.2: 219283000-219283000 on Build GRCh38

View Full Product Details