Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219283178-219283672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB2
Entrez Gene ID
3300
Gene Name
DnaJ heat shock protein family (Hsp40) member B2
Gene Aliases
CMT2T, DSMA5, HSJ-1, HSJ1, HSPF3
UniGene
Hs.77768
Gene Chromosome Location
Chr.2: 219279318-219286900 on Build GRCh38
Transcripts
NM_001039550.1 NM_006736.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6210485 c.544C>T p.R182C Chr.2: 219283231-219283231 on Build GRCh38
5564753 c.530G>A p.R177H Chr.2: 219283217-219283217 on Build GRCh38
4972869 c.571C>T p.R191W Chr.2: 219283441-219283441 on Build GRCh38
6251030 c.533G>T p.R178L Chr.2: 219283220-219283220 on Build GRCh38
6647113 c.619+2T>C p.? Chr.2: 219283491-219283491 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757055788 Chr.2: 219283232-219283232 on Build GRCh38
rs775571150 Chr.2: 219283333-219283333 on Build GRCh38
rs755769067 Chr.2: 219283464-219283464 on Build GRCh38
rs534379119 Chr.2: 219283641-219283641 on Build GRCh38
rs1007135501 Chr.2: 219283366-219283366 on Build GRCh38
rs565672776 Chr.2: 219283636-219283636 on Build GRCh38
rs987760829 Chr.2: 219283560-219283560 on Build GRCh38
rs750128603 Chr.2: 219283457-219283457 on Build GRCh38
rs1006852724 Chr.2: 219283309-219283309 on Build GRCh38
rs781544652 Chr.2: 219283530-219283530 on Build GRCh38
rs998117764 Chr.2: 219283321-219283321 on Build GRCh38
rs776893562 Chr.2: 219283441-219283441 on Build GRCh38
rs376172884 Chr.2: 219283543-219283543 on Build GRCh38
rs377437714 Chr.2: 219283210-219283210 on Build GRCh38
rs1025315551 Chr.2: 219283509-219283509 on Build GRCh38
rs990934672 Chr.2: 219283564-219283564 on Build GRCh38
rs902489268 Chr.2: 219283301-219283301 on Build GRCh38
rs576761656 Chr.2: 219283278-219283278 on Build GRCh38
rs141066032 Chr.2: 219283604-219283604 on Build GRCh38
rs376532188 Chr.2: 219283214-219283214 on Build GRCh38
rs369758068 Chr.2: 219283235-219283235 on Build GRCh38
rs946747306 Chr.2: 219283254-219283254 on Build GRCh38
rs151165500 Chr.2: 219283550-219283550 on Build GRCh38
rs761985475 Chr.2: 219283456-219283456 on Build GRCh38
rs563221082 Chr.2: 219283206-219283206 on Build GRCh38
rs764178720 Chr.2: 219283432-219283432 on Build GRCh38
rs142128824 Chr.2: 219283344-219283344 on Build GRCh38
rs373936498 Chr.2: 219283540-219283540 on Build GRCh38
rs889391320 Chr.2: 219283269-219283269 on Build GRCh38
rs373945499 Chr.2: 219283390-219283390 on Build GRCh38
rs143245353 Chr.2: 219283431-219283431 on Build GRCh38
rs368011088 Chr.2: 219283581-219283584 on Build GRCh38
rs376137700 Chr.2: 219283458-219283458 on Build GRCh38
rs764080481 Chr.2: 219283219-219283219 on Build GRCh38
rs376263765 Chr.2: 219283356-219283356 on Build GRCh38
rs1055176141 Chr.2: 219283231-219283231 on Build GRCh38
rs1035809428 Chr.2: 219283367-219283367 on Build GRCh38
rs762985004 Chr.2: 219283217-219283217 on Build GRCh38
rs1029960371 Chr.2: 219283340-219283340 on Build GRCh38
rs765978583 Chr.2: 219283471-219283471 on Build GRCh38
rs910676731 Chr.2: 219283632-219283632 on Build GRCh38
rs765477783 Chr.2: 219283640-219283640 on Build GRCh38
rs771207537 Chr.2: 219283403-219283403 on Build GRCh38
rs933046075 Chr.2: 219283580-219283583 on Build GRCh38
rs762959655 Chr.2: 219283473-219283475 on Build GRCh38
rs754451895 Chr.2: 219283498-219283498 on Build GRCh38
rs548673998 Chr.2: 219283415-219283415 on Build GRCh38
rs767330115 Chr.2: 219283452-219283452 on Build GRCh38
rs113710471 Chr.2: 219283539-219283539 on Build GRCh38
rs755679147 Chr.2: 219283279-219283279 on Build GRCh38
rs771299169 Chr.2: 219283527-219283527 on Build GRCh38
rs946820850 Chr.2: 219283638-219283638 on Build GRCh38
rs773359849 Chr.2: 219283397-219283397 on Build GRCh38
rs923974092 Chr.2: 219283647-219283647 on Build GRCh38
rs766549564 Chr.2: 219283209-219283209 on Build GRCh38
rs199574391 Chr.2: 219283538-219283538 on Build GRCh38
rs779354826 Chr.2: 219283508-219283508 on Build GRCh38
rs559746196 Chr.2: 219283345-219283345 on Build GRCh38
rs202123709 Chr.2: 219283220-219283220 on Build GRCh38
rs781621441 Chr.2: 219283300-219283300 on Build GRCh38
rs753392588 Chr.2: 219283497-219283497 on Build GRCh38
rs748730652 Chr.2: 219283282-219283282 on Build GRCh38
rs371203834 Chr.2: 219283378-219283378 on Build GRCh38
rs753234090 Chr.2: 219283511-219283511 on Build GRCh38
rs772479925 Chr.2: 219283391-219283391 on Build GRCh38
rs773452073 Chr.2: 219283203-219283203 on Build GRCh38
rs893873016 Chr.2: 219283249-219283249 on Build GRCh38
rs1042383298 Chr.2: 219283259-219283259 on Build GRCh38
rs73079148 Chr.2: 219283558-219283558 on Build GRCh38
rs368989666 Chr.2: 219283514-219283514 on Build GRCh38
rs747335335 Chr.2: 219283524-219283524 on Build GRCh38
rs748401831 Chr.2: 219283597-219283597 on Build GRCh38
rs542308257 Chr.2: 219283410-219283410 on Build GRCh38
rs779554070 Chr.2: 219283281-219283281 on Build GRCh38
rs758770471 Chr.2: 219283513-219283513 on Build GRCh38
rs750259439 Chr.2: 219283383-219283384 on Build GRCh38

View Full Product Details