Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219501751-219502255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMPPA
Entrez Gene ID
29926
Gene Name
GDP-mannose pyrophosphorylase A
Gene Aliases
AAMR
UniGene
Hs.27059
Gene Chromosome Location
Chr.2: 219498865-219506996 on Build GRCh38
Transcripts
NM_013335.3 NM_205847.2 XM_005246483.3 XM_005246486.3 XM_011511032.2 XM_017003906.1 XM_017003907.1 XM_017003908.1 XM_017003909.1 XM_017003910.1 XM_017003911.1 XM_017003912.1 XM_017003913.1 XM_017003914.1 XM_017003915.1 XM_017003916.1 XM_017003917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1016820 c.148G>A p.D50N Chr.2: 219501966-219501966 on Build GRCh38
289446 c.255A>C p.E85D Chr.2: 219501863-219501863 on Build GRCh38
244720 c.296G>T p.R99L Chr.2: 219501904-219501904 on Build GRCh38
5587882 c.197C>T p.P66L Chr.2: 219502015-219502015 on Build GRCh38
7273150 c.337C>T p.L113F Chr.2: 219501945-219501945 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376536579 Chr.2: 219502060-219502060 on Build GRCh38
rs779526489 Chr.2: 219501908-219501908 on Build GRCh38
rs948487637 Chr.2: 219502139-219502139 on Build GRCh38
rs751612064 Chr.2: 219501835-219501835 on Build GRCh38
rs759418965 Chr.2: 219501942-219501942 on Build GRCh38
rs368690353 Chr.2: 219502015-219502015 on Build GRCh38
rs749864028 Chr.2: 219501779-219501779 on Build GRCh38
rs186735505 Chr.2: 219501974-219501974 on Build GRCh38
rs777279135 Chr.2: 219501916-219501916 on Build GRCh38
rs771681259 Chr.2: 219501913-219501913 on Build GRCh38
rs567070877 Chr.2: 219501778-219501778 on Build GRCh38
rs547259164 Chr.2: 219501777-219501777 on Build GRCh38
rs115573631 Chr.2: 219502151-219502151 on Build GRCh38
rs529574707 Chr.2: 219501971-219501971 on Build GRCh38
rs763544554 Chr.2: 219502081-219502081 on Build GRCh38
rs998755895 Chr.2: 219501927-219501927 on Build GRCh38
rs894186395 Chr.2: 219501773-219501773 on Build GRCh38
rs2385539 Chr.2: 219502231-219502231 on Build GRCh38
rs776442349 Chr.2: 219501935-219501935 on Build GRCh38
rs771975056 Chr.2: 219502039-219502039 on Build GRCh38
rs750729833 Chr.2: 219501776-219501776 on Build GRCh38
rs770772263 Chr.2: 219501813-219501813 on Build GRCh38
rs570253568 Chr.2: 219502047-219502047 on Build GRCh38
rs528048995 Chr.2: 219502092-219502092 on Build GRCh38
rs375606944 Chr.2: 219501966-219501966 on Build GRCh38
rs747582518 Chr.2: 219501909-219501909 on Build GRCh38
rs749257889 Chr.2: 219502037-219502037 on Build GRCh38
rs568022094 Chr.2: 219502000-219502000 on Build GRCh38
rs759474217 Chr.2: 219501828-219501828 on Build GRCh38
rs539246802 Chr.2: 219502070-219502070 on Build GRCh38
rs754236303 Chr.2: 219501982-219501982 on Build GRCh38
rs112026034 Chr.2: 219502102-219502102 on Build GRCh38
rs141667745 Chr.2: 219502014-219502014 on Build GRCh38
rs761791232 Chr.2: 219501953-219501953 on Build GRCh38
rs751046456 Chr.2: 219502084-219502084 on Build GRCh38
rs746678477 Chr.2: 219501929-219501929 on Build GRCh38
rs746623283 Chr.2: 219502006-219502006 on Build GRCh38
rs1010281716 Chr.2: 219502133-219502135 on Build GRCh38
rs184836683 Chr.2: 219502158-219502158 on Build GRCh38
rs768219011 Chr.2: 219501782-219501782 on Build GRCh38
rs777343237 Chr.2: 219502004-219502004 on Build GRCh38
rs767718283 Chr.2: 219501858-219501858 on Build GRCh38
rs756530418 Chr.2: 219501855-219501855 on Build GRCh38
rs766680312 Chr.2: 219502046-219502046 on Build GRCh38
rs757789454 Chr.2: 219502003-219502003 on Build GRCh38
rs747791144 Chr.2: 219501785-219501785 on Build GRCh38
rs1021431511 Chr.2: 219501894-219501894 on Build GRCh38
rs945250817 Chr.2: 219501873-219501873 on Build GRCh38
rs753112469 Chr.2: 219501888-219501888 on Build GRCh38
rs770645993 Chr.2: 219501930-219501930 on Build GRCh38
rs755437376 Chr.2: 219501994-219501994 on Build GRCh38
rs894147817 Chr.2: 219502126-219502126 on Build GRCh38
rs397518460 Chr.2: 219501903-219501903 on Build GRCh38
rs769695080 Chr.2: 219502034-219502034 on Build GRCh38
rs542078432 Chr.2: 219502122-219502122 on Build GRCh38
rs773451252 Chr.2: 219502042-219502042 on Build GRCh38
rs760890027 Chr.2: 219502045-219502045 on Build GRCh38
rs372560013 Chr.2: 219501944-219501944 on Build GRCh38
rs34528570 Chr.2: 219501881-219501882 on Build GRCh38
rs544318789 Chr.2: 219502201-219502201 on Build GRCh38
rs747136634 Chr.2: 219501962-219501962 on Build GRCh38
rs756925771 Chr.2: 219502008-219502008 on Build GRCh38
rs765650079 Chr.2: 219502053-219502053 on Build GRCh38
rs372061594 Chr.2: 219502061-219502061 on Build GRCh38
rs1018038018 Chr.2: 219501968-219501968 on Build GRCh38
rs756799215 Chr.2: 219502085-219502085 on Build GRCh38
rs776646514 Chr.2: 219502148-219502148 on Build GRCh38
rs750721385 Chr.2: 219501838-219501838 on Build GRCh38
rs564566105 Chr.2: 219502206-219502206 on Build GRCh38
rs766821786 Chr.2: 219502014-219502017 on Build GRCh38
rs775535431 Chr.2: 219501951-219501951 on Build GRCh38
rs772959218 Chr.2: 219501796-219501796 on Build GRCh38
rs778321872 Chr.2: 219501783-219501783 on Build GRCh38
rs368310157 Chr.2: 219501827-219501827 on Build GRCh38
rs780869528 Chr.2: 219502009-219502009 on Build GRCh38
rs758976144 Chr.2: 219502080-219502080 on Build GRCh38
rs372781760 Chr.2: 219502036-219502036 on Build GRCh38
rs866231169 Chr.2: 219502202-219502202 on Build GRCh38
rs1056776743 Chr.2: 219502106-219502106 on Build GRCh38
rs565848757 Chr.2: 219502002-219502002 on Build GRCh38
rs764193356 Chr.2: 219501834-219501834 on Build GRCh38
rs771604721 Chr.2: 219501789-219501789 on Build GRCh38
rs1047394626 Chr.2: 219502030-219502030 on Build GRCh38
rs929514344 Chr.2: 219502031-219502031 on Build GRCh38
rs867965865 Chr.2: 219501868-219501868 on Build GRCh38
rs760459634 Chr.2: 219501798-219501798 on Build GRCh38
rs562659059 Chr.2: 219501965-219501965 on Build GRCh38
rs759808279 Chr.2: 219502050-219502050 on Build GRCh38
rs571360759 Chr.2: 219502197-219502197 on Build GRCh38
rs779909339 Chr.2: 219501949-219501949 on Build GRCh38

View Full Product Details