Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219505317-219505812 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMPPA
Entrez Gene ID
29926
Gene Name
GDP-mannose pyrophosphorylase A
Gene Aliases
AAMR
UniGene
Hs.27059
Gene Chromosome Location
Chr.2: 219498865-219506996 on Build GRCh38
Transcripts
NM_013335.3 NM_205847.2 XM_005246483.3 XM_005246486.3 XM_011511032.2 XM_017003906.1 XM_017003907.1 XM_017003908.1 XM_017003909.1 XM_017003910.1 XM_017003911.1 XM_017003912.1 XM_017003913.1 XM_017003914.1 XM_017003915.1 XM_017003916.1 XM_017003917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6672041 c.867C>G p.I289M Chr.2: 219505728-219505728 on Build GRCh38
1285186 c.834_835insG p.W282fs*34 Chr.2: 219505536-219505537 on Build GRCh38
5895573 c.845G>A p.W282* Chr.2: 219505547-219505547 on Build GRCh38
1208380 c.863A>G p.Y288C Chr.2: 219505724-219505724 on Build GRCh38
6543970 c.728G>T p.G243V Chr.2: 219505335-219505335 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765252395 Chr.2: 219505396-219505396 on Build GRCh38
rs918927687 Chr.2: 219505425-219505425 on Build GRCh38
rs769745264 Chr.2: 219505734-219505734 on Build GRCh38
rs763135203 Chr.2: 219505435-219505435 on Build GRCh38
rs999512407 Chr.2: 219505543-219505543 on Build GRCh38
rs768769277 Chr.2: 219505401-219505403 on Build GRCh38
rs776643066 Chr.2: 219505699-219505699 on Build GRCh38
rs367640773 Chr.2: 219505357-219505357 on Build GRCh38
rs146215853 Chr.2: 219505470-219505470 on Build GRCh38
rs903900192 Chr.2: 219505772-219505772 on Build GRCh38
rs771870172 Chr.2: 219505373-219505373 on Build GRCh38
rs750790061 Chr.2: 219505530-219505530 on Build GRCh38
rs754262824 Chr.2: 219505662-219505662 on Build GRCh38
rs371497371 Chr.2: 219505360-219505360 on Build GRCh38
rs369043140 Chr.2: 219505573-219505573 on Build GRCh38
rs777036634 Chr.2: 219505759-219505759 on Build GRCh38
rs137863363 Chr.2: 219505477-219505477 on Build GRCh38
rs955950736 Chr.2: 219505586-219505586 on Build GRCh38
rs1005620276 Chr.2: 219505640-219505640 on Build GRCh38
rs751576633 Chr.2: 219505611-219505611 on Build GRCh38
rs754381367 Chr.2: 219505553-219505553 on Build GRCh38
rs778520832 Chr.2: 219505565-219505565 on Build GRCh38
rs61304293 Chr.2: 219505692-219505692 on Build GRCh38
rs139675922 Chr.2: 219505528-219505528 on Build GRCh38
rs200425681 Chr.2: 219505523-219505523 on Build GRCh38
rs554955899 Chr.2: 219505768-219505768 on Build GRCh38
rs762393620 Chr.2: 219505785-219505785 on Build GRCh38
rs752752098 Chr.2: 219505410-219505410 on Build GRCh38
rs369505327 Chr.2: 219505518-219505518 on Build GRCh38
rs139612850 Chr.2: 219505488-219505488 on Build GRCh38
rs746676212 Chr.2: 219505705-219505705 on Build GRCh38
rs147396523 Chr.2: 219505517-219505517 on Build GRCh38
rs776699056 Chr.2: 219505515-219505515 on Build GRCh38
rs972393344 Chr.2: 219505694-219505694 on Build GRCh38
rs768577936 Chr.2: 219505617-219505617 on Build GRCh38
rs755535192 Chr.2: 219505672-219505672 on Build GRCh38
rs781071244 Chr.2: 219505725-219505725 on Build GRCh38
rs762564864 Chr.2: 219505400-219505400 on Build GRCh38
rs773071891 Chr.2: 219505380-219505380 on Build GRCh38
rs757504803 Chr.2: 219505453-219505453 on Build GRCh38
rs569722184 Chr.2: 219505624-219505624 on Build GRCh38
rs762157391 Chr.2: 219505525-219505525 on Build GRCh38
rs79353804 Chr.2: 219505474-219505474 on Build GRCh38
rs953842015 Chr.2: 219505612-219505612 on Build GRCh38
rs762029038 Chr.2: 219505626-219505626 on Build GRCh38
rs776373574 Chr.2: 219505610-219505610 on Build GRCh38
rs961563489 Chr.2: 219505665-219505665 on Build GRCh38
rs746804961 Chr.2: 219505591-219505591 on Build GRCh38
rs372628288 Chr.2: 219505492-219505492 on Build GRCh38
rs550099366 Chr.2: 219505587-219505587 on Build GRCh38
rs368738474 Chr.2: 219505448-219505448 on Build GRCh38
rs200875359 Chr.2: 219505358-219505358 on Build GRCh38
rs761066072 Chr.2: 219505650-219505650 on Build GRCh38
rs567590952 Chr.2: 219505504-219505504 on Build GRCh38
rs371527019 Chr.2: 219505737-219505737 on Build GRCh38
rs766774052 Chr.2: 219505657-219505657 on Build GRCh38
rs968115351 Chr.2: 219505540-219505540 on Build GRCh38
rs372564894 Chr.2: 219505606-219505606 on Build GRCh38
rs781693209 Chr.2: 219505465-219505465 on Build GRCh38
rs1017038834 Chr.2: 219505653-219505653 on Build GRCh38
rs145760617 Chr.2: 219505719-219505719 on Build GRCh38
rs151043830 Chr.2: 219505738-219505738 on Build GRCh38
rs778641701 Chr.2: 219505382-219505382 on Build GRCh38
rs770772480 Chr.2: 219505608-219505608 on Build GRCh38
rs773633201 Chr.2: 219505632-219505632 on Build GRCh38
rs575126365 Chr.2: 219505781-219505781 on Build GRCh38
rs778374979 Chr.2: 219505485-219505485 on Build GRCh38
rs751864983 Chr.2: 219505449-219505449 on Build GRCh38
rs756583765 Chr.2: 219505538-219505538 on Build GRCh38
rs773014626 Chr.2: 219505503-219505503 on Build GRCh38
rs868076850 Chr.2: 219505497-219505497 on Build GRCh38
rs752221989 Chr.2: 219505673-219505673 on Build GRCh38
rs768272917 Chr.2: 219505478-219505478 on Build GRCh38
rs371770464 Chr.2: 219505545-219505545 on Build GRCh38
rs766985965 Chr.2: 219505539-219505539 on Build GRCh38
rs201361535 Chr.2: 219505493-219505493 on Build GRCh38
rs1042729590 Chr.2: 219505521-219505521 on Build GRCh38
rs761012765 Chr.2: 219505754-219505754 on Build GRCh38
rs370776331 Chr.2: 219505511-219505511 on Build GRCh38
rs745743360 Chr.2: 219505733-219505733 on Build GRCh38
rs773504203 Chr.2: 219505750-219505750 on Build GRCh38
rs771155229 Chr.2: 219505755-219505755 on Build GRCh38
rs772333659 Chr.2: 219505742-219505742 on Build GRCh38
rs759858230 Chr.2: 219505765-219505765 on Build GRCh38
rs757972361 Chr.2: 219505677-219505677 on Build GRCh38
rs756708119 Chr.2: 219505471-219505471 on Build GRCh38
rs758025482 Chr.2: 219505575-219505575 on Build GRCh38
rs375575519 Chr.2: 219505509-219505509 on Build GRCh38
rs374903239 Chr.2: 219505552-219505552 on Build GRCh38

View Full Product Details